Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184178660-184179257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2M1
Entrez Gene ID
1173
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
AP50, CLAPM1, mu2
UniGene
Hs.518460
Gene Chromosome Location
Chr.3: 184174846-184184091 on Build GRCh38
Transcripts
NM_001025205.1 NM_001311198.1 NM_004068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601627 c.82G>A p.A28T Chr.3: 184178864-184178864 on Build GRCh38
1421283 c.139G>A p.V47I Chr.3: 184178921-184178921 on Build GRCh38
6601629 c.231G>T p.M77I Chr.3: 184179013-184179013 on Build GRCh38
5595678 c.243C>T p.F81F Chr.3: 184179025-184179025 on Build GRCh38
5637114 c.172G>A p.V58I Chr.3: 184178954-184178954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774420255 Chr.3: 184178811-184178811 on Build GRCh38
rs920508005 Chr.3: 184178687-184178687 on Build GRCh38
rs373489563 Chr.3: 184178848-184178848 on Build GRCh38
rs772071290 Chr.3: 184179001-184179001 on Build GRCh38
rs747675284 Chr.3: 184178911-184178911 on Build GRCh38
rs756456772 Chr.3: 184179205-184179205 on Build GRCh38
rs914732362 Chr.3: 184178852-184178852 on Build GRCh38
rs929246153 Chr.3: 184178690-184178690 on Build GRCh38
rs774293277 Chr.3: 184178954-184178954 on Build GRCh38
rs527670858 Chr.3: 184179137-184179137 on Build GRCh38
rs768273737 Chr.3: 184178694-184178694 on Build GRCh38
rs1054162515 Chr.3: 184178839-184178839 on Build GRCh38
rs1019628464 Chr.3: 184179214-184179214 on Build GRCh38
rs763162356 Chr.3: 184179073-184179073 on Build GRCh38
rs1041873980 Chr.3: 184178914-184178914 on Build GRCh38
rs750708723 Chr.3: 184178841-184178841 on Build GRCh38
rs556633717 Chr.3: 184178863-184178863 on Build GRCh38
rs1025759162 Chr.3: 184179159-184179159 on Build GRCh38
rs757002374 Chr.3: 184179150-184179150 on Build GRCh38
rs112256941 Chr.3: 184178855-184178855 on Build GRCh38
rs1966081 Chr.3: 184178681-184178681 on Build GRCh38
rs555780261 Chr.3: 184178837-184178837 on Build GRCh38
rs111539857 Chr.3: 184179123-184179123 on Build GRCh38
rs1008270810 Chr.3: 184179180-184179180 on Build GRCh38
rs764013574 Chr.3: 184178819-184178819 on Build GRCh38
rs368164763 Chr.3: 184179143-184179143 on Build GRCh38
rs199554477 Chr.3: 184179162-184179162 on Build GRCh38
rs995446453 Chr.3: 184179044-184179044 on Build GRCh38
rs771827784 Chr.3: 184179154-184179154 on Build GRCh38
rs1044542584 Chr.3: 184178804-184178804 on Build GRCh38
rs917096186 Chr.3: 184178719-184178719 on Build GRCh38
rs751993161 Chr.3: 184178851-184178851 on Build GRCh38
rs770921099 Chr.3: 184178809-184178809 on Build GRCh38
rs777442995 Chr.3: 184179144-184179144 on Build GRCh38
rs535484590 Chr.3: 184178736-184178737 on Build GRCh38
rs2231216 Chr.3: 184178853-184178853 on Build GRCh38
rs770792578 Chr.3: 184178953-184178953 on Build GRCh38
rs764872535 Chr.3: 184178905-184178906 on Build GRCh38
rs773307248 Chr.3: 184179172-184179172 on Build GRCh38
rs749062778 Chr.3: 184178807-184178807 on Build GRCh38
rs755622712 Chr.3: 184178920-184178920 on Build GRCh38
rs148249373 Chr.3: 184178926-184178926 on Build GRCh38
rs796948561 Chr.3: 184179179-184179179 on Build GRCh38
rs576897074 Chr.3: 184178847-184178847 on Build GRCh38
rs562501180 Chr.3: 184178998-184178998 on Build GRCh38
rs1010505410 Chr.3: 184178864-184178864 on Build GRCh38
rs11539599 Chr.3: 184179018-184179018 on Build GRCh38
rs375760613 Chr.3: 184179133-184179133 on Build GRCh38
rs374693436 Chr.3: 184179169-184179169 on Build GRCh38
rs776805348 Chr.3: 184178832-184178832 on Build GRCh38
rs761946127 Chr.3: 184179005-184179005 on Build GRCh38
rs780025816 Chr.3: 184179160-184179160 on Build GRCh38
rs778167763 Chr.3: 184179206-184179206 on Build GRCh38
rs11539594 Chr.3: 184178897-184178897 on Build GRCh38
rs143114845 Chr.3: 184178710-184178710 on Build GRCh38
rs986782047 Chr.3: 184178746-184178746 on Build GRCh38
rs199638962 Chr.3: 184178838-184178838 on Build GRCh38
rs540567642 Chr.3: 184179149-184179149 on Build GRCh38
rs759532444 Chr.3: 184178818-184178818 on Build GRCh38
rs777567779 Chr.3: 184178921-184178921 on Build GRCh38
rs775327824 Chr.3: 184179004-184179004 on Build GRCh38
rs200520955 Chr.3: 184179153-184179153 on Build GRCh38
rs766742290 Chr.3: 184178850-184178850 on Build GRCh38
rs766685109 Chr.3: 184179082-184179082 on Build GRCh38
rs2231215 Chr.3: 184178723-184178723 on Build GRCh38
rs768041479 Chr.3: 184179136-184179136 on Build GRCh38
rs975651727 Chr.3: 184178845-184178845 on Build GRCh38
rs1134758 Chr.3: 184179098-184179098 on Build GRCh38
rs2231217 Chr.3: 184179178-184179178 on Build GRCh38
rs938377626 Chr.3: 184178988-184178988 on Build GRCh38
rs560148625 Chr.3: 184179043-184179043 on Build GRCh38
rs761259783 Chr.3: 184178805-184178805 on Build GRCh38
rs370947653 Chr.3: 184179134-184179134 on Build GRCh38
rs11539598 Chr.3: 184179019-184179019 on Build GRCh38

View Full Product Details