Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185204224-185204790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHHADH
Entrez Gene ID
1962
Gene Name
enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
Gene Aliases
ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE
UniGene
Hs.429879
Gene Chromosome Location
Chr.3: 185190624-185282886 on Build GRCh38
Transcripts
NM_001166415.1 NM_001966.3 XM_006713525.1 XM_011512517.1 XM_017005839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3331752 c.819C>T p.F273F Chr.3: 185204507-185204507 on Build GRCh38
1042170 c.667C>A p.Q223K Chr.3: 185204659-185204659 on Build GRCh38
3992912 c.747C>T p.I249I Chr.3: 185204579-185204579 on Build GRCh38
77638 c.901G>C p.G301R Chr.3: 185204425-185204425 on Build GRCh38
4799690 c.895T>A p.S299T Chr.3: 185204431-185204431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022316342 Chr.3: 185204657-185204657 on Build GRCh38
rs771075254 Chr.3: 185204529-185204529 on Build GRCh38
rs17283799 Chr.3: 185204409-185204409 on Build GRCh38
rs765372781 Chr.3: 185204375-185204375 on Build GRCh38
rs140514906 Chr.3: 185204445-185204445 on Build GRCh38
rs767872565 Chr.3: 185204470-185204470 on Build GRCh38
rs144336321 Chr.3: 185204255-185204255 on Build GRCh38
rs780567517 Chr.3: 185204739-185204739 on Build GRCh38
rs760094216 Chr.3: 185204468-185204468 on Build GRCh38
rs763360960 Chr.3: 185204403-185204403 on Build GRCh38
rs781090244 Chr.3: 185204731-185204732 on Build GRCh38
rs937477695 Chr.3: 185204654-185204654 on Build GRCh38
rs560229048 Chr.3: 185204505-185204505 on Build GRCh38
rs144566313 Chr.3: 185204608-185204608 on Build GRCh38
rs550345296 Chr.3: 185204336-185204336 on Build GRCh38
rs145972339 Chr.3: 185204694-185204694 on Build GRCh38
rs140339876 Chr.3: 185204465-185204465 on Build GRCh38
rs185059435 Chr.3: 185204462-185204462 on Build GRCh38
rs752275123 Chr.3: 185204471-185204471 on Build GRCh38
rs113061833 Chr.3: 185204261-185204261 on Build GRCh38
rs111629390 Chr.3: 185204419-185204419 on Build GRCh38
rs930349160 Chr.3: 185204530-185204530 on Build GRCh38
rs538005621 Chr.3: 185204328-185204328 on Build GRCh38
rs758171902 Chr.3: 185204572-185204572 on Build GRCh38
rs764753424 Chr.3: 185204564-185204564 on Build GRCh38
rs753393023 Chr.3: 185204683-185204683 on Build GRCh38
rs199689303 Chr.3: 185204719-185204719 on Build GRCh38
rs753620285 Chr.3: 185204664-185204664 on Build GRCh38
rs6802573 Chr.3: 185204250-185204250 on Build GRCh38
rs374946237 Chr.3: 185204450-185204450 on Build GRCh38
rs139788883 Chr.3: 185204705-185204705 on Build GRCh38
rs753610447 Chr.3: 185204498-185204498 on Build GRCh38
rs777322705 Chr.3: 185204745-185204745 on Build GRCh38
rs751036133 Chr.3: 185204458-185204458 on Build GRCh38
rs748925800 Chr.3: 185204421-185204421 on Build GRCh38
rs761258260 Chr.3: 185204547-185204547 on Build GRCh38
rs978287678 Chr.3: 185204484-185204484 on Build GRCh38
rs370789385 Chr.3: 185204443-185204443 on Build GRCh38
rs773593711 Chr.3: 185204391-185204391 on Build GRCh38
rs966265703 Chr.3: 185204706-185204706 on Build GRCh38
rs769930616 Chr.3: 185204526-185204526 on Build GRCh38
rs1057482044 Chr.3: 185204684-185204684 on Build GRCh38
rs779898866 Chr.3: 185204573-185204573 on Build GRCh38
rs754941485 Chr.3: 185204670-185204670 on Build GRCh38
rs900235331 Chr.3: 185204326-185204326 on Build GRCh38
rs752082700 Chr.3: 185204416-185204416 on Build GRCh38
rs367585416 Chr.3: 185204614-185204614 on Build GRCh38
rs1049200972 Chr.3: 185204368-185204368 on Build GRCh38
rs181605142 Chr.3: 185204253-185204253 on Build GRCh38
rs200735095 Chr.3: 185204634-185204634 on Build GRCh38
rs373788163 Chr.3: 185204585-185204585 on Build GRCh38
rs2302819 Chr.3: 185204506-185204506 on Build GRCh38
rs763665044 Chr.3: 185204496-185204496 on Build GRCh38
rs138063371 Chr.3: 185204543-185204543 on Build GRCh38
rs779909543 Chr.3: 185204522-185204522 on Build GRCh38
rs150375664 Chr.3: 185204537-185204537 on Build GRCh38
rs201465186 Chr.3: 185204507-185204507 on Build GRCh38
rs772810052 Chr.3: 185204652-185204652 on Build GRCh38
rs764777041 Chr.3: 185204638-185204638 on Build GRCh38
rs143364521 Chr.3: 185204418-185204418 on Build GRCh38
rs1001762073 Chr.3: 185204433-185204433 on Build GRCh38
rs961409299 Chr.3: 185204252-185204252 on Build GRCh38
rs762104990 Chr.3: 185204371-185204371 on Build GRCh38
rs757522486 Chr.3: 185204599-185204599 on Build GRCh38
rs750250817 Chr.3: 185204565-185204565 on Build GRCh38
rs773467007 Chr.3: 185204448-185204448 on Build GRCh38
rs999471043 Chr.3: 185204666-185204666 on Build GRCh38
rs148423296 Chr.3: 185204622-185204622 on Build GRCh38
rs754854776 Chr.3: 185204582-185204582 on Build GRCh38
rs746221793 Chr.3: 185204243-185204243 on Build GRCh38
rs567956351 Chr.3: 185204272-185204272 on Build GRCh38
rs766701520 Chr.3: 185204404-185204404 on Build GRCh38
rs758387500 Chr.3: 185204512-185204512 on Build GRCh38
rs752609032 Chr.3: 185204699-185204699 on Build GRCh38
rs748687324 Chr.3: 185204754-185204754 on Build GRCh38
rs749527982 Chr.3: 185204527-185204527 on Build GRCh38
rs763686437 Chr.3: 185204617-185204617 on Build GRCh38
rs149294851 Chr.3: 185204579-185204579 on Build GRCh38
rs752179101 Chr.3: 185204406-185204406 on Build GRCh38
rs745872713 Chr.3: 185204431-185204431 on Build GRCh38
rs762656795 Chr.3: 185204653-185204653 on Build GRCh38
rs114436171 Chr.3: 185204342-185204342 on Build GRCh38
rs114864468 Chr.3: 185204563-185204563 on Build GRCh38
rs868115450 Chr.3: 185204596-185204596 on Build GRCh38
rs761327979 Chr.3: 185204359-185204359 on Build GRCh38
rs1053619517 Chr.3: 185204364-185204364 on Build GRCh38
rs576283902 Chr.3: 185204437-185204437 on Build GRCh38
rs775900075 Chr.3: 185204546-185204546 on Build GRCh38
rs752984075 Chr.3: 185204480-185204480 on Build GRCh38
rs149115164 Chr.3: 185204718-185204718 on Build GRCh38
rs746921626 Chr.3: 185204523-185204523 on Build GRCh38
rs200753408 Chr.3: 185204613-185204613 on Build GRCh38
rs765105505 Chr.3: 185204756-185204756 on Build GRCh38
rs146633671 Chr.3: 185204755-185204755 on Build GRCh38
rs778565475 Chr.3: 185204429-185204429 on Build GRCh38
rs756975818 Chr.3: 185204694-185204697 on Build GRCh38
rs922061056 Chr.3: 185204722-185204722 on Build GRCh38
rs751289858 Chr.3: 185204665-185204665 on Build GRCh38
rs959720474 Chr.3: 185204463-185204463 on Build GRCh38
rs202051821 Chr.3: 185204740-185204740 on Build GRCh38
rs1019192267 Chr.3: 185204696-185204696 on Build GRCh38
rs141664962 Chr.3: 185204623-185204623 on Build GRCh38
rs115754857 Chr.3: 185204479-185204479 on Build GRCh38
rs747529583 Chr.3: 185204617-185204617 on Build GRCh38
rs748132233 Chr.3: 185204444-185204444 on Build GRCh38
rs146062368 Chr.3: 185204548-185204548 on Build GRCh38
rs201955662 Chr.3: 185204425-185204425 on Build GRCh38
rs753338712 Chr.3: 185204553-185204553 on Build GRCh38
rs538717513 Chr.3: 185204394-185204394 on Build GRCh38
rs759769910 Chr.3: 185204541-185204541 on Build GRCh38
rs769215989 Chr.3: 185204619-185204619 on Build GRCh38
rs984969980 Chr.3: 185204763-185204763 on Build GRCh38
rs779053739 Chr.3: 185204600-185204600 on Build GRCh38
rs749328305 Chr.3: 185204590-185204590 on Build GRCh38
rs912571859 Chr.3: 185204316-185204316 on Build GRCh38
rs1013285934 Chr.3: 185204455-185204455 on Build GRCh38
rs779045927 Chr.3: 185204708-185204708 on Build GRCh38

View Full Product Details