Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155088902-155089475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCC3
Entrez Gene ID
79184
Gene Name
BRCA1/BRCA2-containing complex subunit 3
Gene Aliases
BRCC36, C6.1A, CXorf53
UniGene
Hs.558537
Gene Chromosome Location
Chr.X: 155071403-155126766 on Build GRCh38
Transcripts
NM_001018055.2 NM_001242640.1 NM_024332.3 XM_005274751.4 XM_017029838.1

SNP Detail

SNP ID Chromosome Location
rs957746625 Chr.X: 155089114-155089114 on Build GRCh38
rs782639233 Chr.X: 155089238-155089238 on Build GRCh38
rs782706725 Chr.X: 155089047-155089047 on Build GRCh38
rs370260379 Chr.X: 155089384-155089384 on Build GRCh38
rs916458113 Chr.X: 155089004-155089004 on Build GRCh38
rs782606664 Chr.X: 155089180-155089180 on Build GRCh38
rs781964727 Chr.X: 155089096-155089096 on Build GRCh38
rs969183559 Chr.X: 155089438-155089438 on Build GRCh38
rs371170397 Chr.X: 155089225-155089225 on Build GRCh38
rs1012964975 Chr.X: 155089365-155089365 on Build GRCh38
rs899975483 Chr.X: 155089163-155089163 on Build GRCh38
rs782112138 Chr.X: 155089099-155089099 on Build GRCh38
rs782442985 Chr.X: 155089398-155089398 on Build GRCh38
rs782313824 Chr.X: 155089375-155089375 on Build GRCh38
rs782774669 Chr.X: 155089153-155089153 on Build GRCh38
rs1039428602 Chr.X: 155089157-155089157 on Build GRCh38
rs781851163 Chr.X: 155089353-155089353 on Build GRCh38
rs782129717 Chr.X: 155089373-155089373 on Build GRCh38
rs201427873 Chr.X: 155089237-155089237 on Build GRCh38
rs969213223 Chr.X: 155089036-155089036 on Build GRCh38
rs782818838 Chr.X: 155089299-155089299 on Build GRCh38
rs1023064832 Chr.X: 155089374-155089374 on Build GRCh38
rs782516571 Chr.X: 155089140-155089140 on Build GRCh38
rs12833808 Chr.X: 155089288-155089288 on Build GRCh38
rs782330342 Chr.X: 155089010-155089010 on Build GRCh38
rs781946105 Chr.X: 155089395-155089395 on Build GRCh38
rs781952593 Chr.X: 155089367-155089368 on Build GRCh38
rs989711651 Chr.X: 155088936-155088936 on Build GRCh38
rs782237616 Chr.X: 155089231-155089231 on Build GRCh38
rs143567106 Chr.X: 155089069-155089069 on Build GRCh38
rs377415541 Chr.X: 155089273-155089273 on Build GRCh38
rs12839639 Chr.X: 155089449-155089449 on Build GRCh38
rs782041937 Chr.X: 155089181-155089181 on Build GRCh38
rs957684045 Chr.X: 155089364-155089364 on Build GRCh38
rs371817972 Chr.X: 155089365-155089366 on Build GRCh38
rs927880706 Chr.X: 155089056-155089056 on Build GRCh38
rs929610129 Chr.X: 155089061-155089061 on Build GRCh38

View Full Product Details