Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 138631758-138632255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF13
Entrez Gene ID
2258
Gene Name
fibroblast growth factor 13
Gene Aliases
FGF-13, FGF2, FHF-2, FHF2
UniGene
Hs.6540
Gene Chromosome Location
Chr.X: 138631573-139222889 on Build GRCh38
Transcripts
NM_001139498.1 NM_001139500.1 NM_001139501.1 NM_001139502.1 NM_004114.3 NM_033642.2 XM_005262399.1

SNP Detail

SNP ID Chromosome Location
rs985919816 Chr.X: 138632047-138632047 on Build GRCh38
rs17284572 Chr.X: 138632223-138632223 on Build GRCh38
rs34170693 Chr.X: 138631982-138631983 on Build GRCh38
rs772141893 Chr.X: 138632195-138632195 on Build GRCh38
rs756781729 Chr.X: 138631832-138631832 on Build GRCh38
rs894675053 Chr.X: 138631985-138631985 on Build GRCh38
rs975169483 Chr.X: 138632191-138632191 on Build GRCh38
rs1003709650 Chr.X: 138631816-138631816 on Build GRCh38
rs1031219515 Chr.X: 138631853-138631853 on Build GRCh38
rs1062711 Chr.X: 138632123-138632123 on Build GRCh38
rs746331030 Chr.X: 138632166-138632166 on Build GRCh38
rs962344614 Chr.X: 138632173-138632173 on Build GRCh38
rs953220946 Chr.X: 138632038-138632038 on Build GRCh38
rs928959850 Chr.X: 138631899-138631899 on Build GRCh38
rs998825925 Chr.X: 138632188-138632188 on Build GRCh38
rs956985121 Chr.X: 138631874-138631874 on Build GRCh38
rs367783929 Chr.X: 138632020-138632020 on Build GRCh38
rs937783887 Chr.X: 138631956-138631956 on Build GRCh38
rs757536081 Chr.X: 138631946-138631946 on Build GRCh38
rs764688277 Chr.X: 138631977-138631977 on Build GRCh38
rs903440882 Chr.X: 138632131-138632131 on Build GRCh38
rs1045953429 Chr.X: 138632118-138632118 on Build GRCh38
rs754274254 Chr.X: 138631929-138631929 on Build GRCh38
rs1033465023 Chr.X: 138632218-138632218 on Build GRCh38
rs149912958 Chr.X: 138632201-138632201 on Build GRCh38
rs183821429 Chr.X: 138632091-138632091 on Build GRCh38
rs984873174 Chr.X: 138631904-138631904 on Build GRCh38
rs1010972297 Chr.X: 138631939-138631939 on Build GRCh38
rs781722025 Chr.X: 138632176-138632176 on Build GRCh38
rs911675660 Chr.X: 138632074-138632074 on Build GRCh38
rs919188041 Chr.X: 138631855-138631855 on Build GRCh38

View Full Product Details