Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109659953-109660417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCUB
Entrez Gene ID
55013
Gene Name
mitochondrial calcium uniporter dominant negative beta subunit
Gene Aliases
CCDC109B
UniGene
Hs.234149
Gene Chromosome Location
Chr.4: 109560199-109688719 on Build GRCh38
Transcripts
NM_017918.4 XM_006714246.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1426335 c.223T>C p.L75L Chr.4: 109660242-109660242 on Build GRCh38
3760330 c.188T>A p.I63N Chr.4: 109660207-109660207 on Build GRCh38
331391 c.30G>T p.Q10H Chr.4: 109660313-109660313 on Build GRCh38
6260086 c.331G>T p.A111S Chr.4: 109660350-109660350 on Build GRCh38
4941484 c.74T>C p.F25S Chr.4: 109660357-109660357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11373410 Chr.4: 109660178-109660178 on Build GRCh38
rs1012368346 Chr.4: 109660079-109660079 on Build GRCh38
rs763610302 Chr.4: 109660261-109660261 on Build GRCh38
rs768258227 Chr.4: 109660322-109660322 on Build GRCh38
rs757102911 Chr.4: 109660164-109660164 on Build GRCh38
rs537942250 Chr.4: 109660184-109660184 on Build GRCh38
rs747094722 Chr.4: 109660212-109660212 on Build GRCh38
rs149127194 Chr.4: 109660301-109660301 on Build GRCh38
rs397813223 Chr.4: 109660177-109660178 on Build GRCh38
rs763875854 Chr.4: 109660176-109660177 on Build GRCh38
rs767209760 Chr.4: 109660169-109660169 on Build GRCh38
rs764800511 Chr.4: 109660338-109660338 on Build GRCh38
rs911075348 Chr.4: 109660271-109660271 on Build GRCh38
rs775027003 Chr.4: 109660320-109660320 on Build GRCh38
rs35661664 Chr.4: 109660004-109660005 on Build GRCh38
rs1040918201 Chr.4: 109659977-109659977 on Build GRCh38
rs370601686 Chr.4: 109660316-109660316 on Build GRCh38
rs367793330 Chr.4: 109660381-109660381 on Build GRCh38
rs577695498 Chr.4: 109660321-109660321 on Build GRCh38
rs760280798 Chr.4: 109660260-109660260 on Build GRCh38
rs34754720 Chr.4: 109660168-109660169 on Build GRCh38
rs1051388565 Chr.4: 109660384-109660384 on Build GRCh38
rs753363097 Chr.4: 109660214-109660215 on Build GRCh38
rs555100175 Chr.4: 109660215-109660215 on Build GRCh38
rs755146159 Chr.4: 109660213-109660213 on Build GRCh38
rs771571379 Chr.4: 109660252-109660252 on Build GRCh38
rs771513878 Chr.4: 109660317-109660317 on Build GRCh38
rs761592489 Chr.4: 109660336-109660336 on Build GRCh38
rs958597249 Chr.4: 109660144-109660144 on Build GRCh38
rs775080152 Chr.4: 109660255-109660255 on Build GRCh38
rs760378581 Chr.4: 109660175-109660178 on Build GRCh38
rs992789851 Chr.4: 109660173-109660173 on Build GRCh38
rs999185546 Chr.4: 109660020-109660020 on Build GRCh38
rs201688118 Chr.4: 109660179-109660179 on Build GRCh38
rs62327694 Chr.4: 109660363-109660363 on Build GRCh38
rs1032539057 Chr.4: 109660032-109660032 on Build GRCh38
rs968489235 Chr.4: 109660143-109660144 on Build GRCh38
rs776383064 Chr.4: 109660265-109660265 on Build GRCh38
rs765136791 Chr.4: 109660268-109660268 on Build GRCh38
rs772931200 Chr.4: 109660344-109660344 on Build GRCh38
rs386401131 Chr.4: 109660171-109660172 on Build GRCh38
rs770243966 Chr.4: 109660224-109660224 on Build GRCh38
rs761255523 Chr.4: 109660343-109660344 on Build GRCh38
rs902213224 Chr.4: 109659999-109659999 on Build GRCh38
rs748430752 Chr.4: 109660217-109660217 on Build GRCh38
rs893551346 Chr.4: 109660045-109660045 on Build GRCh38
rs374193327 Chr.4: 109660282-109660282 on Build GRCh38
rs200586659 Chr.4: 109660376-109660376 on Build GRCh38
rs375817848 Chr.4: 109660143-109660143 on Build GRCh38
rs576092543 Chr.4: 109660144-109660147 on Build GRCh38
rs4698744 Chr.4: 109660207-109660207 on Build GRCh38
rs776134828 Chr.4: 109660324-109660324 on Build GRCh38
rs766328651 Chr.4: 109660295-109660295 on Build GRCh38
rs761207399 Chr.4: 109660269-109660269 on Build GRCh38
rs756433544 Chr.4: 109660312-109660312 on Build GRCh38
rs370786108 Chr.4: 109660234-109660234 on Build GRCh38
rs781249038 Chr.4: 109660304-109660304 on Build GRCh38
rs146159118 Chr.4: 109660314-109660314 on Build GRCh38
rs543404522 Chr.4: 109660351-109660351 on Build GRCh38
rs867856625 Chr.4: 109660200-109660200 on Build GRCh38
rs778679968 Chr.4: 109660168-109660168 on Build GRCh38
rs199863518 Chr.4: 109660243-109660243 on Build GRCh38
rs773582703 Chr.4: 109660239-109660239 on Build GRCh38
rs17040705 Chr.4: 109660092-109660092 on Build GRCh38

View Full Product Details