Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80944684-80945237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf22
Entrez Gene ID
255119
Gene Name
chromosome 4 open reading frame 22
Gene Aliases
-
UniGene
Hs.527104
Gene Chromosome Location
Chr.4: 80321265-80963756 on Build GRCh38
Transcripts
NM_001206997.1 NM_152770.2 XM_017007972.1 XM_017007973.1 XM_017007975.1 XM_017007978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3606428 c.598C>T p.P200S Chr.4: 80944880-80944880 on Build GRCh38
5551771 c.628G>A p.D210N Chr.4: 80944910-80944910 on Build GRCh38
5544579 c.540G>A p.W180* Chr.4: 80944822-80944822 on Build GRCh38
3606426 c.592G>A p.D198N Chr.4: 80944874-80944874 on Build GRCh38
3606424 c.580C>T p.Q194* Chr.4: 80944862-80944862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770005602 Chr.4: 80944765-80944765 on Build GRCh38
rs972085615 Chr.4: 80944758-80944758 on Build GRCh38
rs756154032 Chr.4: 80944971-80944971 on Build GRCh38
rs756472910 Chr.4: 80944958-80944961 on Build GRCh38
rs367718282 Chr.4: 80944927-80944927 on Build GRCh38
rs768032524 Chr.4: 80944964-80944964 on Build GRCh38
rs758672351 Chr.4: 80944833-80944833 on Build GRCh38
rs1015516583 Chr.4: 80945140-80945140 on Build GRCh38
rs993496501 Chr.4: 80945163-80945163 on Build GRCh38
rs559540033 Chr.4: 80944939-80944939 on Build GRCh38
rs539984599 Chr.4: 80944961-80944961 on Build GRCh38
rs201959026 Chr.4: 80944856-80944856 on Build GRCh38
rs762559834 Chr.4: 80944892-80944892 on Build GRCh38
rs766163358 Chr.4: 80944809-80944809 on Build GRCh38
rs765967357 Chr.4: 80944941-80944941 on Build GRCh38
rs756278492 Chr.4: 80944893-80944893 on Build GRCh38
rs768544459 Chr.4: 80944770-80944770 on Build GRCh38
rs767123092 Chr.4: 80944777-80944777 on Build GRCh38
rs919173766 Chr.4: 80944947-80944947 on Build GRCh38
rs760339841 Chr.4: 80944805-80944805 on Build GRCh38
rs547107467 Chr.4: 80945186-80945186 on Build GRCh38
rs1042576999 Chr.4: 80945114-80945114 on Build GRCh38
rs376053352 Chr.4: 80945149-80945149 on Build GRCh38
rs748042577 Chr.4: 80944917-80944917 on Build GRCh38
rs774260298 Chr.4: 80944912-80944912 on Build GRCh38
rs1027674090 Chr.4: 80945165-80945165 on Build GRCh38
rs745976654 Chr.4: 80944892-80944892 on Build GRCh38
rs531597141 Chr.4: 80945107-80945107 on Build GRCh38
rs976379492 Chr.4: 80944903-80944903 on Build GRCh38
rs772810572 Chr.4: 80944937-80944937 on Build GRCh38
rs762677729 Chr.4: 80944769-80944769 on Build GRCh38
rs751992783 Chr.4: 80944873-80944873 on Build GRCh38
rs1013899063 Chr.4: 80945127-80945127 on Build GRCh38
rs551860650 Chr.4: 80945079-80945079 on Build GRCh38
rs867338718 Chr.4: 80944838-80944838 on Build GRCh38
rs769966886 Chr.4: 80944853-80944853 on Build GRCh38
rs764384536 Chr.4: 80944978-80944979 on Build GRCh38
rs768332413 Chr.4: 80944910-80944910 on Build GRCh38
rs187409875 Chr.4: 80945204-80945204 on Build GRCh38
rs989538618 Chr.4: 80945081-80945081 on Build GRCh38
rs189134114 Chr.4: 80945067-80945067 on Build GRCh38
rs1042627656 Chr.4: 80944745-80944745 on Build GRCh38
rs761876765 Chr.4: 80944774-80944774 on Build GRCh38
rs143788216 Chr.4: 80945184-80945184 on Build GRCh38
rs1018165456 Chr.4: 80945197-80945197 on Build GRCh38
rs369510444 Chr.4: 80944951-80944951 on Build GRCh38
rs138962192 Chr.4: 80944741-80944741 on Build GRCh38
rs181571041 Chr.4: 80945158-80945158 on Build GRCh38
rs772863648 Chr.4: 80944801-80944801 on Build GRCh38
rs78647918 Chr.4: 80945102-80945102 on Build GRCh38
rs943425760 Chr.4: 80945003-80945004 on Build GRCh38
rs757771954 Chr.4: 80944874-80944874 on Build GRCh38
rs568310862 Chr.4: 80945180-80945180 on Build GRCh38
rs781178654 Chr.4: 80944886-80944886 on Build GRCh38
rs749063568 Chr.4: 80945074-80945074 on Build GRCh38
rs937810123 Chr.4: 80944980-80944980 on Build GRCh38
rs780247297 Chr.4: 80944985-80944985 on Build GRCh38
rs1035398525 Chr.4: 80944784-80944784 on Build GRCh38
rs776120582 Chr.4: 80944946-80944946 on Build GRCh38
rs376774145 Chr.4: 80944950-80944950 on Build GRCh38
rs574682803 Chr.4: 80944957-80944957 on Build GRCh38
rs149006937 Chr.4: 80944825-80944825 on Build GRCh38
rs751171265 Chr.4: 80944941-80944941 on Build GRCh38
rs780115293 Chr.4: 80944901-80944901 on Build GRCh38
rs374375143 Chr.4: 80944776-80944776 on Build GRCh38
rs1055397982 Chr.4: 80945000-80945000 on Build GRCh38
rs766039933 Chr.4: 80944935-80944954 on Build GRCh38
rs201740556 Chr.4: 80944969-80944969 on Build GRCh38

View Full Product Details