Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 13691510-13692109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
UniGene
Hs.212360
Gene Chromosome Location
Chr.5: 13690328-14011829 on Build GRCh38
Transcripts
NM_001369.2 XM_005248262.3 XM_017009177.1 XM_017009178.1 XM_017009179.1 XM_017009185.1 XM_017009186.1 XM_017009188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
375337 c.13843C>T p.R4615C Chr.5: 13692016-13692016 on Build GRCh38
5596602 c.13849G>A p.V4617I Chr.5: 13692010-13692010 on Build GRCh38
4585414 c.13778C>T p.T4593M Chr.5: 13692081-13692081 on Build GRCh38
5598233 c.13840C>T p.L4614F Chr.5: 13692019-13692019 on Build GRCh38
4648839 c.13798G>A p.A4600T Chr.5: 13692061-13692061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753263224 Chr.5: 13691935-13691935 on Build GRCh38
rs936169293 Chr.5: 13691703-13691703 on Build GRCh38
rs770244453 Chr.5: 13692042-13692042 on Build GRCh38
rs778223884 Chr.5: 13691988-13691988 on Build GRCh38
rs1046104837 Chr.5: 13691567-13691567 on Build GRCh38
rs866966083 Chr.5: 13691952-13691952 on Build GRCh38
rs866931796 Chr.5: 13691635-13691635 on Build GRCh38
rs201246418 Chr.5: 13691986-13691986 on Build GRCh38
rs372688121 Chr.5: 13691934-13691934 on Build GRCh38
rs752872674 Chr.5: 13692067-13692067 on Build GRCh38
rs1028972712 Chr.5: 13691854-13691854 on Build GRCh38
rs749171065 Chr.5: 13691970-13691970 on Build GRCh38
rs963719711 Chr.5: 13691689-13691689 on Build GRCh38
rs1038240439 Chr.5: 13691810-13691810 on Build GRCh38
rs777715365 Chr.5: 13691964-13691964 on Build GRCh38
rs375643128 Chr.5: 13692062-13692062 on Build GRCh38
rs771108785 Chr.5: 13692055-13692055 on Build GRCh38
rs1018054486 Chr.5: 13692006-13692006 on Build GRCh38
rs986567874 Chr.5: 13691685-13691685 on Build GRCh38
rs915053894 Chr.5: 13691596-13691596 on Build GRCh38
rs774279588 Chr.5: 13692061-13692061 on Build GRCh38
rs994437800 Chr.5: 13691652-13691652 on Build GRCh38
rs527863586 Chr.5: 13691884-13691884 on Build GRCh38
rs529580704 Chr.5: 13691731-13691731 on Build GRCh38
rs371639785 Chr.5: 13692044-13692044 on Build GRCh38
rs923678039 Chr.5: 13692010-13692010 on Build GRCh38
rs926216540 Chr.5: 13691694-13691694 on Build GRCh38
rs997910703 Chr.5: 13691761-13691761 on Build GRCh38
rs886059934 Chr.5: 13692034-13692034 on Build GRCh38
rs569850644 Chr.5: 13691910-13691910 on Build GRCh38
rs778207515 Chr.5: 13691629-13691629 on Build GRCh38
rs868385128 Chr.5: 13691916-13691916 on Build GRCh38
rs375610185 Chr.5: 13691984-13691984 on Build GRCh38
rs142036266 Chr.5: 13692081-13692081 on Build GRCh38
rs371038556 Chr.5: 13691549-13691549 on Build GRCh38
rs779043106 Chr.5: 13691885-13691885 on Build GRCh38
rs530012105 Chr.5: 13691643-13691643 on Build GRCh38
rs753887600 Chr.5: 13691945-13691945 on Build GRCh38
rs368120810 Chr.5: 13691869-13691869 on Build GRCh38
rs756083157 Chr.5: 13691946-13691946 on Build GRCh38
rs927275157 Chr.5: 13691754-13691754 on Build GRCh38
rs886059933 Chr.5: 13691892-13691892 on Build GRCh38
rs199643112 Chr.5: 13691938-13691938 on Build GRCh38
rs771806858 Chr.5: 13692082-13692082 on Build GRCh38
rs898453783 Chr.5: 13691861-13691861 on Build GRCh38
rs913403268 Chr.5: 13691724-13691724 on Build GRCh38
rs559328186 Chr.5: 13691612-13691612 on Build GRCh38
rs372556312 Chr.5: 13691688-13691688 on Build GRCh38
rs533564551 Chr.5: 13691879-13691879 on Build GRCh38
rs543361829 Chr.5: 13691608-13691608 on Build GRCh38
rs745602916 Chr.5: 13692011-13692011 on Build GRCh38
rs1005054424 Chr.5: 13691728-13691728 on Build GRCh38
rs905881566 Chr.5: 13691598-13691598 on Build GRCh38
rs752463884 Chr.5: 13691942-13691942 on Build GRCh38
rs771015963 Chr.5: 13691693-13691693 on Build GRCh38
rs139979856 Chr.5: 13691539-13691539 on Build GRCh38
rs976147107 Chr.5: 13691534-13691534 on Build GRCh38
rs564203224 Chr.5: 13691557-13691557 on Build GRCh38
rs955336890 Chr.5: 13691679-13691679 on Build GRCh38
rs367709427 Chr.5: 13692084-13692084 on Build GRCh38
rs143451462 Chr.5: 13691737-13691737 on Build GRCh38
rs147583407 Chr.5: 13692022-13692022 on Build GRCh38
rs867446476 Chr.5: 13692050-13692050 on Build GRCh38
rs151143347 Chr.5: 13691900-13691900 on Build GRCh38
rs771613197 Chr.5: 13692016-13692016 on Build GRCh38
rs541323896 Chr.5: 13691558-13691558 on Build GRCh38
rs760482011 Chr.5: 13692079-13692079 on Build GRCh38
rs191662365 Chr.5: 13692052-13692052 on Build GRCh38
rs1025381931 Chr.5: 13691655-13691655 on Build GRCh38
rs374253051 Chr.5: 13691789-13691789 on Build GRCh38
rs748494424 Chr.5: 13692032-13692032 on Build GRCh38
rs756808089 Chr.5: 13691973-13691973 on Build GRCh38
rs989424256 Chr.5: 13691718-13691718 on Build GRCh38
rs994011012 Chr.5: 13691883-13691883 on Build GRCh38
rs200555581 Chr.5: 13692080-13692080 on Build GRCh38
rs937367904 Chr.5: 13691799-13691799 on Build GRCh38
rs530814189 Chr.5: 13691924-13691924 on Build GRCh38
rs778096108 Chr.5: 13692009-13692009 on Build GRCh38

View Full Product Details