Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65658231-65658777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC13
Entrez Gene ID
80006
Gene Name
trafficking protein particle complex 13
Gene Aliases
C5orf44
UniGene
Hs.591760
Gene Chromosome Location
Chr.5: 65624731-65666136 on Build GRCh38
Transcripts
NM_001093755.1 NM_001093756.1 NM_001243737.1 NM_024941.3 XM_011543652.2 XM_017009890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4398081 c.609C>G p.T203T Chr.5: 65658412-65658412 on Build GRCh38
6807530 c.608C>T p.T203I Chr.5: 65658411-65658411 on Build GRCh38
3776791 c.591G>C p.Q197H Chr.5: 65658394-65658394 on Build GRCh38
6807531 c.611C>T p.T204I Chr.5: 65658411-65658411 on Build GRCh38
4398083 c.612C>G p.T204T Chr.5: 65658412-65658412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573154657 Chr.5: 65658411-65658411 on Build GRCh38
rs765612665 Chr.5: 65658331-65658331 on Build GRCh38
rs533037939 Chr.5: 65658689-65658689 on Build GRCh38
rs772147476 Chr.5: 65658509-65658509 on Build GRCh38
rs759264849 Chr.5: 65658533-65658533 on Build GRCh38
rs1000821510 Chr.5: 65658399-65658399 on Build GRCh38
rs748030671 Chr.5: 65658487-65658487 on Build GRCh38
rs398109019 Chr.5: 65658704-65658705 on Build GRCh38
rs765944642 Chr.5: 65658318-65658318 on Build GRCh38
rs146672885 Chr.5: 65658732-65658732 on Build GRCh38
rs781238575 Chr.5: 65658394-65658394 on Build GRCh38
rs942553310 Chr.5: 65658568-65658568 on Build GRCh38
rs745951858 Chr.5: 65658719-65658722 on Build GRCh38
rs540363733 Chr.5: 65658722-65658722 on Build GRCh38
rs925207592 Chr.5: 65658581-65658584 on Build GRCh38
rs386418378 Chr.5: 65658696-65658697 on Build GRCh38
rs758914399 Chr.5: 65658343-65658343 on Build GRCh38
rs56189866 Chr.5: 65658576-65658576 on Build GRCh38
rs34351655 Chr.5: 65658603-65658603 on Build GRCh38
rs1043509687 Chr.5: 65658581-65658581 on Build GRCh38
rs903427662 Chr.5: 65658344-65658344 on Build GRCh38
rs994187883 Chr.5: 65658722-65658722 on Build GRCh38
rs904913929 Chr.5: 65658682-65658683 on Build GRCh38
rs747080723 Chr.5: 65658358-65658358 on Build GRCh38
rs768283426 Chr.5: 65658416-65658416 on Build GRCh38
rs750852999 Chr.5: 65658695-65658695 on Build GRCh38
rs773338876 Chr.5: 65658521-65658521 on Build GRCh38
rs28084 Chr.5: 65658332-65658332 on Build GRCh38
rs202127914 Chr.5: 65658705-65658705 on Build GRCh38
rs1055910976 Chr.5: 65658693-65658693 on Build GRCh38
rs552930982 Chr.5: 65658391-65658391 on Build GRCh38
rs200115020 Chr.5: 65658710-65658710 on Build GRCh38
rs974373141 Chr.5: 65658671-65658671 on Build GRCh38
rs774317847 Chr.5: 65658428-65658428 on Build GRCh38
rs868421007 Chr.5: 65658491-65658491 on Build GRCh38
rs950299231 Chr.5: 65658711-65658711 on Build GRCh38
rs193271237 Chr.5: 65658603-65658603 on Build GRCh38
rs1044856334 Chr.5: 65658673-65658673 on Build GRCh38
rs753570022 Chr.5: 65658326-65658326 on Build GRCh38
rs910646783 Chr.5: 65658746-65658746 on Build GRCh38
rs955245723 Chr.5: 65658565-65658565 on Build GRCh38
rs1052355882 Chr.5: 65658635-65658635 on Build GRCh38
rs748632343 Chr.5: 65658489-65658489 on Build GRCh38
rs755275585 Chr.5: 65658330-65658330 on Build GRCh38
rs896170764 Chr.5: 65658660-65658660 on Build GRCh38
rs777774809 Chr.5: 65658357-65658357 on Build GRCh38
rs17830886 Chr.5: 65658547-65658547 on Build GRCh38
rs781707860 Chr.5: 65658405-65658412 on Build GRCh38
rs372504723 Chr.5: 65658304-65658304 on Build GRCh38
rs541877316 Chr.5: 65658422-65658422 on Build GRCh38
rs528854615 Chr.5: 65658742-65658742 on Build GRCh38
rs937311096 Chr.5: 65658692-65658692 on Build GRCh38
rs144581442 Chr.5: 65658651-65658651 on Build GRCh38
rs972092612 Chr.5: 65658269-65658269 on Build GRCh38
rs143790811 Chr.5: 65658698-65658699 on Build GRCh38
rs564319255 Chr.5: 65658667-65658667 on Build GRCh38
rs371092558 Chr.5: 65658527-65658527 on Build GRCh38
rs908335893 Chr.5: 65658665-65658665 on Build GRCh38
rs776098349 Chr.5: 65658530-65658530 on Build GRCh38

View Full Product Details