Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114178509-114179059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
UniGene
Hs.121478
Gene Chromosome Location
Chr.3: 114127623-114199407 on Build GRCh38
Transcripts
NM_000796.5 NM_001282563.2 NM_001290809.1 NM_033663.5 XM_017005829.1

SNP Detail

SNP ID Chromosome Location
rs201872343 Chr.3: 114178659-114178659 on Build GRCh38
rs1015453851 Chr.3: 114178620-114178620 on Build GRCh38
rs374173438 Chr.3: 114178602-114178602 on Build GRCh38
rs199937463 Chr.3: 114178971-114178971 on Build GRCh38
rs936213275 Chr.3: 114178995-114178995 on Build GRCh38
rs542599152 Chr.3: 114178732-114178732 on Build GRCh38
rs201380131 Chr.3: 114178859-114178859 on Build GRCh38
rs993178144 Chr.3: 114178941-114178941 on Build GRCh38
rs879885446 Chr.3: 114178533-114178533 on Build GRCh38
rs36211803 Chr.3: 114178873-114178873 on Build GRCh38
rs973896900 Chr.3: 114178918-114178918 on Build GRCh38
rs200161999 Chr.3: 114178946-114178946 on Build GRCh38
rs201591907 Chr.3: 114178795-114178795 on Build GRCh38
rs546525325 Chr.3: 114178779-114178779 on Build GRCh38
rs36212174 Chr.3: 114178875-114178875 on Build GRCh38
rs945017268 Chr.3: 114178958-114178958 on Build GRCh38
rs138571838 Chr.3: 114178963-114178963 on Build GRCh38
rs200452733 Chr.3: 114179012-114179012 on Build GRCh38
rs534477556 Chr.3: 114178933-114178933 on Build GRCh38
rs201861362 Chr.3: 114178900-114178900 on Build GRCh38
rs199852439 Chr.3: 114178902-114178902 on Build GRCh38
rs200881851 Chr.3: 114178668-114178668 on Build GRCh38
rs920752039 Chr.3: 114178687-114178687 on Build GRCh38
rs180684888 Chr.3: 114178638-114178638 on Build GRCh38
rs1012968398 Chr.3: 114178868-114178868 on Build GRCh38
rs202121521 Chr.3: 114178981-114178981 on Build GRCh38
rs200225823 Chr.3: 114178993-114178993 on Build GRCh38
rs199877784 Chr.3: 114178662-114178662 on Build GRCh38
rs201561838 Chr.3: 114178939-114178939 on Build GRCh38
rs886057790 Chr.3: 114178735-114178735 on Build GRCh38
rs978647536 Chr.3: 114178585-114178585 on Build GRCh38
rs199762499 Chr.3: 114178813-114178813 on Build GRCh38
rs36211802 Chr.3: 114178726-114178726 on Build GRCh38
rs184745368 Chr.3: 114178765-114178765 on Build GRCh38
rs771302402 Chr.3: 114178541-114178541 on Build GRCh38
rs200565231 Chr.3: 114178852-114178852 on Build GRCh38
rs894549950 Chr.3: 114178789-114178789 on Build GRCh38
rs201848348 Chr.3: 114178671-114178671 on Build GRCh38
rs752323522 Chr.3: 114178869-114178869 on Build GRCh38
rs550508277 Chr.3: 114178863-114178863 on Build GRCh38
rs182068801 Chr.3: 114178908-114178908 on Build GRCh38
rs190248679 Chr.3: 114178907-114178907 on Build GRCh38
rs200345655 Chr.3: 114178774-114178774 on Build GRCh38
rs1047969695 Chr.3: 114178735-114178735 on Build GRCh38
rs202143450 Chr.3: 114178745-114178745 on Build GRCh38
rs936160182 Chr.3: 114178747-114178747 on Build GRCh38
rs920679921 Chr.3: 114178986-114178986 on Build GRCh38
rs201785376 Chr.3: 114178959-114178959 on Build GRCh38
rs953938637 Chr.3: 114178945-114178945 on Build GRCh38
rs36212175 Chr.3: 114178954-114178954 on Build GRCh38
rs751251746 Chr.3: 114178890-114178890 on Build GRCh38
rs759059024 Chr.3: 114178622-114178622 on Build GRCh38
rs34804942 Chr.3: 114178605-114178605 on Build GRCh38
rs917298298 Chr.3: 114178637-114178637 on Build GRCh38
rs917245768 Chr.3: 114178948-114178948 on Build GRCh38
rs973846146 Chr.3: 114178625-114178625 on Build GRCh38
rs202129893 Chr.3: 114179006-114179006 on Build GRCh38
rs962558292 Chr.3: 114178914-114178914 on Build GRCh38
rs986866007 Chr.3: 114178643-114178643 on Build GRCh38
rs201385820 Chr.3: 114179004-114179004 on Build GRCh38
rs559171520 Chr.3: 114178552-114178552 on Build GRCh38
rs968558547 Chr.3: 114178563-114178563 on Build GRCh38
rs201251384 Chr.3: 114178974-114178974 on Build GRCh38
rs200384587 Chr.3: 114178698-114178698 on Build GRCh38
rs944963552 Chr.3: 114178704-114178704 on Build GRCh38
rs532308344 Chr.3: 114178862-114178862 on Build GRCh38
rs948834038 Chr.3: 114179018-114179018 on Build GRCh38
rs1024260193 Chr.3: 114178604-114178604 on Build GRCh38
rs982655380 Chr.3: 114178612-114178612 on Build GRCh38
rs924848577 Chr.3: 114178967-114178967 on Build GRCh38
rs1040732859 Chr.3: 114178816-114178816 on Build GRCh38
rs886057791 Chr.3: 114179009-114179009 on Build GRCh38

View Full Product Details