Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112162387-112162943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L4A
Entrez Gene ID
64097
Gene Name
erythrocyte membrane protein band 4.1 like 4A
Gene Aliases
EPB41L4, NBL4
UniGene
Hs.584954
Gene Chromosome Location
Chr.5: 112141829-112419490 on Build GRCh38
Transcripts
NM_022140.3 XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 XM_017009689.1 NR_015370.2

Gene Symbol
EPB41L4A-AS1
Entrez Gene ID
114915
Gene Name
EPB41L4A antisense RNA 1
Gene Aliases
C5orf26, NCRNA00219, TIGA1
UniGene
Hs.12082
Gene Chromosome Location
Chr.5: 112160526-112162501 on Build GRCh38
Transcripts
NM_022140.3 XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 XM_017009689.1 NR_015370.2

SNP Detail

SNP ID Chromosome Location
rs556689028 Chr.5: 112162482-112162482 on Build GRCh38
rs527291652 Chr.5: 112162826-112162826 on Build GRCh38
rs139215886 Chr.5: 112162415-112162415 on Build GRCh38
rs985452758 Chr.5: 112162784-112162784 on Build GRCh38
rs912554082 Chr.5: 112162789-112162789 on Build GRCh38
rs999268789 Chr.5: 112162907-112162907 on Build GRCh38
rs11558487 Chr.5: 112162856-112162856 on Build GRCh38
rs886606849 Chr.5: 112162774-112162774 on Build GRCh38
rs780836725 Chr.5: 112162484-112162484 on Build GRCh38
rs886490566 Chr.5: 112162535-112162535 on Build GRCh38
rs906203685 Chr.5: 112162906-112162906 on Build GRCh38
rs923789661 Chr.5: 112162496-112162496 on Build GRCh38
rs566526010 Chr.5: 112162801-112162801 on Build GRCh38
rs763749155 Chr.5: 112162897-112162900 on Build GRCh38
rs911591041 Chr.5: 112162689-112162689 on Build GRCh38
rs551448143 Chr.5: 112162760-112162760 on Build GRCh38
rs775694046 Chr.5: 112162862-112162862 on Build GRCh38
rs866927200 Chr.5: 112162475-112162475 on Build GRCh38
rs532920837 Chr.5: 112162753-112162753 on Build GRCh38
rs143331986 Chr.5: 112162548-112162548 on Build GRCh38
rs1030801472 Chr.5: 112162721-112162721 on Build GRCh38
rs752830857 Chr.5: 112162891-112162892 on Build GRCh38
rs544097675 Chr.5: 112162713-112162713 on Build GRCh38
rs770799415 Chr.5: 112162618-112162618 on Build GRCh38
rs930808092 Chr.5: 112162766-112162766 on Build GRCh38
rs966932282 Chr.5: 112162889-112162889 on Build GRCh38
rs1048614579 Chr.5: 112162866-112162866 on Build GRCh38
rs1004994184 Chr.5: 112162809-112162809 on Build GRCh38
rs915068230 Chr.5: 112162642-112162642 on Build GRCh38
rs542567728 Chr.5: 112162636-112162636 on Build GRCh38
rs769433387 Chr.5: 112162560-112162560 on Build GRCh38
rs759543193 Chr.5: 112162726-112162726 on Build GRCh38
rs914570632 Chr.5: 112162541-112162541 on Build GRCh38
rs900685949 Chr.5: 112162527-112162527 on Build GRCh38
rs945001745 Chr.5: 112162500-112162500 on Build GRCh38
rs373214341 Chr.5: 112162920-112162920 on Build GRCh38
rs368967446 Chr.5: 112162626-112162626 on Build GRCh38
rs190650076 Chr.5: 112162644-112162644 on Build GRCh38
rs538800952 Chr.5: 112162873-112162873 on Build GRCh38
rs1024749633 Chr.5: 112162465-112162465 on Build GRCh38
rs747855413 Chr.5: 112162540-112162540 on Build GRCh38
rs545465472 Chr.5: 112162570-112162570 on Build GRCh38
rs944343721 Chr.5: 112162710-112162710 on Build GRCh38
rs1050255726 Chr.5: 112162554-112162554 on Build GRCh38
rs148402953 Chr.5: 112162859-112162859 on Build GRCh38
rs561660033 Chr.5: 112162672-112162672 on Build GRCh38
rs774309179 Chr.5: 112162715-112162715 on Build GRCh38
rs994982330 Chr.5: 112162528-112162528 on Build GRCh38
rs761041755 Chr.5: 112162871-112162871 on Build GRCh38
rs1048665155 Chr.5: 112162670-112162670 on Build GRCh38
rs560610715 Chr.5: 112162736-112162742 on Build GRCh38
rs1027745028 Chr.5: 112162530-112162530 on Build GRCh38
rs571801190 Chr.5: 112162870-112162870 on Build GRCh38
rs79658925 Chr.5: 112162582-112162582 on Build GRCh38
rs144816254 Chr.5: 112162679-112162679 on Build GRCh38
rs1039291403 Chr.5: 112162522-112162522 on Build GRCh38
rs571719305 Chr.5: 112162752-112162753 on Build GRCh38
rs911608624 Chr.5: 112162909-112162909 on Build GRCh38
rs914952215 Chr.5: 112162437-112162437 on Build GRCh38
rs186408051 Chr.5: 112162579-112162579 on Build GRCh38
rs1025075471 Chr.5: 112162709-112162712 on Build GRCh38
rs989238735 Chr.5: 112162631-112162631 on Build GRCh38
rs141653088 Chr.5: 112162902-112162902 on Build GRCh38
rs868646457 Chr.5: 112162529-112162529 on Build GRCh38
rs556510792 Chr.5: 112162861-112162861 on Build GRCh38
rs765619268 Chr.5: 112162856-112162857 on Build GRCh38
rs989207663 Chr.5: 112162418-112162418 on Build GRCh38
rs945044055 Chr.5: 112162468-112162468 on Build GRCh38
rs999685728 Chr.5: 112162712-112162712 on Build GRCh38
rs977924193 Chr.5: 112162477-112162477 on Build GRCh38
rs890115384 Chr.5: 112162485-112162485 on Build GRCh38
rs1037555428 Chr.5: 112162842-112162842 on Build GRCh38
rs944101245 Chr.5: 112162799-112162799 on Build GRCh38
rs1037162631 Chr.5: 112162607-112162607 on Build GRCh38
rs1033532291 Chr.5: 112162608-112162608 on Build GRCh38
rs1049294193 Chr.5: 112162767-112162767 on Build GRCh38
rs897335850 Chr.5: 112162625-112162625 on Build GRCh38
rs945185386 Chr.5: 112162660-112162660 on Build GRCh38
rs1022108086 Chr.5: 112162872-112162872 on Build GRCh38
rs182888525 Chr.5: 112162895-112162895 on Build GRCh38
rs562681794 Chr.5: 112162745-112162745 on Build GRCh38
rs548964300 Chr.5: 112162832-112162832 on Build GRCh38
rs965744657 Chr.5: 112162922-112162922 on Build GRCh38

View Full Product Details