Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147881038-147881626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf46
Entrez Gene ID
389336
Gene Name
chromosome 5 open reading frame 46
Gene Aliases
SSSP1
UniGene
Hs.660038
Gene Chromosome Location
Chr.5: 147850463-147906594 on Build GRCh38
Transcripts
XM_017009459.1 NM_054023.4

Gene Symbol
SCGB3A2
Entrez Gene ID
117156
Gene Name
secretoglobin family 3A member 2
Gene Aliases
LU103, PNSP1, UGRP1, pnSP-1
UniGene
Hs.483765
Gene Chromosome Location
Chr.5: 147878711-147882193 on Build GRCh38
Transcripts
XM_017009459.1 NM_054023.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3852526 c.181G>C p.E61Q Chr.5: 147881571-147881571 on Build GRCh38
1063952 c.73A>G p.N25D Chr.5: 147881463-147881463 on Build GRCh38
6772683 c.82C>T p.P28S Chr.5: 147881472-147881472 on Build GRCh38
3827348 c.134T>G p.F45C Chr.5: 147881524-147881524 on Build GRCh38
5887911 c.133T>C p.F45L Chr.5: 147881523-147881523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007646539 Chr.5: 147881169-147881169 on Build GRCh38
rs758462858 Chr.5: 147881446-147881446 on Build GRCh38
rs919483211 Chr.5: 147881322-147881322 on Build GRCh38
rs772649271 Chr.5: 147881426-147881426 on Build GRCh38
rs770723371 Chr.5: 147881493-147881493 on Build GRCh38
rs1029558729 Chr.5: 147881211-147881211 on Build GRCh38
rs991184983 Chr.5: 147881270-147881270 on Build GRCh38
rs774916078 Chr.5: 147881517-147881517 on Build GRCh38
rs757836250 Chr.5: 147881427-147881427 on Build GRCh38
rs1022681621 Chr.5: 147881284-147881284 on Build GRCh38
rs531673765 Chr.5: 147881263-147881263 on Build GRCh38
rs765774292 Chr.5: 147881441-147881441 on Build GRCh38
rs892571867 Chr.5: 147881363-147881363 on Build GRCh38
rs140212139 Chr.5: 147881597-147881597 on Build GRCh38
rs60460686 Chr.5: 147881173-147881173 on Build GRCh38
rs757143528 Chr.5: 147881541-147881546 on Build GRCh38
rs73261387 Chr.5: 147881394-147881394 on Build GRCh38
rs1049266394 Chr.5: 147881151-147881151 on Build GRCh38
rs984549634 Chr.5: 147881196-147881196 on Build GRCh38
rs922958334 Chr.5: 147881499-147881499 on Build GRCh38
rs140936862 Chr.5: 147881463-147881463 on Build GRCh38
rs747232043 Chr.5: 147881464-147881464 on Build GRCh38
rs147045939 Chr.5: 147881412-147881412 on Build GRCh38
rs554915459 Chr.5: 147881259-147881259 on Build GRCh38
rs74826199 Chr.5: 147881139-147881139 on Build GRCh38
rs372140237 Chr.5: 147881384-147881384 on Build GRCh38
rs763908581 Chr.5: 147881539-147881539 on Build GRCh38
rs182450127 Chr.5: 147881153-147881153 on Build GRCh38
rs200621359 Chr.5: 147881457-147881457 on Build GRCh38
rs576910137 Chr.5: 147881116-147881116 on Build GRCh38
rs142388312 Chr.5: 147881485-147881485 on Build GRCh38
rs757007939 Chr.5: 147881599-147881599 on Build GRCh38
rs774298931 Chr.5: 147881496-147881496 on Build GRCh38
rs151333009 Chr.5: 147881550-147881550 on Build GRCh38
rs780518946 Chr.5: 147881540-147881540 on Build GRCh38
rs771584795 Chr.5: 147881514-147881514 on Build GRCh38
rs374818057 Chr.5: 147881437-147881437 on Build GRCh38
rs750911275 Chr.5: 147881562-147881562 on Build GRCh38
rs745415240 Chr.5: 147881541-147881541 on Build GRCh38
rs940324159 Chr.5: 147881230-147881230 on Build GRCh38
rs746750493 Chr.5: 147881472-147881472 on Build GRCh38
rs750914244 Chr.5: 147881445-147881445 on Build GRCh38
rs140599638 Chr.5: 147881560-147881560 on Build GRCh38
rs576780929 Chr.5: 147881421-147881421 on Build GRCh38
rs766960636 Chr.5: 147881401-147881401 on Build GRCh38
rs6859391 Chr.5: 147881252-147881252 on Build GRCh38
rs760362085 Chr.5: 147881520-147881520 on Build GRCh38
rs576050539 Chr.5: 147881356-147881356 on Build GRCh38
rs41291427 Chr.5: 147881293-147881293 on Build GRCh38
rs919520136 Chr.5: 147881103-147881107 on Build GRCh38
rs755052157 Chr.5: 147881586-147881586 on Build GRCh38
rs186588392 Chr.5: 147881181-147881181 on Build GRCh38
rs113436975 Chr.5: 147881407-147881407 on Build GRCh38
rs923164117 Chr.5: 147881251-147881251 on Build GRCh38
rs558660994 Chr.5: 147881113-147881113 on Build GRCh38
rs1004343272 Chr.5: 147881460-147881460 on Build GRCh38
rs6859234 Chr.5: 147881175-147881175 on Build GRCh38
rs750594552 Chr.5: 147881092-147881092 on Build GRCh38
rs1002458498 Chr.5: 147881409-147881409 on Build GRCh38
rs763418998 Chr.5: 147881571-147881571 on Build GRCh38
rs925130391 Chr.5: 147881305-147881305 on Build GRCh38
rs556241547 Chr.5: 147881481-147881481 on Build GRCh38
rs191891444 Chr.5: 147881369-147881369 on Build GRCh38
rs745826855 Chr.5: 147881513-147881513 on Build GRCh38
rs41291429 Chr.5: 147881205-147881205 on Build GRCh38
rs150100560 Chr.5: 147881582-147881582 on Build GRCh38
rs971063440 Chr.5: 147881301-147881301 on Build GRCh38
rs755248204 Chr.5: 147881472-147881472 on Build GRCh38
rs139166445 Chr.5: 147881328-147881328 on Build GRCh38
rs760289285 Chr.5: 147881406-147881406 on Build GRCh38
rs386693356 Chr.5: 147881173-147881175 on Build GRCh38
rs775173133 Chr.5: 147881405-147881405 on Build GRCh38
rs975741002 Chr.5: 147881241-147881241 on Build GRCh38
rs142978672 Chr.5: 147881340-147881340 on Build GRCh38
rs745799049 Chr.5: 147881602-147881602 on Build GRCh38
rs778616621 Chr.5: 147881601-147881601 on Build GRCh38
rs146666696 Chr.5: 147881585-147881585 on Build GRCh38
rs550250313 Chr.5: 147881291-147881291 on Build GRCh38
rs961784042 Chr.5: 147881473-147881473 on Build GRCh38
rs755395634 Chr.5: 147881358-147881358 on Build GRCh38

View Full Product Details