Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171448991-171449567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF18
Entrez Gene ID
8817
Gene Name
fibroblast growth factor 18
Gene Aliases
FGF-18, ZFGF5
UniGene
Hs.87191
Gene Chromosome Location
Chr.5: 171419663-171457626 on Build GRCh38
Transcripts
NM_003862.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3827683 c.286C>G p.Q96E Chr.5: 171449182-171449182 on Build GRCh38
6311910 c.335G>T p.R112L Chr.5: 171449231-171449231 on Build GRCh38
5497814 c.260T>C p.L87P Chr.5: 171449156-171449156 on Build GRCh38
3339485 c.312G>A p.T104T Chr.5: 171449208-171449208 on Build GRCh38
1436012 c.352G>T p.G118W Chr.5: 171449248-171449248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113795603 Chr.5: 171449451-171449451 on Build GRCh38
rs771665409 Chr.5: 171449138-171449138 on Build GRCh38
rs908750637 Chr.5: 171449242-171449242 on Build GRCh38
rs980389634 Chr.5: 171449421-171449421 on Build GRCh38
rs56932885 Chr.5: 171449401-171449402 on Build GRCh38
rs978540481 Chr.5: 171449360-171449360 on Build GRCh38
rs146062033 Chr.5: 171449241-171449241 on Build GRCh38
rs574620399 Chr.5: 171449486-171449486 on Build GRCh38
rs758140567 Chr.5: 171449289-171449289 on Build GRCh38
rs199946177 Chr.5: 171449286-171449286 on Build GRCh38
rs888254877 Chr.5: 171449056-171449056 on Build GRCh38
rs377015632 Chr.5: 171449492-171449492 on Build GRCh38
rs977378302 Chr.5: 171449154-171449154 on Build GRCh38
rs878902685 Chr.5: 171449408-171449408 on Build GRCh38
rs534322394 Chr.5: 171449143-171449143 on Build GRCh38
rs967251892 Chr.5: 171449440-171449440 on Build GRCh38
rs778403912 Chr.5: 171449121-171449121 on Build GRCh38
rs992524872 Chr.5: 171449398-171449398 on Build GRCh38
rs375630229 Chr.5: 171449175-171449175 on Build GRCh38
rs796123762 Chr.5: 171449402-171449404 on Build GRCh38
rs747946480 Chr.5: 171449267-171449268 on Build GRCh38
rs879786440 Chr.5: 171449039-171449039 on Build GRCh38
rs1031456852 Chr.5: 171449387-171449387 on Build GRCh38
rs372395610 Chr.5: 171449346-171449346 on Build GRCh38
rs759836429 Chr.5: 171449212-171449212 on Build GRCh38
rs760200784 Chr.5: 171449532-171449533 on Build GRCh38
rs186978754 Chr.5: 171449369-171449369 on Build GRCh38
rs775139267 Chr.5: 171449360-171449361 on Build GRCh38
rs1018779832 Chr.5: 171449416-171449416 on Build GRCh38
rs112415649 Chr.5: 171449401-171449406 on Build GRCh38
rs56323021 Chr.5: 171449402-171449402 on Build GRCh38
rs746517764 Chr.5: 171449305-171449305 on Build GRCh38
rs890431790 Chr.5: 171449524-171449524 on Build GRCh38
rs867474061 Chr.5: 171449189-171449189 on Build GRCh38
rs373312537 Chr.5: 171449157-171449157 on Build GRCh38
rs62383996 Chr.5: 171449404-171449404 on Build GRCh38
rs62383997 Chr.5: 171449425-171449425 on Build GRCh38
rs375522342 Chr.5: 171449422-171449425 on Build GRCh38
rs757697994 Chr.5: 171449120-171449120 on Build GRCh38
rs1036864516 Chr.5: 171449475-171449475 on Build GRCh38
rs778716813 Chr.5: 171449131-171449133 on Build GRCh38
rs142736846 Chr.5: 171449223-171449223 on Build GRCh38
rs942131105 Chr.5: 171449073-171449073 on Build GRCh38
rs888883202 Chr.5: 171449381-171449404 on Build GRCh38
rs567881592 Chr.5: 171449276-171449276 on Build GRCh38
rs969395443 Chr.5: 171449410-171449410 on Build GRCh38
rs1009122951 Chr.5: 171449385-171449385 on Build GRCh38
rs953315108 Chr.5: 171449104-171449104 on Build GRCh38
rs558603829 Chr.5: 171449347-171449347 on Build GRCh38
rs772036466 Chr.5: 171449059-171449059 on Build GRCh38
rs144451411 Chr.5: 171449400-171449400 on Build GRCh38
rs1038624204 Chr.5: 171449275-171449275 on Build GRCh38
rs929379968 Chr.5: 171449491-171449491 on Build GRCh38
rs879430876 Chr.5: 171449401-171449402 on Build GRCh38
rs765232190 Chr.5: 171449245-171449245 on Build GRCh38
rs1030270383 Chr.5: 171449274-171449274 on Build GRCh38
rs745570086 Chr.5: 171449125-171449125 on Build GRCh38
rs776780469 Chr.5: 171449155-171449155 on Build GRCh38
rs200833479 Chr.5: 171449277-171449277 on Build GRCh38
rs764539070 Chr.5: 171449445-171449446 on Build GRCh38
rs774406179 Chr.5: 171449187-171449187 on Build GRCh38
rs769680920 Chr.5: 171449169-171449169 on Build GRCh38
rs757526091 Chr.5: 171449285-171449285 on Build GRCh38
rs201988531 Chr.5: 171449231-171449231 on Build GRCh38
rs753821843 Chr.5: 171449428-171449431 on Build GRCh38
rs779730682 Chr.5: 171449142-171449142 on Build GRCh38
rs779646948 Chr.5: 171449296-171449296 on Build GRCh38
rs112697159 Chr.5: 171449406-171449406 on Build GRCh38
rs560807495 Chr.5: 171449477-171449477 on Build GRCh38
rs148266720 Chr.5: 171449470-171449470 on Build GRCh38
rs369769318 Chr.5: 171449262-171449262 on Build GRCh38
rs571271571 Chr.5: 171449081-171449081 on Build GRCh38
rs955758340 Chr.5: 171449397-171449397 on Build GRCh38
rs1051728225 Chr.5: 171449523-171449523 on Build GRCh38
rs1017788821 Chr.5: 171449340-171449340 on Build GRCh38
rs916027671 Chr.5: 171449200-171449200 on Build GRCh38
rs747809232 Chr.5: 171449135-171449135 on Build GRCh38
rs576812134 Chr.5: 171449331-171449331 on Build GRCh38
rs1032967381 Chr.5: 171449452-171449452 on Build GRCh38
rs879169582 Chr.5: 171449387-171449388 on Build GRCh38
rs1038158120 Chr.5: 171449079-171449079 on Build GRCh38

View Full Product Details