Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89268848-89269346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR2
Entrez Gene ID
2570
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Gene Aliases
-
UniGene
Hs.379330
Gene Chromosome Location
Chr.6: 89254464-89315299 on Build GRCh38
Transcripts
NM_002043.4 XM_011535713.2 XM_011535714.2 XM_011535715.2 XM_011535716.2 XM_011535717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434573 c.584T>C p.M195T Chr.6: 89269014-89269014 on Build GRCh38
3721884 c.439T>A p.F147I Chr.6: 89269084-89269084 on Build GRCh38
5434574 c.509T>C p.M170T Chr.6: 89269014-89269014 on Build GRCh38
6514206 c.401G>T p.W134L Chr.6: 89269122-89269122 on Build GRCh38
5521152 c.426C>T p.S142S Chr.6: 89269172-89269172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027204872 Chr.6: 89268904-89268904 on Build GRCh38
rs777756660 Chr.6: 89269260-89269260 on Build GRCh38
rs752323967 Chr.6: 89268977-89268977 on Build GRCh38
rs750928468 Chr.6: 89269143-89269143 on Build GRCh38
rs759263104 Chr.6: 89268969-89268969 on Build GRCh38
rs753459276 Chr.6: 89269133-89269133 on Build GRCh38
rs372349024 Chr.6: 89268980-89268980 on Build GRCh38
rs147512746 Chr.6: 89269140-89269140 on Build GRCh38
rs767516561 Chr.6: 89269246-89269246 on Build GRCh38
rs566218184 Chr.6: 89269209-89269209 on Build GRCh38
rs74555654 Chr.6: 89268916-89268916 on Build GRCh38
rs763564107 Chr.6: 89268993-89268993 on Build GRCh38
rs754367318 Chr.6: 89268994-89268994 on Build GRCh38
rs990484317 Chr.6: 89269027-89269027 on Build GRCh38
rs758697296 Chr.6: 89269045-89269045 on Build GRCh38
rs1031054343 Chr.6: 89269301-89269301 on Build GRCh38
rs202108132 Chr.6: 89268952-89268952 on Build GRCh38
rs1012224907 Chr.6: 89268925-89268925 on Build GRCh38
rs961902991 Chr.6: 89268924-89268924 on Build GRCh38
rs755210127 Chr.6: 89269170-89269170 on Build GRCh38
rs185917468 Chr.6: 89269220-89269220 on Build GRCh38
rs181484637 Chr.6: 89269185-89269185 on Build GRCh38
rs140140640 Chr.6: 89269141-89269141 on Build GRCh38
rs763733884 Chr.6: 89269277-89269277 on Build GRCh38
rs763695466 Chr.6: 89269125-89269125 on Build GRCh38
rs780270041 Chr.6: 89269053-89269053 on Build GRCh38
rs569842676 Chr.6: 89269245-89269245 on Build GRCh38
rs774187983 Chr.6: 89268967-89268967 on Build GRCh38
rs189434112 Chr.6: 89268995-89268995 on Build GRCh38
rs894496255 Chr.6: 89269107-89269107 on Build GRCh38
rs372516073 Chr.6: 89269117-89269117 on Build GRCh38
rs776431412 Chr.6: 89269213-89269213 on Build GRCh38
rs369521101 Chr.6: 89268964-89268964 on Build GRCh38
rs756164179 Chr.6: 89269255-89269255 on Build GRCh38
rs779410330 Chr.6: 89269023-89269023 on Build GRCh38
rs369231038 Chr.6: 89269109-89269109 on Build GRCh38
rs770795229 Chr.6: 89269088-89269088 on Build GRCh38
rs768368381 Chr.6: 89269210-89269210 on Build GRCh38
rs542856438 Chr.6: 89268966-89268966 on Build GRCh38
rs977549914 Chr.6: 89268995-89268996 on Build GRCh38
rs764885638 Chr.6: 89269228-89269228 on Build GRCh38
rs932538951 Chr.6: 89269037-89269037 on Build GRCh38
rs749207947 Chr.6: 89269086-89269086 on Build GRCh38
rs181374667 Chr.6: 89269249-89269249 on Build GRCh38
rs752938192 Chr.6: 89269252-89269252 on Build GRCh38
rs750763228 Chr.6: 89269028-89269028 on Build GRCh38
rs778773400 Chr.6: 89269187-89269187 on Build GRCh38
rs775218481 Chr.6: 89269199-89269199 on Build GRCh38
rs200936065 Chr.6: 89268907-89268907 on Build GRCh38
rs953060479 Chr.6: 89268949-89268949 on Build GRCh38
rs35013548 Chr.6: 89269108-89269109 on Build GRCh38
rs751799384 Chr.6: 89269148-89269148 on Build GRCh38
rs773734376 Chr.6: 89269231-89269231 on Build GRCh38
rs752214685 Chr.6: 89269172-89269172 on Build GRCh38
rs938956769 Chr.6: 89269035-89269035 on Build GRCh38
rs766749657 Chr.6: 89269246-89269246 on Build GRCh38
rs761966019 Chr.6: 89269159-89269169 on Build GRCh38
rs897653981 Chr.6: 89269261-89269261 on Build GRCh38
rs745843321 Chr.6: 89269194-89269194 on Build GRCh38
rs1030444452 Chr.6: 89268946-89268946 on Build GRCh38
rs768747095 Chr.6: 89269071-89269071 on Build GRCh38
rs996165332 Chr.6: 89268895-89268895 on Build GRCh38
rs576789300 Chr.6: 89268879-89268879 on Build GRCh38
rs751271291 Chr.6: 89269292-89269292 on Build GRCh38
rs953178358 Chr.6: 89269310-89269310 on Build GRCh38
rs997416301 Chr.6: 89269298-89269298 on Build GRCh38
rs955863014 Chr.6: 89269025-89269025 on Build GRCh38
rs781404279 Chr.6: 89269171-89269171 on Build GRCh38
rs747240789 Chr.6: 89269070-89269070 on Build GRCh38
rs1024918819 Chr.6: 89268943-89268943 on Build GRCh38
rs771786628 Chr.6: 89269113-89269113 on Build GRCh38
rs369162796 Chr.6: 89269264-89269264 on Build GRCh38
rs74923708 Chr.6: 89269147-89269147 on Build GRCh38
rs548869077 Chr.6: 89268887-89268887 on Build GRCh38
rs199577106 Chr.6: 89269197-89269197 on Build GRCh38
rs188424932 Chr.6: 89269227-89269227 on Build GRCh38
rs759334518 Chr.6: 89269111-89269111 on Build GRCh38
rs202047754 Chr.6: 89269248-89269248 on Build GRCh38
rs751786690 Chr.6: 89269126-89269128 on Build GRCh38
rs763874884 Chr.6: 89269045-89269047 on Build GRCh38
rs531769395 Chr.6: 89269085-89269085 on Build GRCh38
rs373992917 Chr.6: 89268962-89268962 on Build GRCh38
rs529728554 Chr.6: 89268934-89268934 on Build GRCh38

View Full Product Details