Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87681417-87681966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKIRIN2
Entrez Gene ID
55122
Gene Name
akirin 2
Gene Aliases
C6orf166, FBI1, dJ486L4.2
UniGene
Hs.485915
Gene Chromosome Location
Chr.6: 87674860-87702267 on Build GRCh38
Transcripts
NM_018064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256281 c.272G>A p.R91Q Chr.6: 87681727-87681727 on Build GRCh38
6566186 c.338C>T p.A113V Chr.6: 87681661-87681661 on Build GRCh38
6175113 c.361A>T p.S121C Chr.6: 87681638-87681638 on Build GRCh38
4826634 c.374C>T p.S125L Chr.6: 87681625-87681625 on Build GRCh38
1082382 c.295G>T p.E99* Chr.6: 87681704-87681704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766062158 Chr.6: 87681578-87681578 on Build GRCh38
rs530761426 Chr.6: 87681516-87681516 on Build GRCh38
rs112707559 Chr.6: 87681538-87681538 on Build GRCh38
rs11666 Chr.6: 87681659-87681659 on Build GRCh38
rs892039726 Chr.6: 87681930-87681930 on Build GRCh38
rs774245889 Chr.6: 87681676-87681676 on Build GRCh38
rs772546657 Chr.6: 87681738-87681739 on Build GRCh38
rs755037981 Chr.6: 87681787-87681787 on Build GRCh38
rs757849354 Chr.6: 87681723-87681723 on Build GRCh38
rs755711101 Chr.6: 87681596-87681596 on Build GRCh38
rs1045172342 Chr.6: 87681846-87681846 on Build GRCh38
rs751618895 Chr.6: 87681504-87681504 on Build GRCh38
rs766210052 Chr.6: 87681571-87681571 on Build GRCh38
rs528541660 Chr.6: 87681816-87681816 on Build GRCh38
rs55882516 Chr.6: 87681938-87681938 on Build GRCh38
rs911061943 Chr.6: 87681652-87681652 on Build GRCh38
rs760192931 Chr.6: 87681697-87681697 on Build GRCh38
rs780185634 Chr.6: 87681771-87681771 on Build GRCh38
rs780083608 Chr.6: 87681650-87681650 on Build GRCh38
rs200639997 Chr.6: 87681640-87681640 on Build GRCh38
rs62000376 Chr.6: 87681687-87681687 on Build GRCh38
rs149384256 Chr.6: 87681616-87681616 on Build GRCh38
rs779388400 Chr.6: 87681632-87681632 on Build GRCh38
rs1000357252 Chr.6: 87681890-87681890 on Build GRCh38
rs985926151 Chr.6: 87681639-87681639 on Build GRCh38
rs746073678 Chr.6: 87681638-87681638 on Build GRCh38
rs779115184 Chr.6: 87681724-87681724 on Build GRCh38
rs150426711 Chr.6: 87681674-87681674 on Build GRCh38
rs762459527 Chr.6: 87681546-87681546 on Build GRCh38
rs768086157 Chr.6: 87681771-87681773 on Build GRCh38
rs748551748 Chr.6: 87681453-87681453 on Build GRCh38
rs778382256 Chr.6: 87681597-87681597 on Build GRCh38
rs767108648 Chr.6: 87681579-87681579 on Build GRCh38
rs370136919 Chr.6: 87681470-87681470 on Build GRCh38
rs745498729 Chr.6: 87681809-87681810 on Build GRCh38
rs747247893 Chr.6: 87681774-87681774 on Build GRCh38
rs1033286102 Chr.6: 87681903-87681903 on Build GRCh38
rs754283062 Chr.6: 87681699-87681699 on Build GRCh38
rs763707805 Chr.6: 87681698-87681698 on Build GRCh38
rs930188008 Chr.6: 87681625-87681625 on Build GRCh38
rs947846489 Chr.6: 87681827-87681827 on Build GRCh38
rs750847976 Chr.6: 87681748-87681748 on Build GRCh38
rs561882457 Chr.6: 87681557-87681557 on Build GRCh38
rs752307157 Chr.6: 87681592-87681592 on Build GRCh38
rs1053519479 Chr.6: 87681781-87681781 on Build GRCh38
rs781399884 Chr.6: 87681793-87681793 on Build GRCh38
rs754370623 Chr.6: 87681602-87681602 on Build GRCh38
rs906001398 Chr.6: 87681469-87681469 on Build GRCh38
rs906239089 Chr.6: 87681926-87681926 on Build GRCh38
rs1009454995 Chr.6: 87681876-87681876 on Build GRCh38
rs573759898 Chr.6: 87681559-87681559 on Build GRCh38
rs944028378 Chr.6: 87681679-87681679 on Build GRCh38
rs752431667 Chr.6: 87681692-87681692 on Build GRCh38
rs370962803 Chr.6: 87681573-87681573 on Build GRCh38
rs749153357 Chr.6: 87681804-87681804 on Build GRCh38
rs772966634 Chr.6: 87681667-87681667 on Build GRCh38
rs758783280 Chr.6: 87681753-87681753 on Build GRCh38
rs557334415 Chr.6: 87681463-87681463 on Build GRCh38
rs772319870 Chr.6: 87681649-87681649 on Build GRCh38
rs754310451 Chr.6: 87681800-87681800 on Build GRCh38
rs9450770 Chr.6: 87681518-87681518 on Build GRCh38
rs547291561 Chr.6: 87681860-87681860 on Build GRCh38
rs564463375 Chr.6: 87681691-87681691 on Build GRCh38
rs1025305236 Chr.6: 87681923-87681923 on Build GRCh38
rs770627861 Chr.6: 87681675-87681675 on Build GRCh38
rs368094512 Chr.6: 87681663-87681663 on Build GRCh38

View Full Product Details