Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 100516192-100516599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC3
Entrez Gene ID
10973
Gene Name
activating signal cointegrator 1 complex subunit 3
Gene Aliases
ASC1p200, HELIC1, RNAH
UniGene
Hs.486031
Gene Chromosome Location
Chr.6: 100508194-100881372 on Build GRCh38
Transcripts
NM_006828.3 XM_011535394.2 XM_011535395.2 XM_011535396.2 XM_017010205.1 XM_017010206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828676 c.5967T>G p.G1989G Chr.6: 100516288-100516288 on Build GRCh38
1132075 c.5984C>G p.S1995C Chr.6: 100516271-100516271 on Build GRCh38
4663669 c.5968C>T p.R1990W Chr.6: 100516287-100516287 on Build GRCh38
1697681 c.5963_5964GG>AA p.R1988K Chr.6: 100516291-100516292 on Build GRCh38
7113428 c.6003T>G p.H2001Q Chr.6: 100516252-100516252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981441145 Chr.6: 100516287-100516287 on Build GRCh38
rs148099735 Chr.6: 100516342-100516342 on Build GRCh38
rs746173351 Chr.6: 100516335-100516335 on Build GRCh38
rs757380124 Chr.6: 100516320-100516320 on Build GRCh38
rs569508246 Chr.6: 100516454-100516454 on Build GRCh38
rs240780 Chr.6: 100516271-100516271 on Build GRCh38
rs887061253 Chr.6: 100516449-100516449 on Build GRCh38
rs539303132 Chr.6: 100516249-100516249 on Build GRCh38
rs33988845 Chr.6: 100516282-100516282 on Build GRCh38
rs766331817 Chr.6: 100516296-100516296 on Build GRCh38
rs931143539 Chr.6: 100516391-100516391 on Build GRCh38
rs771231178 Chr.6: 100516232-100516232 on Build GRCh38
rs762849129 Chr.6: 100516295-100516295 on Build GRCh38
rs765006481 Chr.6: 100516286-100516286 on Build GRCh38
rs573779798 Chr.6: 100516301-100516301 on Build GRCh38
rs747609990 Chr.6: 100516269-100516269 on Build GRCh38
rs779112446 Chr.6: 100516328-100516328 on Build GRCh38
rs141842488 Chr.6: 100516490-100516490 on Build GRCh38
rs917290128 Chr.6: 100516254-100516254 on Build GRCh38
rs753973276 Chr.6: 100516316-100516316 on Build GRCh38
rs773105470 Chr.6: 100516289-100516289 on Build GRCh38
rs555202785 Chr.6: 100516290-100516290 on Build GRCh38
rs772500713 Chr.6: 100516251-100516251 on Build GRCh38
rs576122470 Chr.6: 100516272-100516272 on Build GRCh38
rs1008959153 Chr.6: 100516353-100516355 on Build GRCh38
rs1019314027 Chr.6: 100516457-100516457 on Build GRCh38
rs779198711 Chr.6: 100516234-100516234 on Build GRCh38
rs240781 Chr.6: 100516315-100516315 on Build GRCh38
rs984353322 Chr.6: 100516576-100516576 on Build GRCh38
rs779701858 Chr.6: 100516419-100516419 on Build GRCh38
rs777971180 Chr.6: 100516218-100516218 on Build GRCh38
rs775988870 Chr.6: 100516260-100516260 on Build GRCh38
rs116759347 Chr.6: 100516392-100516392 on Build GRCh38
rs369040191 Chr.6: 100516231-100516231 on Build GRCh38
rs995943593 Chr.6: 100516489-100516489 on Build GRCh38
rs780464565 Chr.6: 100516363-100516363 on Build GRCh38
rs898555138 Chr.6: 100516387-100516387 on Build GRCh38
rs755586100 Chr.6: 100516383-100516383 on Build GRCh38
rs561085972 Chr.6: 100516570-100516570 on Build GRCh38
rs374214153 Chr.6: 100516294-100516294 on Build GRCh38
rs754980276 Chr.6: 100516306-100516306 on Build GRCh38
rs545340166 Chr.6: 100516471-100516471 on Build GRCh38
rs1044580852 Chr.6: 100516276-100516276 on Build GRCh38
rs543202961 Chr.6: 100516545-100516545 on Build GRCh38
rs899518380 Chr.6: 100516484-100516484 on Build GRCh38
rs563989642 Chr.6: 100516474-100516474 on Build GRCh38

View Full Product Details