Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73682406-73682955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC30
Entrez Gene ID
84277
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
Gene Aliases
WBSCR18
UniGene
Hs.647046
Gene Chromosome Location
Chr.7: 73680918-73683451 on Build GRCh38
Transcripts
NM_032317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3882257 c.532C>T p.R178W Chr.7: 73682892-73682892 on Build GRCh38
747161 c.505C>G p.Q169E Chr.7: 73682919-73682919 on Build GRCh38
5615865 c.598G>C p.G200R Chr.7: 73682826-73682826 on Build GRCh38
3833181 c.497C>A p.A166D Chr.7: 73682927-73682927 on Build GRCh38
3640381 c.610G>A p.E204K Chr.7: 73682814-73682814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961476487 Chr.7: 73682540-73682540 on Build GRCh38
rs1003583624 Chr.7: 73682550-73682550 on Build GRCh38
rs370444492 Chr.7: 73682812-73682812 on Build GRCh38
rs959210087 Chr.7: 73682608-73682608 on Build GRCh38
rs573532722 Chr.7: 73682519-73682519 on Build GRCh38
rs568368328 Chr.7: 73682784-73682784 on Build GRCh38
rs8891 Chr.7: 73682663-73682663 on Build GRCh38
rs781804312 Chr.7: 73682768-73682768 on Build GRCh38
rs13244259 Chr.7: 73682923-73682923 on Build GRCh38
rs1003468683 Chr.7: 73682807-73682807 on Build GRCh38
rs781850653 Chr.7: 73682913-73682913 on Build GRCh38
rs782338163 Chr.7: 73682850-73682850 on Build GRCh38
rs1040133653 Chr.7: 73682716-73682716 on Build GRCh38
rs562570211 Chr.7: 73682632-73682632 on Build GRCh38
rs781833425 Chr.7: 73682830-73682830 on Build GRCh38
rs555076484 Chr.7: 73682490-73682490 on Build GRCh38
rs944461785 Chr.7: 73682670-73682670 on Build GRCh38
rs372985556 Chr.7: 73682701-73682701 on Build GRCh38
rs770045692 Chr.7: 73682610-73682610 on Build GRCh38
rs1015516447 Chr.7: 73682520-73682520 on Build GRCh38
rs782609685 Chr.7: 73682774-73682774 on Build GRCh38
rs371617893 Chr.7: 73682739-73682739 on Build GRCh38
rs781998031 Chr.7: 73682796-73682796 on Build GRCh38
rs782770783 Chr.7: 73682756-73682756 on Build GRCh38
rs782558149 Chr.7: 73682838-73682838 on Build GRCh38
rs782325183 Chr.7: 73682870-73682870 on Build GRCh38
rs782382339 Chr.7: 73682880-73682880 on Build GRCh38
rs1034841389 Chr.7: 73682603-73682603 on Build GRCh38
rs978016076 Chr.7: 73682901-73682901 on Build GRCh38
rs782603180 Chr.7: 73682918-73682918 on Build GRCh38
rs782506661 Chr.7: 73682827-73682827 on Build GRCh38
rs534498812 Chr.7: 73682905-73682905 on Build GRCh38
rs532848960 Chr.7: 73682641-73682641 on Build GRCh38
rs550681883 Chr.7: 73682837-73682837 on Build GRCh38
rs782550393 Chr.7: 73682770-73682770 on Build GRCh38
rs967955806 Chr.7: 73682621-73682621 on Build GRCh38
rs782478459 Chr.7: 73682766-73682766 on Build GRCh38
rs372858734 Chr.7: 73682872-73682872 on Build GRCh38
rs201962971 Chr.7: 73682871-73682871 on Build GRCh38
rs201805928 Chr.7: 73682792-73682792 on Build GRCh38
rs782113420 Chr.7: 73682823-73682823 on Build GRCh38
rs782310475 Chr.7: 73682779-73682779 on Build GRCh38
rs79849491 Chr.7: 73682752-73682752 on Build GRCh38
rs782620253 Chr.7: 73682841-73682841 on Build GRCh38
rs782073520 Chr.7: 73682907-73682907 on Build GRCh38
rs782615015 Chr.7: 73682855-73682855 on Build GRCh38
rs782685455 Chr.7: 73682771-73682771 on Build GRCh38
rs782515545 Chr.7: 73682800-73682800 on Build GRCh38
rs781973239 Chr.7: 73682711-73682711 on Build GRCh38
rs1005969809 Chr.7: 73682504-73682504 on Build GRCh38
rs147002145 Chr.7: 73682822-73682822 on Build GRCh38
rs569400604 Chr.7: 73682846-73682846 on Build GRCh38
rs782183147 Chr.7: 73682861-73682861 on Build GRCh38
rs782781775 Chr.7: 73682827-73682827 on Build GRCh38
rs781916547 Chr.7: 73682811-73682811 on Build GRCh38
rs145908448 Chr.7: 73682914-73682914 on Build GRCh38
rs200098566 Chr.7: 73682717-73682717 on Build GRCh38
rs200600889 Chr.7: 73682705-73682705 on Build GRCh38
rs374274920 Chr.7: 73682758-73682758 on Build GRCh38
rs782439339 Chr.7: 73682920-73682920 on Build GRCh38
rs782283976 Chr.7: 73682780-73682780 on Build GRCh38
rs782348689 Chr.7: 73682706-73682706 on Build GRCh38
rs782246935 Chr.7: 73682772-73682772 on Build GRCh38
rs781869572 Chr.7: 73682721-73682722 on Build GRCh38
rs189100172 Chr.7: 73682461-73682461 on Build GRCh38
rs376974280 Chr.7: 73682707-73682707 on Build GRCh38
rs782704770 Chr.7: 73682767-73682767 on Build GRCh38
rs782589087 Chr.7: 73682692-73682692 on Build GRCh38
rs782791140 Chr.7: 73682741-73682741 on Build GRCh38
rs200043253 Chr.7: 73682932-73682932 on Build GRCh38
rs782368766 Chr.7: 73682794-73682794 on Build GRCh38
rs192814756 Chr.7: 73682607-73682607 on Build GRCh38
rs1041273304 Chr.7: 73682435-73682435 on Build GRCh38
rs782526757 Chr.7: 73682773-73682773 on Build GRCh38
rs201353139 Chr.7: 73682891-73682891 on Build GRCh38
rs901323437 Chr.7: 73682458-73682458 on Build GRCh38
rs990642334 Chr.7: 73682609-73682609 on Build GRCh38
rs782000390 Chr.7: 73682886-73682886 on Build GRCh38
rs1037036120 Chr.7: 73682790-73682790 on Build GRCh38
rs782630153 Chr.7: 73682935-73682935 on Build GRCh38
rs145658326 Chr.7: 73682750-73682750 on Build GRCh38
rs782573050 Chr.7: 73682835-73682835 on Build GRCh38
rs782279189 Chr.7: 73682927-73682927 on Build GRCh38
rs1569062 Chr.7: 73682908-73682908 on Build GRCh38
rs781836501 Chr.7: 73682919-73682919 on Build GRCh38
rs782810760 Chr.7: 73682701-73682701 on Build GRCh38
rs541001286 Chr.7: 73682484-73682484 on Build GRCh38
rs137909145 Chr.7: 73682904-73682904 on Build GRCh38
rs529261927 Chr.7: 73682834-73682834 on Build GRCh38

View Full Product Details