Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183954375-183954916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT2
Entrez Gene ID
23127
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
C1orf17, GLT25D2
UniGene
Hs.387995
Gene Chromosome Location
Chr.1: 183929662-184051612 on Build GRCh38
Transcripts
NM_001303420.1 NM_001303421.1 NM_015101.3 XM_011509332.2 XM_011509334.2 XM_011509335.2 XM_011509336.2 XM_011509337.2 XM_011509338.2 XM_017000750.1 XM_017000751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5888562 c.228G>A p.M76I Chr.1: 183954774-183954774 on Build GRCh38
6060424 c.225G>C p.K75N Chr.1: 183954777-183954777 on Build GRCh38
5888561 c.1017G>A p.M339I Chr.1: 183954774-183954774 on Build GRCh38
4514439 c.974C>T p.P325L Chr.1: 183954817-183954817 on Build GRCh38
6060423 c.1014G>C p.K338N Chr.1: 183954777-183954777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747201570 Chr.1: 183954724-183954724 on Build GRCh38
rs928372324 Chr.1: 183954671-183954671 on Build GRCh38
rs141867153 Chr.1: 183954492-183954492 on Build GRCh38
rs375823749 Chr.1: 183954800-183954800 on Build GRCh38
rs371890638 Chr.1: 183954799-183954799 on Build GRCh38
rs763589154 Chr.1: 183954402-183954402 on Build GRCh38
rs776403169 Chr.1: 183954824-183954824 on Build GRCh38
rs147542828 Chr.1: 183954672-183954672 on Build GRCh38
rs990050998 Chr.1: 183954613-183954613 on Build GRCh38
rs375108147 Chr.1: 183954421-183954421 on Build GRCh38
rs777860348 Chr.1: 183954723-183954723 on Build GRCh38
rs1005694010 Chr.1: 183954505-183954505 on Build GRCh38
rs1029707441 Chr.1: 183954530-183954530 on Build GRCh38
rs114422863 Chr.1: 183954603-183954603 on Build GRCh38
rs116104334 Chr.1: 183954509-183954509 on Build GRCh38
rs184755208 Chr.1: 183954838-183954838 on Build GRCh38
rs957296720 Chr.1: 183954604-183954604 on Build GRCh38
rs552945541 Chr.1: 183954843-183954843 on Build GRCh38
rs936981432 Chr.1: 183954471-183954471 on Build GRCh38
rs1054267275 Chr.1: 183954410-183954410 on Build GRCh38
rs994694482 Chr.1: 183954529-183954529 on Build GRCh38
rs114095532 Chr.1: 183954832-183954832 on Build GRCh38
rs554619315 Chr.1: 183954427-183954427 on Build GRCh38
rs757429561 Chr.1: 183954851-183954851 on Build GRCh38
rs149857645 Chr.1: 183954582-183954582 on Build GRCh38
rs75151947 Chr.1: 183954727-183954727 on Build GRCh38
rs748138630 Chr.1: 183954738-183954738 on Build GRCh38
rs759870668 Chr.1: 183954826-183954826 on Build GRCh38
rs765716895 Chr.1: 183954718-183954718 on Build GRCh38
rs981603392 Chr.1: 183954656-183954656 on Build GRCh38
rs906900561 Chr.1: 183954467-183954467 on Build GRCh38
rs150617278 Chr.1: 183954510-183954510 on Build GRCh38
rs1055791104 Chr.1: 183954485-183954485 on Build GRCh38
rs749316096 Chr.1: 183954627-183954627 on Build GRCh38
rs773393449 Chr.1: 183954432-183954432 on Build GRCh38
rs560956973 Chr.1: 183954669-183954669 on Build GRCh38
rs753250795 Chr.1: 183954720-183954720 on Build GRCh38
rs906514877 Chr.1: 183954618-183954618 on Build GRCh38
rs977503572 Chr.1: 183954431-183954431 on Build GRCh38
rs770484843 Chr.1: 183954816-183954816 on Build GRCh38
rs778884970 Chr.1: 183954881-183954881 on Build GRCh38
rs779835647 Chr.1: 183954726-183954726 on Build GRCh38
rs754790475 Chr.1: 183954878-183954878 on Build GRCh38
rs74767569 Chr.1: 183954428-183954428 on Build GRCh38
rs965621429 Chr.1: 183954456-183954456 on Build GRCh38
rs768969742 Chr.1: 183954695-183954695 on Build GRCh38
rs754156438 Chr.1: 183954712-183954712 on Build GRCh38
rs368173013 Chr.1: 183954629-183954629 on Build GRCh38
rs17570110 Chr.1: 183954572-183954572 on Build GRCh38
rs755436127 Chr.1: 183954714-183954714 on Build GRCh38
rs367634486 Chr.1: 183954759-183954759 on Build GRCh38
rs181809768 Chr.1: 183954833-183954833 on Build GRCh38
rs201373411 Chr.1: 183954876-183954876 on Build GRCh38
rs765628889 Chr.1: 183954830-183954830 on Build GRCh38
rs564699958 Chr.1: 183954729-183954729 on Build GRCh38
rs1045480225 Chr.1: 183954459-183954459 on Build GRCh38
rs780498445 Chr.1: 183954843-183954843 on Build GRCh38
rs534186263 Chr.1: 183954433-183954433 on Build GRCh38
rs937560320 Chr.1: 183954653-183954653 on Build GRCh38
rs975621495 Chr.1: 183954473-183954473 on Build GRCh38
rs926355494 Chr.1: 183954624-183954624 on Build GRCh38

View Full Product Details