Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179393890-179394139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
UniGene
Hs.658505
Gene Chromosome Location
Chr.1: 179365705-179554735 on Build GRCh38
Transcripts
NM_144696.5 NR_073544.1 XM_011509165.1 XM_011509166.2 XM_011509167.2 XM_011509168.1 XM_011509169.1 XM_011509170.1 XM_011509171.1 XM_011509174.1 XM_011509175.1 XM_011509176.1 XM_011509178.2 XM_011509179.2 XM_011509180.1 XM_017000257.1 XM_017000258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5968995 c.960_962delAGA p.E322delE Chr.1: 179393999-179394001 on Build GRCh38
900261 c.994G>T p.E332* Chr.1: 179394033-179394033 on Build GRCh38
4836718 c.870G>C p.R290S Chr.1: 179393909-179393909 on Build GRCh38
6302299 c.864-9G>T p.? Chr.1: 179393894-179393894 on Build GRCh38
6609010 c.943A>G p.M315V Chr.1: 179393982-179393982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202170595 Chr.1: 179393977-179393977 on Build GRCh38
rs370650077 Chr.1: 179394080-179394080 on Build GRCh38
rs568558494 Chr.1: 179394100-179394100 on Build GRCh38
rs752752345 Chr.1: 179394010-179394010 on Build GRCh38
rs753955745 Chr.1: 179393991-179393991 on Build GRCh38
rs561147430 Chr.1: 179393956-179393956 on Build GRCh38
rs532828641 Chr.1: 179394020-179394020 on Build GRCh38
rs759831232 Chr.1: 179394006-179394006 on Build GRCh38
rs766288405 Chr.1: 179393965-179393965 on Build GRCh38
rs148772825 Chr.1: 179394003-179394003 on Build GRCh38
rs77282993 Chr.1: 179394008-179394008 on Build GRCh38
rs771708407 Chr.1: 179394056-179394056 on Build GRCh38
rs768005250 Chr.1: 179393970-179393970 on Build GRCh38
rs781502885 Chr.1: 179393999-179394001 on Build GRCh38
rs753160885 Chr.1: 179393945-179393945 on Build GRCh38
rs778985705 Chr.1: 179393938-179393938 on Build GRCh38
rs747654129 Chr.1: 179394048-179394048 on Build GRCh38
rs747360696 Chr.1: 179393947-179393947 on Build GRCh38
rs541494490 Chr.1: 179393937-179393937 on Build GRCh38
rs778762762 Chr.1: 179394046-179394046 on Build GRCh38
rs139341288 Chr.1: 179393979-179393979 on Build GRCh38
rs748558161 Chr.1: 179394052-179394053 on Build GRCh38
rs369586960 Chr.1: 179393971-179393971 on Build GRCh38
rs1026789611 Chr.1: 179394024-179394024 on Build GRCh38
rs754633857 Chr.1: 179394044-179394044 on Build GRCh38
rs752115906 Chr.1: 179394039-179394039 on Build GRCh38
rs571709171 Chr.1: 179394023-179394023 on Build GRCh38
rs374737211 Chr.1: 179394083-179394083 on Build GRCh38
rs377419373 Chr.1: 179394087-179394087 on Build GRCh38
rs754094622 Chr.1: 179393927-179393927 on Build GRCh38
rs760320063 Chr.1: 179393984-179393984 on Build GRCh38
rs952614902 Chr.1: 179394027-179394027 on Build GRCh38
rs748855561 Chr.1: 179394043-179394043 on Build GRCh38
rs146917473 Chr.1: 179393976-179393976 on Build GRCh38
rs758873487 Chr.1: 179393946-179393946 on Build GRCh38
rs766209621 Chr.1: 179393989-179393989 on Build GRCh38
rs771397143 Chr.1: 179393948-179393948 on Build GRCh38
rs779510365 Chr.1: 179393952-179393952 on Build GRCh38
rs201487374 Chr.1: 179394058-179394058 on Build GRCh38
rs201762163 Chr.1: 179394041-179394041 on Build GRCh38
rs200125774 Chr.1: 179393992-179393992 on Build GRCh38
rs904803787 Chr.1: 179393918-179393918 on Build GRCh38
rs937581534 Chr.1: 179393922-179393922 on Build GRCh38

View Full Product Details