Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212624475-212625073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71A
Entrez Gene ID
149647
Gene Name
family with sequence similarity 71 member A
Gene Aliases
-
UniGene
Hs.129293
Gene Chromosome Location
Chr.1: 212624447-212626778 on Build GRCh38
Transcripts
NM_153606.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5128343 c.88C>T p.R30* Chr.1: 212624965-212624965 on Build GRCh38
5585169 c.15T>A p.F5L Chr.1: 212624892-212624892 on Build GRCh38
1206426 c.89G>A p.R30Q Chr.1: 212624966-212624966 on Build GRCh38
425255 c.111C>T p.Y37Y Chr.1: 212624988-212624988 on Build GRCh38
1206427 c.107A>G p.E36G Chr.1: 212624984-212624984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200565640 Chr.1: 212624966-212624966 on Build GRCh38
rs948514554 Chr.1: 212625003-212625003 on Build GRCh38
rs1054172349 Chr.1: 212624694-212624694 on Build GRCh38
rs548872168 Chr.1: 212624578-212624578 on Build GRCh38
rs772952524 Chr.1: 212625007-212625007 on Build GRCh38
rs7533306 Chr.1: 212624548-212624548 on Build GRCh38
rs77184091 Chr.1: 212624551-212624551 on Build GRCh38
rs760104113 Chr.1: 212624997-212624997 on Build GRCh38
rs17853363 Chr.1: 212624941-212624941 on Build GRCh38
rs28620301 Chr.1: 212624650-212624650 on Build GRCh38
rs1044080007 Chr.1: 212624804-212624804 on Build GRCh38
rs199963724 Chr.1: 212624929-212624929 on Build GRCh38
rs986135908 Chr.1: 212624637-212624637 on Build GRCh38
rs768834193 Chr.1: 212624806-212624806 on Build GRCh38
rs189613224 Chr.1: 212625033-212625033 on Build GRCh38
rs377562239 Chr.1: 212624857-212624857 on Build GRCh38
rs562388020 Chr.1: 212624667-212624667 on Build GRCh38
rs769503977 Chr.1: 212624905-212624905 on Build GRCh38
rs140634697 Chr.1: 212624901-212624901 on Build GRCh38
rs1015440773 Chr.1: 212624550-212624550 on Build GRCh38
rs767847858 Chr.1: 212624852-212624852 on Build GRCh38
rs974046717 Chr.1: 212624527-212624527 on Build GRCh38
rs765362354 Chr.1: 212624961-212624961 on Build GRCh38
rs748984968 Chr.1: 212624680-212624680 on Build GRCh38
rs761147277 Chr.1: 212624856-212624856 on Build GRCh38
rs777545222 Chr.1: 212624625-212624625 on Build GRCh38
rs1045830404 Chr.1: 212624619-212624619 on Build GRCh38
rs762253961 Chr.1: 212624839-212624839 on Build GRCh38
rs756158237 Chr.1: 212625048-212625048 on Build GRCh38
rs1023325314 Chr.1: 212624542-212624542 on Build GRCh38
rs763928601 Chr.1: 212624967-212624967 on Build GRCh38
rs1007411588 Chr.1: 212624760-212624760 on Build GRCh38
rs757220768 Chr.1: 212624971-212624971 on Build GRCh38
rs970245317 Chr.1: 212624838-212624838 on Build GRCh38
rs759629500 Chr.1: 212624951-212624951 on Build GRCh38
rs149508727 Chr.1: 212624994-212624994 on Build GRCh38
rs551458200 Chr.1: 212624992-212624992 on Build GRCh38
rs909144984 Chr.1: 212624525-212624525 on Build GRCh38
rs750379097 Chr.1: 212625034-212625034 on Build GRCh38
rs1041987483 Chr.1: 212624670-212624670 on Build GRCh38
rs143410969 Chr.1: 212624899-212624899 on Build GRCh38
rs1044505992 Chr.1: 212625006-212625006 on Build GRCh38
rs753851923 Chr.1: 212624940-212624940 on Build GRCh38
rs779494352 Chr.1: 212624982-212624982 on Build GRCh38
rs139614117 Chr.1: 212624909-212624909 on Build GRCh38
rs1042017113 Chr.1: 212624960-212624960 on Build GRCh38
rs771373425 Chr.1: 212624993-212624993 on Build GRCh38
rs952166984 Chr.1: 212624636-212624636 on Build GRCh38
rs947536416 Chr.1: 212624931-212624931 on Build GRCh38
rs948679103 Chr.1: 212624503-212624551 on Build GRCh38
rs184461395 Chr.1: 212625002-212625002 on Build GRCh38
rs200833097 Chr.1: 212624858-212624858 on Build GRCh38
rs376265759 Chr.1: 212624976-212624976 on Build GRCh38
rs956228696 Chr.1: 212624979-212624979 on Build GRCh38
rs200700327 Chr.1: 212625008-212625008 on Build GRCh38
rs892866271 Chr.1: 212624736-212624736 on Build GRCh38
rs370646615 Chr.1: 212624983-212624983 on Build GRCh38
rs751300602 Chr.1: 212625009-212625009 on Build GRCh38
rs181910238 Chr.1: 212625051-212625051 on Build GRCh38
rs907103866 Chr.1: 212624545-212624545 on Build GRCh38
rs777485101 Chr.1: 212624897-212624897 on Build GRCh38
rs1016472837 Chr.1: 212624798-212624798 on Build GRCh38
rs906837033 Chr.1: 212624823-212624823 on Build GRCh38
rs953263933 Chr.1: 212624516-212624517 on Build GRCh38
rs142855237 Chr.1: 212624854-212624854 on Build GRCh38
rs771031665 Chr.1: 212624900-212624900 on Build GRCh38
rs934822264 Chr.1: 212624684-212624684 on Build GRCh38
rs202022393 Chr.1: 212624968-212624968 on Build GRCh38
rs768492352 Chr.1: 212625015-212625015 on Build GRCh38
rs976264777 Chr.1: 212624663-212624663 on Build GRCh38
rs190061957 Chr.1: 212624624-212624624 on Build GRCh38
rs201256590 Chr.1: 212624988-212624988 on Build GRCh38
rs147433240 Chr.1: 212624859-212624859 on Build GRCh38
rs199558191 Chr.1: 212625040-212625040 on Build GRCh38
rs1012579066 Chr.1: 212624508-212624508 on Build GRCh38
rs924789523 Chr.1: 212624681-212624681 on Build GRCh38
rs1056336994 Chr.1: 212624661-212624662 on Build GRCh38
rs944996931 Chr.1: 212624653-212624653 on Build GRCh38
rs374436561 Chr.1: 212624937-212624937 on Build GRCh38
rs963603103 Chr.1: 212624526-212624526 on Build GRCh38
rs533334594 Chr.1: 212624989-212624989 on Build GRCh38
rs150185683 Chr.1: 212624918-212624918 on Build GRCh38
rs768238530 Chr.1: 212624987-212624987 on Build GRCh38
rs779118426 Chr.1: 212624876-212624876 on Build GRCh38
rs149868185 Chr.1: 212624877-212624877 on Build GRCh38
rs745401565 Chr.1: 212624902-212624902 on Build GRCh38
rs986357426 Chr.1: 212624517-212624517 on Build GRCh38
rs775317074 Chr.1: 212624965-212624965 on Build GRCh38
rs1026215800 Chr.1: 212625022-212625022 on Build GRCh38
rs1030115373 Chr.1: 212624724-212624724 on Build GRCh38
rs910516157 Chr.1: 212624647-212624647 on Build GRCh38
rs141841205 Chr.1: 212625021-212625021 on Build GRCh38
rs781279141 Chr.1: 212624973-212624973 on Build GRCh38
rs79381401 Chr.1: 212624749-212624749 on Build GRCh38
rs111627662 Chr.1: 212624978-212624978 on Build GRCh38
rs972179759 Chr.1: 212624552-212624552 on Build GRCh38

View Full Product Details