Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222749117-222749699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177B
Entrez Gene ID
400823
Gene Name
family with sequence similarity 177 member B
Gene Aliases
-
UniGene
Hs.697608
Gene Chromosome Location
Chr.1: 222737216-222772193 on Build GRCh38
Transcripts
NM_001324080.1 NM_207468.2 NR_136691.1 XM_017001279.1 XM_017001280.1 XM_017001281.1 XM_017001282.1 XM_017001283.1 XM_017001284.1 XM_017001285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1637754 c.339+1G>A p.? Chr.1: 222749563-222749563 on Build GRCh38
6541804 c.312A>T p.L104F Chr.1: 222749535-222749535 on Build GRCh38
3484009 c.327G>A p.R109R Chr.1: 222749550-222749550 on Build GRCh38
6658954 c.327G>T p.R109S Chr.1: 222749550-222749550 on Build GRCh38
904337 c.321C>A p.F107L Chr.1: 222749544-222749544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568229622 Chr.1: 222749613-222749613 on Build GRCh38
rs755572856 Chr.1: 222749607-222749607 on Build GRCh38
rs780763892 Chr.1: 222749499-222749499 on Build GRCh38
rs765237569 Chr.1: 222749452-222749452 on Build GRCh38
rs529782735 Chr.1: 222749283-222749283 on Build GRCh38
rs957898381 Chr.1: 222749442-222749442 on Build GRCh38
rs540426903 Chr.1: 222749200-222749200 on Build GRCh38
rs758258868 Chr.1: 222749404-222749404 on Build GRCh38
rs1016962799 Chr.1: 222749229-222749230 on Build GRCh38
rs114063646 Chr.1: 222749253-222749253 on Build GRCh38
rs894649132 Chr.1: 222749172-222749172 on Build GRCh38
rs749563194 Chr.1: 222749544-222749544 on Build GRCh38
rs745358743 Chr.1: 222749528-222749528 on Build GRCh38
rs993927951 Chr.1: 222749277-222749277 on Build GRCh38
rs535536977 Chr.1: 222749627-222749627 on Build GRCh38
rs761004278 Chr.1: 222749547-222749547 on Build GRCh38
rs762133837 Chr.1: 222749426-222749426 on Build GRCh38
rs756926579 Chr.1: 222749494-222749494 on Build GRCh38
rs779352097 Chr.1: 222749535-222749535 on Build GRCh38
rs945043010 Chr.1: 222749464-222749464 on Build GRCh38
rs151202441 Chr.1: 222749451-222749451 on Build GRCh38
rs374953254 Chr.1: 222749441-222749441 on Build GRCh38
rs200531384 Chr.1: 222749587-222749587 on Build GRCh38
rs774886487 Chr.1: 222749420-222749420 on Build GRCh38
rs561540048 Chr.1: 222749487-222749487 on Build GRCh38
rs887698041 Chr.1: 222749400-222749400 on Build GRCh38
rs971563880 Chr.1: 222749143-222749143 on Build GRCh38
rs530760022 Chr.1: 222749475-222749475 on Build GRCh38
rs1036255375 Chr.1: 222749661-222749661 on Build GRCh38
rs954746830 Chr.1: 222749330-222749330 on Build GRCh38
rs552438163 Chr.1: 222749491-222749491 on Build GRCh38
rs766579012 Chr.1: 222749450-222749450 on Build GRCh38
rs2068814 Chr.1: 222749659-222749659 on Build GRCh38
rs990102777 Chr.1: 222749222-222749222 on Build GRCh38
rs771164107 Chr.1: 222749548-222749548 on Build GRCh38
rs927647605 Chr.1: 222749503-222749503 on Build GRCh38
rs1001709521 Chr.1: 222749495-222749495 on Build GRCh38
rs772259032 Chr.1: 222749544-222749544 on Build GRCh38
rs750333295 Chr.1: 222749334-222749334 on Build GRCh38
rs757053095 Chr.1: 222749588-222749588 on Build GRCh38
rs371681063 Chr.1: 222749440-222749440 on Build GRCh38
rs765140454 Chr.1: 222749166-222749166 on Build GRCh38
rs201349132 Chr.1: 222749577-222749577 on Build GRCh38
rs771827014 Chr.1: 222749553-222749553 on Build GRCh38
rs188726674 Chr.1: 222749538-222749538 on Build GRCh38
rs141328108 Chr.1: 222749636-222749636 on Build GRCh38
rs773623646 Chr.1: 222749546-222749546 on Build GRCh38
rs750108621 Chr.1: 222749601-222749601 on Build GRCh38
rs927552169 Chr.1: 222749159-222749159 on Build GRCh38
rs373347408 Chr.1: 222749560-222749560 on Build GRCh38
rs759652487 Chr.1: 222749555-222749555 on Build GRCh38
rs762984818 Chr.1: 222749579-222749579 on Build GRCh38
rs769136651 Chr.1: 222749417-222749417 on Build GRCh38
rs765433277 Chr.1: 222749561-222749561 on Build GRCh38
rs1050795521 Chr.1: 222749348-222749348 on Build GRCh38
rs982161291 Chr.1: 222749467-222749470 on Build GRCh38
rs980792463 Chr.1: 222749152-222749152 on Build GRCh38
rs986220230 Chr.1: 222749365-222749365 on Build GRCh38
rs1006469725 Chr.1: 222749175-222749175 on Build GRCh38
rs931821018 Chr.1: 222749342-222749342 on Build GRCh38
rs777036351 Chr.1: 222749552-222749552 on Build GRCh38
rs894637837 Chr.1: 222749572-222749572 on Build GRCh38
rs745446811 Chr.1: 222749414-222749414 on Build GRCh38
rs764001300 Chr.1: 222749580-222749580 on Build GRCh38
rs780868721 Chr.1: 222749590-222749590 on Build GRCh38
rs1007648013 Chr.1: 222749662-222749662 on Build GRCh38
rs779543964 Chr.1: 222749610-222749610 on Build GRCh38
rs774482730 Chr.1: 222749396-222749396 on Build GRCh38
rs569647776 Chr.1: 222749458-222749458 on Build GRCh38
rs957409569 Chr.1: 222749214-222749214 on Build GRCh38
rs923110942 Chr.1: 222749324-222749324 on Build GRCh38
rs946138227 Chr.1: 222749258-222749258 on Build GRCh38
rs758222413 Chr.1: 222749466-222749466 on Build GRCh38
rs780782763 Chr.1: 222749668-222749668 on Build GRCh38
rs941710159 Chr.1: 222749435-222749435 on Build GRCh38
rs756320603 Chr.1: 222749455-222749455 on Build GRCh38
rs769332929 Chr.1: 222749534-222749534 on Build GRCh38
rs777632900 Chr.1: 222749467-222749467 on Build GRCh38
rs771432829 Chr.1: 222749509-222749512 on Build GRCh38

View Full Product Details