Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235550873-235551207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG4
Entrez Gene ID
2786
Gene Name
G protein subunit gamma 4
Gene Aliases
-
UniGene
Hs.159711
Gene Chromosome Location
Chr.1: 235547685-235650754 on Build GRCh38
Transcripts
NM_001098721.1 NM_001098722.1 NM_004485.3 XM_006711761.2 XM_011544167.2

SNP Detail

SNP ID Chromosome Location
rs918105380 Chr.1: 235550948-235550948 on Build GRCh38
rs767248144 Chr.1: 235551156-235551156 on Build GRCh38
rs1050482372 Chr.1: 235551108-235551108 on Build GRCh38
rs944137017 Chr.1: 235551096-235551096 on Build GRCh38
rs1023165048 Chr.1: 235551157-235551157 on Build GRCh38
rs373091149 Chr.1: 235551083-235551083 on Build GRCh38
rs762893699 Chr.1: 235551076-235551076 on Build GRCh38
rs1041251737 Chr.1: 235551109-235551109 on Build GRCh38
rs997115039 Chr.1: 235551111-235551111 on Build GRCh38
rs772634123 Chr.1: 235550968-235550968 on Build GRCh38
rs959749289 Chr.1: 235551150-235551150 on Build GRCh38
rs933563977 Chr.1: 235551098-235551098 on Build GRCh38
rs551505035 Chr.1: 235550969-235550969 on Build GRCh38
rs554340073 Chr.1: 235551087-235551087 on Build GRCh38
rs770545913 Chr.1: 235551116-235551116 on Build GRCh38
rs992702090 Chr.1: 235550924-235550924 on Build GRCh38
rs998484372 Chr.1: 235551134-235551134 on Build GRCh38
rs1052695565 Chr.1: 235551081-235551081 on Build GRCh38
rs879213690 Chr.1: 235551112-235551112 on Build GRCh38
rs931309987 Chr.1: 235551073-235551073 on Build GRCh38
rs775482113 Chr.1: 235551019-235551019 on Build GRCh38
rs147909932 Chr.1: 235550906-235550906 on Build GRCh38
rs935463520 Chr.1: 235551061-235551061 on Build GRCh38
rs993259484 Chr.1: 235551149-235551149 on Build GRCh38
rs968532755 Chr.1: 235551000-235551000 on Build GRCh38
rs748961193 Chr.1: 235551110-235551110 on Build GRCh38
rs925466248 Chr.1: 235551047-235551047 on Build GRCh38
rs113467123 Chr.1: 235551008-235551008 on Build GRCh38
rs978495101 Chr.1: 235551034-235551034 on Build GRCh38
rs920212236 Chr.1: 235551063-235551063 on Build GRCh38
rs45467094 Chr.1: 235551140-235551140 on Build GRCh38
rs577325077 Chr.1: 235551169-235551169 on Build GRCh38
rs534218849 Chr.1: 235551117-235551117 on Build GRCh38
rs566567188 Chr.1: 235550957-235550957 on Build GRCh38

View Full Product Details