Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12196014-12196585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC123
Entrez Gene ID
8872
Gene Name
cell division cycle 123
Gene Aliases
C10orf7, D123
UniGene
Hs.412842
Gene Chromosome Location
Chr.10: 12195962-12250590 on Build GRCh38
Transcripts
NM_006023.2 XM_005252638.3 NM_001321647.1 NM_001321648.1 NM_014142.3

Gene Symbol
NUDT5
Entrez Gene ID
11164
Gene Name
nudix hydrolase 5
Gene Aliases
YSA1, YSA1H, YSAH1
UniGene
Hs.555956
Gene Chromosome Location
Chr.10: 12165328-12196144 on Build GRCh38
Transcripts
NM_006023.2 XM_005252638.3 NM_001321647.1 NM_001321648.1 NM_014142.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012134 c.48G>A p.P16P Chr.10: 12196293-12196293 on Build GRCh38
3806589 c.24C>T p.H8H Chr.10: 12196269-12196269 on Build GRCh38
7134284 c.10G>A p.E4K Chr.10: 12196255-12196255 on Build GRCh38
1733007 c.52T>A p.F18I Chr.10: 12196297-12196297 on Build GRCh38
127583 c.39G>A p.A13A Chr.10: 12196284-12196284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141093124 Chr.10: 12196327-12196330 on Build GRCh38
rs890261015 Chr.10: 12196540-12196540 on Build GRCh38
rs751471870 Chr.10: 12196210-12196210 on Build GRCh38
rs202058655 Chr.10: 12196342-12196342 on Build GRCh38
rs997994594 Chr.10: 12196433-12196433 on Build GRCh38
rs765971286 Chr.10: 12196335-12196335 on Build GRCh38
rs769978738 Chr.10: 12196268-12196268 on Build GRCh38
rs758743330 Chr.10: 12196241-12196241 on Build GRCh38
rs144048523 Chr.10: 12196065-12196065 on Build GRCh38
rs1001856522 Chr.10: 12196075-12196075 on Build GRCh38
rs763646823 Chr.10: 12196306-12196306 on Build GRCh38
rs770053299 Chr.10: 12196216-12196216 on Build GRCh38
rs907601044 Chr.10: 12196494-12196494 on Build GRCh38
rs764952080 Chr.10: 12196259-12196260 on Build GRCh38
rs553708650 Chr.10: 12196359-12196359 on Build GRCh38
rs890302591 Chr.10: 12196354-12196354 on Build GRCh38
rs904599151 Chr.10: 12196564-12196564 on Build GRCh38
rs900409546 Chr.10: 12196245-12196245 on Build GRCh38
rs761832328 Chr.10: 12196302-12196302 on Build GRCh38
rs973061222 Chr.10: 12196516-12196516 on Build GRCh38
rs1030201851 Chr.10: 12196083-12196083 on Build GRCh38
rs1010322033 Chr.10: 12196104-12196104 on Build GRCh38
rs760058625 Chr.10: 12196235-12196236 on Build GRCh38
rs752355230 Chr.10: 12196326-12196327 on Build GRCh38
rs752327673 Chr.10: 12196301-12196301 on Build GRCh38
rs893522005 Chr.10: 12196117-12196117 on Build GRCh38
rs746679242 Chr.10: 12196437-12196437 on Build GRCh38
rs1046620861 Chr.10: 12196042-12196042 on Build GRCh38
rs904629142 Chr.10: 12196403-12196403 on Build GRCh38
rs945882091 Chr.10: 12196549-12196549 on Build GRCh38
rs191671538 Chr.10: 12196260-12196260 on Build GRCh38
rs963699061 Chr.10: 12196507-12196507 on Build GRCh38
rs954955133 Chr.10: 12196088-12196088 on Build GRCh38
rs138632064 Chr.10: 12196408-12196408 on Build GRCh38
rs185382241 Chr.10: 12196498-12196498 on Build GRCh38
rs371415716 Chr.10: 12196228-12196228 on Build GRCh38
rs978959593 Chr.10: 12196141-12196141 on Build GRCh38
rs764504123 Chr.10: 12196348-12196348 on Build GRCh38
rs949117625 Chr.10: 12196214-12196214 on Build GRCh38
rs1053417361 Chr.10: 12196386-12196386 on Build GRCh38
rs968759972 Chr.10: 12196123-12196123 on Build GRCh38
rs1020670108 Chr.10: 12196110-12196110 on Build GRCh38
rs577288946 Chr.10: 12196394-12196395 on Build GRCh38
rs200140568 Chr.10: 12196341-12196341 on Build GRCh38
rs931851573 Chr.10: 12196528-12196528 on Build GRCh38
rs138564420 Chr.10: 12196264-12196264 on Build GRCh38
rs779431973 Chr.10: 12196330-12196330 on Build GRCh38
rs184139810 Chr.10: 12196378-12196378 on Build GRCh38
rs761416072 Chr.10: 12196212-12196212 on Build GRCh38
rs767206968 Chr.10: 12196304-12196304 on Build GRCh38
rs780672584 Chr.10: 12196261-12196261 on Build GRCh38
rs750162429 Chr.10: 12196235-12196235 on Build GRCh38
rs372795785 Chr.10: 12196478-12196478 on Build GRCh38
rs958977689 Chr.10: 12196166-12196166 on Build GRCh38
rs771253432 Chr.10: 12196218-12196219 on Build GRCh38
rs188885120 Chr.10: 12196400-12196400 on Build GRCh38
rs983585217 Chr.10: 12196487-12196487 on Build GRCh38
rs1051693590 Chr.10: 12196531-12196531 on Build GRCh38
rs577659934 Chr.10: 12196324-12196324 on Build GRCh38
rs769642833 Chr.10: 12196197-12196197 on Build GRCh38
rs1037637939 Chr.10: 12196043-12196043 on Build GRCh38
rs921598861 Chr.10: 12196523-12196523 on Build GRCh38
rs1041970517 Chr.10: 12196561-12196561 on Build GRCh38
rs186669832 Chr.10: 12196203-12196203 on Build GRCh38
rs113790347 Chr.10: 12196367-12196367 on Build GRCh38
rs1032438803 Chr.10: 12196116-12196116 on Build GRCh38
rs550785421 Chr.10: 12196439-12196439 on Build GRCh38
rs759440755 Chr.10: 12196514-12196514 on Build GRCh38
rs992949375 Chr.10: 12196179-12196179 on Build GRCh38
rs561230007 Chr.10: 12196545-12196545 on Build GRCh38
rs759008639 Chr.10: 12196414-12196414 on Build GRCh38
rs774171426 Chr.10: 12196281-12196281 on Build GRCh38
rs755827727 Chr.10: 12196237-12196237 on Build GRCh38
rs768428123 Chr.10: 12196280-12196280 on Build GRCh38
rs763958291 Chr.10: 12196208-12196208 on Build GRCh38
rs370213215 Chr.10: 12196339-12196339 on Build GRCh38
rs374983987 Chr.10: 12196207-12196207 on Build GRCh38
rs1046569072 Chr.10: 12196360-12196360 on Build GRCh38
rs749739963 Chr.10: 12196311-12196311 on Build GRCh38
rs531377832 Chr.10: 12196147-12196147 on Build GRCh38
rs141874461 Chr.10: 12196416-12196416 on Build GRCh38
rs779376119 Chr.10: 12196239-12196239 on Build GRCh38
rs200043765 Chr.10: 12196218-12196218 on Build GRCh38
rs993044262 Chr.10: 12196442-12196442 on Build GRCh38
rs995995224 Chr.10: 12196080-12196080 on Build GRCh38
rs953951137 Chr.10: 12196449-12196449 on Build GRCh38
rs192964684 Chr.10: 12196471-12196471 on Build GRCh38
rs565289269 Chr.10: 12196120-12196120 on Build GRCh38
rs919321254 Chr.10: 12196547-12196547 on Build GRCh38
rs777026580 Chr.10: 12196222-12196223 on Build GRCh38
rs1051556057 Chr.10: 12196305-12196305 on Build GRCh38
rs201298397 Chr.10: 12196320-12196320 on Build GRCh38
rs565281996 Chr.10: 12196468-12196468 on Build GRCh38
rs201353624 Chr.10: 12196240-12196240 on Build GRCh38
rs762685799 Chr.10: 12196278-12196278 on Build GRCh38
rs34811487 Chr.10: 12196083-12196084 on Build GRCh38
rs547765286 Chr.10: 12196152-12196152 on Build GRCh38
rs976609047 Chr.10: 12196183-12196183 on Build GRCh38
rs778175088 Chr.10: 12196249-12196249 on Build GRCh38
rs755814515 Chr.10: 12196450-12196451 on Build GRCh38
rs758701338 Chr.10: 12196343-12196343 on Build GRCh38
rs747491256 Chr.10: 12196254-12196254 on Build GRCh38
rs1020742544 Chr.10: 12196142-12196142 on Build GRCh38
rs762580148 Chr.10: 12196303-12196303 on Build GRCh38
rs375416169 Chr.10: 12196272-12196272 on Build GRCh38

View Full Product Details