Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88934947-88935543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTA2
Entrez Gene ID
59
Gene Name
actin, alpha 2, smooth muscle, aorta
Gene Aliases
AAT6, ACTSA, MYMY5
UniGene
Hs.500483
Gene Chromosome Location
Chr.10: 88935074-88991397 on Build GRCh38
Transcripts
NM_001141945.2 NM_001320855.1 NM_001613.2 NR_125373.1

Gene Symbol
ACTA2-AS1
Entrez Gene ID
100132116
Gene Name
ACTA2 antisense RNA 1
Gene Aliases
ZXF1, uc001kfo.1
UniGene
Hs.713690
Gene Chromosome Location
Chr.10: 88932684-88939974 on Build GRCh38
Transcripts
NM_001141945.2 NM_001320855.1 NM_001613.2 NR_125373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868020 c.1073G>A p.W358* Chr.10: 88935284-88935284 on Build GRCh38
3441364 c.1001C>T p.P334L Chr.10: 88935356-88935356 on Build GRCh38
4659089 c.1032T>C p.G344G Chr.10: 88935325-88935325 on Build GRCh38
158657 c.1005G>A p.P335P Chr.10: 88935352-88935352 on Build GRCh38
5050384 c.999C>T p.A333A Chr.10: 88935358-88935358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141343503 Chr.10: 88935520-88935520 on Build GRCh38
rs749013931 Chr.10: 88935382-88935382 on Build GRCh38
rs920817987 Chr.10: 88935061-88935061 on Build GRCh38
rs760778864 Chr.10: 88935357-88935357 on Build GRCh38
rs772067065 Chr.10: 88935350-88935350 on Build GRCh38
rs794728031 Chr.10: 88935367-88935367 on Build GRCh38
rs1021650257 Chr.10: 88935137-88935137 on Build GRCh38
rs752879983 Chr.10: 88935211-88935211 on Build GRCh38
rs111352790 Chr.10: 88935310-88935310 on Build GRCh38
rs543809234 Chr.10: 88935121-88935121 on Build GRCh38
rs926445020 Chr.10: 88935126-88935126 on Build GRCh38
rs148204227 Chr.10: 88935488-88935488 on Build GRCh38
rs920243723 Chr.10: 88935344-88935344 on Build GRCh38
rs774969469 Chr.10: 88935375-88935379 on Build GRCh38
rs947727930 Chr.10: 88935346-88935346 on Build GRCh38
rs367977687 Chr.10: 88935065-88935065 on Build GRCh38
rs368262366 Chr.10: 88935396-88935396 on Build GRCh38
rs775803770 Chr.10: 88935419-88935419 on Build GRCh38
rs757859002 Chr.10: 88935250-88935250 on Build GRCh38
rs78651566 Chr.10: 88935426-88935426 on Build GRCh38
rs769623133 Chr.10: 88935083-88935083 on Build GRCh38
rs779707072 Chr.10: 88935260-88935260 on Build GRCh38
rs3781212 Chr.10: 88935486-88935486 on Build GRCh38
rs775006794 Chr.10: 88935328-88935328 on Build GRCh38
rs886047452 Chr.10: 88935180-88935180 on Build GRCh38
rs761599964 Chr.10: 88935370-88935370 on Build GRCh38
rs149059995 Chr.10: 88935118-88935118 on Build GRCh38
rs571562251 Chr.10: 88934982-88934982 on Build GRCh38
rs1022141809 Chr.10: 88935453-88935453 on Build GRCh38
rs956182522 Chr.10: 88935119-88935119 on Build GRCh38
rs1057518929 Chr.10: 88935354-88935354 on Build GRCh38
rs77086884 Chr.10: 88935012-88935012 on Build GRCh38
rs781108632 Chr.10: 88935173-88935173 on Build GRCh38
rs749601245 Chr.10: 88935418-88935418 on Build GRCh38
rs375064015 Chr.10: 88935349-88935349 on Build GRCh38
rs878854465 Chr.10: 88935225-88935225 on Build GRCh38
rs749867078 Chr.10: 88935248-88935248 on Build GRCh38
rs778887472 Chr.10: 88935245-88935245 on Build GRCh38
rs775742052 Chr.10: 88935352-88935352 on Build GRCh38
rs111317533 Chr.10: 88935369-88935369 on Build GRCh38
rs201489517 Chr.10: 88935287-88935287 on Build GRCh38
rs772860893 Chr.10: 88935521-88935521 on Build GRCh38
rs771562943 Chr.10: 88935327-88935327 on Build GRCh38
rs983390127 Chr.10: 88935066-88935066 on Build GRCh38
rs143005546 Chr.10: 88935195-88935195 on Build GRCh38
rs1062398 Chr.10: 88935239-88935239 on Build GRCh38
rs1010348403 Chr.10: 88935450-88935450 on Build GRCh38
rs794728032 Chr.10: 88935331-88935331 on Build GRCh38
rs994981043 Chr.10: 88935505-88935505 on Build GRCh38
rs372406288 Chr.10: 88935365-88935365 on Build GRCh38
rs1042056574 Chr.10: 88935404-88935404 on Build GRCh38
rs139352781 Chr.10: 88935334-88935334 on Build GRCh38
rs375863298 Chr.10: 88935389-88935389 on Build GRCh38
rs1054117798 Chr.10: 88935379-88935379 on Build GRCh38
rs765771552 Chr.10: 88935393-88935393 on Build GRCh38
rs202020476 Chr.10: 88935207-88935207 on Build GRCh38
rs1062399 Chr.10: 88935226-88935226 on Build GRCh38
rs185128929 Chr.10: 88935403-88935403 on Build GRCh38
rs778257647 Chr.10: 88935270-88935270 on Build GRCh38
rs374643433 Chr.10: 88935438-88935438 on Build GRCh38
rs746668428 Chr.10: 88935264-88935264 on Build GRCh38
rs965025659 Chr.10: 88935036-88935037 on Build GRCh38
rs767716156 Chr.10: 88935196-88935196 on Build GRCh38
rs774562884 Chr.10: 88935186-88935186 on Build GRCh38
rs535799514 Chr.10: 88934997-88934997 on Build GRCh38
rs764351626 Chr.10: 88935359-88935359 on Build GRCh38
rs745310943 Chr.10: 88935202-88935205 on Build GRCh38
rs190827603 Chr.10: 88935475-88935475 on Build GRCh38
rs762441210 Chr.10: 88935208-88935208 on Build GRCh38
rs768058257 Chr.10: 88935219-88935219 on Build GRCh38
rs11202904 Chr.10: 88935283-88935283 on Build GRCh38
rs754695149 Chr.10: 88935265-88935265 on Build GRCh38
rs3781211 Chr.10: 88935508-88935508 on Build GRCh38
rs367887292 Chr.10: 88935213-88935213 on Build GRCh38
rs538985736 Chr.10: 88935417-88935417 on Build GRCh38
rs754661727 Chr.10: 88935401-88935401 on Build GRCh38
rs969027170 Chr.10: 88935178-88935178 on Build GRCh38
rs1009054226 Chr.10: 88935466-88935466 on Build GRCh38
rs113464077 Chr.10: 88935005-88935005 on Build GRCh38
rs7908852 Chr.10: 88935036-88935036 on Build GRCh38
rs180730588 Chr.10: 88935081-88935081 on Build GRCh38
rs765100909 Chr.10: 88935373-88935373 on Build GRCh38
rs794728034 Chr.10: 88935337-88935338 on Build GRCh38
rs750349531 Chr.10: 88935383-88935383 on Build GRCh38
rs3208386 Chr.10: 88935300-88935300 on Build GRCh38
rs538956017 Chr.10: 88935001-88935001 on Build GRCh38
rs757166963 Chr.10: 88935237-88935237 on Build GRCh38
rs1018175426 Chr.10: 88935017-88935017 on Build GRCh38
rs769364282 Chr.10: 88935373-88935373 on Build GRCh38
rs886047451 Chr.10: 88935101-88935101 on Build GRCh38

View Full Product Details