Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113849060-113849585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLRE1A
Entrez Gene ID
9937
Gene Name
DNA cross-link repair 1A
Gene Aliases
PSO2, SNM1, SNM1A
UniGene
Hs.1560
Gene Chromosome Location
Chr.10: 113834724-113854404 on Build GRCh38
Transcripts
NM_001271816.1 NM_014881.4 XM_006718090.1 XM_011540429.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6908033 c.1760C>T p.P587L Chr.10: 113849345-113849345 on Build GRCh38
1703061 c.1696_1697CC>TT p.P566F Chr.10: 113849408-113849409 on Build GRCh38
5521161 c.1831G>A p.E611K Chr.10: 113849274-113849274 on Build GRCh38
1675569 c.1936G>A p.E646K Chr.10: 113849169-113849169 on Build GRCh38
5461807 c.1793A>G p.Q598R Chr.10: 113849312-113849312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764940693 Chr.10: 113849129-113849129 on Build GRCh38
rs377032576 Chr.10: 113849162-113849162 on Build GRCh38
rs751572525 Chr.10: 113849259-113849259 on Build GRCh38
rs757385140 Chr.10: 113849518-113849518 on Build GRCh38
rs776458581 Chr.10: 113849478-113849506 on Build GRCh38
rs761761851 Chr.10: 113849455-113849455 on Build GRCh38
rs749722646 Chr.10: 113849171-113849171 on Build GRCh38
rs761668585 Chr.10: 113849285-113849285 on Build GRCh38
rs772818660 Chr.10: 113849463-113849463 on Build GRCh38
rs747137758 Chr.10: 113849397-113849397 on Build GRCh38
rs764138267 Chr.10: 113849146-113849146 on Build GRCh38
rs779544665 Chr.10: 113849177-113849177 on Build GRCh38
rs772181460 Chr.10: 113849096-113849096 on Build GRCh38
rs138749190 Chr.10: 113849428-113849428 on Build GRCh38
rs17228693 Chr.10: 113849161-113849161 on Build GRCh38
rs200571623 Chr.10: 113849286-113849286 on Build GRCh38
rs771161130 Chr.10: 113849229-113849229 on Build GRCh38
rs144798018 Chr.10: 113849173-113849173 on Build GRCh38
rs777776912 Chr.10: 113849394-113849394 on Build GRCh38
rs17855759 Chr.10: 113849361-113849361 on Build GRCh38
rs376036672 Chr.10: 113849536-113849536 on Build GRCh38
rs755807571 Chr.10: 113849339-113849339 on Build GRCh38
rs765600708 Chr.10: 113849507-113849507 on Build GRCh38
rs748166686 Chr.10: 113849191-113849191 on Build GRCh38
rs987726875 Chr.10: 113849413-113849413 on Build GRCh38
rs754152153 Chr.10: 113849346-113849346 on Build GRCh38
rs774020173 Chr.10: 113849454-113849454 on Build GRCh38
rs570459342 Chr.10: 113849378-113849378 on Build GRCh38
rs200449329 Chr.10: 113849510-113849510 on Build GRCh38
rs575498667 Chr.10: 113849186-113849186 on Build GRCh38
rs893916745 Chr.10: 113849315-113849315 on Build GRCh38
rs1036040801 Chr.10: 113849445-113849445 on Build GRCh38
rs376208580 Chr.10: 113849477-113849477 on Build GRCh38
rs781144369 Chr.10: 113849528-113849528 on Build GRCh38
rs771107901 Chr.10: 113849415-113849415 on Build GRCh38
rs767096421 Chr.10: 113849291-113849291 on Build GRCh38
rs149353240 Chr.10: 113849438-113849438 on Build GRCh38
rs147445182 Chr.10: 113849547-113849547 on Build GRCh38
rs760329839 Chr.10: 113849465-113849465 on Build GRCh38
rs566231940 Chr.10: 113849159-113849159 on Build GRCh38
rs759214264 Chr.10: 113849537-113849537 on Build GRCh38
rs370847347 Chr.10: 113849491-113849491 on Build GRCh38
rs375534266 Chr.10: 113849111-113849111 on Build GRCh38
rs769015000 Chr.10: 113849175-113849175 on Build GRCh38
rs780518186 Chr.10: 113849166-113849166 on Build GRCh38
rs761996874 Chr.10: 113849093-113849093 on Build GRCh38
rs779162731 Chr.10: 113849370-113849370 on Build GRCh38
rs150640935 Chr.10: 113849238-113849238 on Build GRCh38
rs753021054 Chr.10: 113849508-113849508 on Build GRCh38
rs776904276 Chr.10: 113849430-113849430 on Build GRCh38
rs554432161 Chr.10: 113849140-113849140 on Build GRCh38
rs779894331 Chr.10: 113849340-113849340 on Build GRCh38
rs772136993 Chr.10: 113849192-113849192 on Build GRCh38
rs773485376 Chr.10: 113849103-113849103 on Build GRCh38
rs749021441 Chr.10: 113849443-113849443 on Build GRCh38
rs981300245 Chr.10: 113849181-113849181 on Build GRCh38
rs555219952 Chr.10: 113849207-113849207 on Build GRCh38
rs773288594 Chr.10: 113849204-113849204 on Build GRCh38
rs760903290 Chr.10: 113849110-113849110 on Build GRCh38
rs141907465 Chr.10: 113849210-113849210 on Build GRCh38
rs767613365 Chr.10: 113849458-113849458 on Build GRCh38
rs747086391 Chr.10: 113849512-113849512 on Build GRCh38
rs751001765 Chr.10: 113849158-113849158 on Build GRCh38
rs747223502 Chr.10: 113849198-113849198 on Build GRCh38
rs762747479 Chr.10: 113849142-113849142 on Build GRCh38
rs754595563 Chr.10: 113849494-113849494 on Build GRCh38
rs552125051 Chr.10: 113849170-113849170 on Build GRCh38
rs940385031 Chr.10: 113849123-113849123 on Build GRCh38
rs796808132 Chr.10: 113849382-113849382 on Build GRCh38
rs746035540 Chr.10: 113849541-113849541 on Build GRCh38
rs1050209994 Chr.10: 113849420-113849420 on Build GRCh38
rs755431606 Chr.10: 113849359-113849359 on Build GRCh38
rs763931810 Chr.10: 113849252-113849252 on Build GRCh38
rs770766223 Chr.10: 113849302-113849302 on Build GRCh38
rs138658617 Chr.10: 113849208-113849208 on Build GRCh38
rs181214695 Chr.10: 113849509-113849509 on Build GRCh38
rs768440076 Chr.10: 113849453-113849453 on Build GRCh38
rs749976175 Chr.10: 113849324-113849324 on Build GRCh38
rs368906155 Chr.10: 113849234-113849234 on Build GRCh38

View Full Product Details