Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125836573-125837049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCCIP
Entrez Gene ID
56647
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
TOK-1, TOK1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125823535-125853695 on Build GRCh38
Transcripts
NM_016567.3 NM_078469.2 NM_018180.2 XM_017016404.1 XM_017016405.1

Gene Symbol
DHX32
Entrez Gene ID
55760
Gene Name
DEAH-box helicase 32 (putative)
Gene Aliases
DDX32, DHLP1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125836337-125896483 on Build GRCh38
Transcripts
NM_016567.3 NM_078469.2 NM_018180.2 XM_017016404.1 XM_017016405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297048 c.2203G>A p.E735K Chr.10: 125836716-125836716 on Build GRCh38
4012520 c.2143G>A p.D715N Chr.10: 125836776-125836776 on Build GRCh38
5836474 c.2096G>A p.S699N Chr.10: 125836823-125836823 on Build GRCh38
7092721 c.2065T>G p.F689V Chr.10: 125836854-125836854 on Build GRCh38
6023170 c.2195C>T p.T732M Chr.10: 125836724-125836724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755612965 Chr.10: 125836964-125836964 on Build GRCh38
rs752785187 Chr.10: 125837020-125837021 on Build GRCh38
rs765297092 Chr.10: 125836663-125836663 on Build GRCh38
rs767417807 Chr.10: 125836755-125836755 on Build GRCh38
rs1043173759 Chr.10: 125837000-125837000 on Build GRCh38
rs192701781 Chr.10: 125836914-125836914 on Build GRCh38
rs368995976 Chr.10: 125836710-125836710 on Build GRCh38
rs372141280 Chr.10: 125836680-125836680 on Build GRCh38
rs767167630 Chr.10: 125836806-125836806 on Build GRCh38
rs773988110 Chr.10: 125836613-125836613 on Build GRCh38
rs1016957247 Chr.10: 125836878-125836878 on Build GRCh38
rs767375243 Chr.10: 125836678-125836678 on Build GRCh38
rs531809485 Chr.10: 125836776-125836776 on Build GRCh38
rs545907861 Chr.10: 125836607-125836607 on Build GRCh38
rs372661896 Chr.10: 125836787-125836787 on Build GRCh38
rs377298429 Chr.10: 125836862-125836862 on Build GRCh38
rs142010622 Chr.10: 125836732-125836732 on Build GRCh38
rs771329770 Chr.10: 125836891-125836891 on Build GRCh38
rs917709466 Chr.10: 125836803-125836803 on Build GRCh38
rs555335198 Chr.10: 125836985-125836985 on Build GRCh38
rs1014374149 Chr.10: 125836813-125836813 on Build GRCh38
rs773018038 Chr.10: 125836761-125836761 on Build GRCh38
rs921826660 Chr.10: 125836934-125836934 on Build GRCh38
rs747897636 Chr.10: 125836828-125836828 on Build GRCh38
rs752862867 Chr.10: 125836768-125836768 on Build GRCh38
rs748920499 Chr.10: 125836733-125836733 on Build GRCh38
rs755174172 Chr.10: 125836762-125836763 on Build GRCh38
rs913022122 Chr.10: 125836974-125836974 on Build GRCh38
rs374910849 Chr.10: 125836697-125836697 on Build GRCh38
rs748816351 Chr.10: 125836815-125836815 on Build GRCh38
rs375257974 Chr.10: 125836656-125836656 on Build GRCh38
rs976026931 Chr.10: 125836662-125836662 on Build GRCh38
rs542077938 Chr.10: 125836599-125836599 on Build GRCh38
rs757734941 Chr.10: 125836723-125836723 on Build GRCh38
rs527851917 Chr.10: 125836605-125836605 on Build GRCh38
rs763866774 Chr.10: 125836657-125836657 on Build GRCh38
rs778249535 Chr.10: 125836702-125836702 on Build GRCh38
rs548053605 Chr.10: 125837020-125837020 on Build GRCh38
rs1006012583 Chr.10: 125836866-125836866 on Build GRCh38
rs199520716 Chr.10: 125836730-125836730 on Build GRCh38
rs781672078 Chr.10: 125836780-125836780 on Build GRCh38
rs761387659 Chr.10: 125836766-125836766 on Build GRCh38
rs771750217 Chr.10: 125836829-125836829 on Build GRCh38
rs954564580 Chr.10: 125836946-125836946 on Build GRCh38
rs745318505 Chr.10: 125836728-125836728 on Build GRCh38
rs751893829 Chr.10: 125836777-125836777 on Build GRCh38
rs964141130 Chr.10: 125836623-125836623 on Build GRCh38
rs931201763 Chr.10: 125836859-125836863 on Build GRCh38
rs138878584 Chr.10: 125836724-125836724 on Build GRCh38
rs941963264 Chr.10: 125836608-125836608 on Build GRCh38
rs772571642 Chr.10: 125836743-125836743 on Build GRCh38
rs72830596 Chr.10: 125837010-125837010 on Build GRCh38
rs974635581 Chr.10: 125836673-125836673 on Build GRCh38
rs560318203 Chr.10: 125836600-125836600 on Build GRCh38
rs763140267 Chr.10: 125836881-125836881 on Build GRCh38
rs760722649 Chr.10: 125836762-125836762 on Build GRCh38
rs765384472 Chr.10: 125836765-125836765 on Build GRCh38
rs35941699 Chr.10: 125836703-125836703 on Build GRCh38
rs768237004 Chr.10: 125836819-125836819 on Build GRCh38
rs936991459 Chr.10: 125837017-125837017 on Build GRCh38
rs760761419 Chr.10: 125836859-125836862 on Build GRCh38
rs761768015 Chr.10: 125836669-125836669 on Build GRCh38
rs974688124 Chr.10: 125836928-125836928 on Build GRCh38
rs898397383 Chr.10: 125836845-125836845 on Build GRCh38
rs758741108 Chr.10: 125836698-125836698 on Build GRCh38
rs150831914 Chr.10: 125836737-125836737 on Build GRCh38
rs138137798 Chr.10: 125836791-125836791 on Build GRCh38
rs15281 Chr.10: 125836679-125836679 on Build GRCh38
rs771021543 Chr.10: 125836838-125836838 on Build GRCh38
rs34866812 Chr.10: 125836825-125836825 on Build GRCh38
rs773108161 Chr.10: 125836836-125836836 on Build GRCh38
rs984131005 Chr.10: 125836606-125836606 on Build GRCh38
rs774144111 Chr.10: 125836744-125836744 on Build GRCh38
rs145358444 Chr.10: 125836963-125836963 on Build GRCh38
rs761523347 Chr.10: 125836753-125836753 on Build GRCh38

View Full Product Details