Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14966946-14967487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCA
Entrez Gene ID
796
Gene Name
calcitonin related polypeptide alpha
Gene Aliases
CALC1, CGRP, CGRP-I, CGRP1, CT, KC, PCT
UniGene
Hs.37058
Gene Chromosome Location
Chr.11: 14966668-14972361 on Build GRCh38
Transcripts
NM_001033953.2 NR_125898.1 XM_017018283.1 XM_017018284.1

SNP Detail

SNP ID Chromosome Location
rs782136764 Chr.11: 14967239-14967239 on Build GRCh38
rs574014704 Chr.11: 14967303-14967303 on Build GRCh38
rs782713493 Chr.11: 14967460-14967460 on Build GRCh38
rs995766683 Chr.11: 14967215-14967215 on Build GRCh38
rs117456639 Chr.11: 14966999-14966999 on Build GRCh38
rs534395483 Chr.11: 14967146-14967146 on Build GRCh38
rs782764528 Chr.11: 14967242-14967242 on Build GRCh38
rs980764953 Chr.11: 14967395-14967395 on Build GRCh38
rs545365122 Chr.11: 14966989-14966989 on Build GRCh38
rs1038591742 Chr.11: 14967173-14967173 on Build GRCh38
rs35096144 Chr.11: 14967275-14967275 on Build GRCh38
rs989822227 Chr.11: 14967290-14967290 on Build GRCh38
rs1028735665 Chr.11: 14967231-14967231 on Build GRCh38
rs954460836 Chr.11: 14967236-14967236 on Build GRCh38
rs5242 Chr.11: 14967452-14967452 on Build GRCh38
rs939539128 Chr.11: 14967140-14967140 on Build GRCh38
rs887394313 Chr.11: 14967161-14967161 on Build GRCh38
rs555849992 Chr.11: 14967291-14967291 on Build GRCh38
rs190911508 Chr.11: 14967211-14967211 on Build GRCh38
rs1047655438 Chr.11: 14967153-14967153 on Build GRCh38
rs781926828 Chr.11: 14967046-14967046 on Build GRCh38
rs150481800 Chr.11: 14966994-14966994 on Build GRCh38
rs5243 Chr.11: 14967167-14967167 on Build GRCh38
rs540184879 Chr.11: 14967202-14967202 on Build GRCh38
rs782039756 Chr.11: 14967149-14967149 on Build GRCh38
rs187521976 Chr.11: 14967039-14967039 on Build GRCh38
rs781881627 Chr.11: 14967185-14967185 on Build GRCh38
rs928025287 Chr.11: 14967065-14967065 on Build GRCh38
rs529506549 Chr.11: 14967003-14967003 on Build GRCh38
rs776627405 Chr.11: 14967459-14967459 on Build GRCh38
rs570039297 Chr.11: 14967264-14967264 on Build GRCh38
rs782100310 Chr.11: 14966985-14966985 on Build GRCh38
rs948013246 Chr.11: 14967007-14967007 on Build GRCh38
rs928056285 Chr.11: 14967413-14967413 on Build GRCh38
rs1045175572 Chr.11: 14967040-14967040 on Build GRCh38
rs56147952 Chr.11: 14967085-14967085 on Build GRCh38
rs939403413 Chr.11: 14967439-14967439 on Build GRCh38

View Full Product Details