Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31431877-31432443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
UniGene
Hs.187269
Gene Chromosome Location
Chr.11: 31369829-31432835 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1 NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

Gene Symbol
IMMP1L
Entrez Gene ID
196294
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
IMMP1, IMP1, IMP1-LIKE
UniGene
Hs.502223
Gene Chromosome Location
Chr.11: 31432399-31509644 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1 NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

SNP Detail

SNP ID Chromosome Location
rs571901692 Chr.11: 31432381-31432381 on Build GRCh38
rs1030874383 Chr.11: 31432314-31432315 on Build GRCh38
rs989563375 Chr.11: 31432024-31432025 on Build GRCh38
rs568541397 Chr.11: 31432205-31432205 on Build GRCh38
rs74622402 Chr.11: 31432141-31432141 on Build GRCh38
rs553932330 Chr.11: 31432267-31432267 on Build GRCh38
rs551492344 Chr.11: 31431937-31431937 on Build GRCh38
rs977176147 Chr.11: 31432055-31432055 on Build GRCh38
rs956349668 Chr.11: 31431923-31431923 on Build GRCh38
rs535906988 Chr.11: 31432388-31432388 on Build GRCh38
rs542619704 Chr.11: 31432413-31432413 on Build GRCh38
rs1024349378 Chr.11: 31432325-31432325 on Build GRCh38
rs964106079 Chr.11: 31432037-31432037 on Build GRCh38
rs534219350 Chr.11: 31432005-31432005 on Build GRCh38
rs1011144487 Chr.11: 31432336-31432336 on Build GRCh38
rs774865053 Chr.11: 31432195-31432195 on Build GRCh38
rs980536395 Chr.11: 31431995-31431995 on Build GRCh38
rs963321121 Chr.11: 31432103-31432103 on Build GRCh38
rs893674892 Chr.11: 31432323-31432323 on Build GRCh38
rs987237613 Chr.11: 31432180-31432180 on Build GRCh38
rs1000644461 Chr.11: 31431912-31431912 on Build GRCh38
rs762633201 Chr.11: 31432254-31432254 on Build GRCh38
rs1018768077 Chr.11: 31432035-31432035 on Build GRCh38
rs943552029 Chr.11: 31432198-31432198 on Build GRCh38
rs957828834 Chr.11: 31432004-31432004 on Build GRCh38
rs1037483405 Chr.11: 31432179-31432179 on Build GRCh38
rs922598235 Chr.11: 31432069-31432069 on Build GRCh38
rs1018083108 Chr.11: 31432372-31432372 on Build GRCh38
rs1030801957 Chr.11: 31431977-31431977 on Build GRCh38
rs936255371 Chr.11: 31432237-31432237 on Build GRCh38
rs999403759 Chr.11: 31432290-31432290 on Build GRCh38
rs373337947 Chr.11: 31431938-31431938 on Build GRCh38
rs770823738 Chr.11: 31431993-31431993 on Build GRCh38
rs73467223 Chr.11: 31431921-31431921 on Build GRCh38
rs1034045715 Chr.11: 31432070-31432070 on Build GRCh38
rs73467225 Chr.11: 31431948-31431948 on Build GRCh38
rs577939220 Chr.11: 31432226-31432226 on Build GRCh38
rs74481088 Chr.11: 31432240-31432240 on Build GRCh38
rs1033498723 Chr.11: 31432407-31432407 on Build GRCh38
rs1024323203 Chr.11: 31431954-31431954 on Build GRCh38
rs139678308 Chr.11: 31432172-31432172 on Build GRCh38
rs891815807 Chr.11: 31432246-31432246 on Build GRCh38
rs773534978 Chr.11: 31432185-31432185 on Build GRCh38
rs188411167 Chr.11: 31432153-31432153 on Build GRCh38
rs982992072 Chr.11: 31432129-31432129 on Build GRCh38
rs1039219064 Chr.11: 31432213-31432213 on Build GRCh38
rs894844062 Chr.11: 31431898-31431898 on Build GRCh38
rs970535652 Chr.11: 31431991-31431991 on Build GRCh38
rs111706082 Chr.11: 31432012-31432012 on Build GRCh38
rs150617237 Chr.11: 31432105-31432105 on Build GRCh38
rs1012311114 Chr.11: 31431900-31431900 on Build GRCh38
rs16922168 Chr.11: 31432191-31432191 on Build GRCh38
rs776678478 Chr.11: 31432136-31432136 on Build GRCh38

View Full Product Details