Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36594103-36594671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf74
Entrez Gene ID
119710
Gene Name
chromosome 11 open reading frame 74
Gene Aliases
HEPIS
UniGene
Hs.159376
Gene Chromosome Location
Chr.11: 36594245-36659291 on Build GRCh38
Transcripts
NM_001276722.1 NM_001276723.1 NM_001276725.1 NM_001276726.1 NM_001276727.1 NM_138787.3 NR_077243.1 XM_011519885.2 NM_000536.3 NM_001243785.1 NM_001243786.1

Gene Symbol
RAG2
Entrez Gene ID
5897
Gene Name
recombination activating gene 2
Gene Aliases
RAG-2
UniGene
Hs.714519
Gene Chromosome Location
Chr.11: 36591943-36598279 on Build GRCh38
Transcripts
NM_001276722.1 NM_001276723.1 NM_001276725.1 NM_001276726.1 NM_001276727.1 NM_138787.3 NR_077243.1 XM_011519885.2 NM_000536.3 NM_001243785.1 NM_001243786.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623773 c.30T>C p.N10N Chr.11: 36594139-36594139 on Build GRCh38
3979607 c.48G>T p.Q16H Chr.11: 36594121-36594121 on Build GRCh38
1289551 c.3G>A p.M1I Chr.11: 36594166-36594166 on Build GRCh38
328290 c.22G>A p.V8I Chr.11: 36594147-36594147 on Build GRCh38
6279112 c.49C>A p.P17T Chr.11: 36594120-36594120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011560887 Chr.11: 36594560-36594560 on Build GRCh38
rs934947896 Chr.11: 36594536-36594536 on Build GRCh38
rs908902943 Chr.11: 36594412-36594412 on Build GRCh38
rs149524940 Chr.11: 36594409-36594409 on Build GRCh38
rs544520901 Chr.11: 36594415-36594415 on Build GRCh38
rs548845589 Chr.11: 36594589-36594589 on Build GRCh38
rs972161006 Chr.11: 36594331-36594331 on Build GRCh38
rs3740957 Chr.11: 36594562-36594562 on Build GRCh38
rs1051690453 Chr.11: 36594381-36594381 on Build GRCh38
rs556083127 Chr.11: 36594383-36594383 on Build GRCh38
rs766352307 Chr.11: 36594183-36594183 on Build GRCh38
rs146584017 Chr.11: 36594134-36594134 on Build GRCh38
rs370164311 Chr.11: 36594408-36594408 on Build GRCh38
rs972592602 Chr.11: 36594582-36594582 on Build GRCh38
rs1031785070 Chr.11: 36594202-36594202 on Build GRCh38
rs1052868623 Chr.11: 36594448-36594448 on Build GRCh38
rs139508378 Chr.11: 36594527-36594527 on Build GRCh38
rs375822686 Chr.11: 36594379-36594379 on Build GRCh38
rs577304779 Chr.11: 36594184-36594184 on Build GRCh38
rs750499687 Chr.11: 36594148-36594148 on Build GRCh38
rs753764938 Chr.11: 36594130-36594130 on Build GRCh38
rs780060712 Chr.11: 36594137-36594137 on Build GRCh38
rs1017689956 Chr.11: 36594260-36594260 on Build GRCh38
rs150762709 Chr.11: 36594147-36594147 on Build GRCh38
rs754870165 Chr.11: 36594319-36594319 on Build GRCh38
rs773712265 Chr.11: 36594213-36594213 on Build GRCh38
rs975892505 Chr.11: 36594298-36594298 on Build GRCh38
rs376003428 Chr.11: 36594485-36594485 on Build GRCh38
rs758700459 Chr.11: 36594229-36594229 on Build GRCh38
rs1030206921 Chr.11: 36594627-36594627 on Build GRCh38
rs559735360 Chr.11: 36594543-36594543 on Build GRCh38
rs373777467 Chr.11: 36594324-36594324 on Build GRCh38
rs919290967 Chr.11: 36594352-36594352 on Build GRCh38
rs377614322 Chr.11: 36594506-36594506 on Build GRCh38
rs895503827 Chr.11: 36594467-36594467 on Build GRCh38
rs781432309 Chr.11: 36594170-36594170 on Build GRCh38
rs138100839 Chr.11: 36594242-36594242 on Build GRCh38
rs753715989 Chr.11: 36594236-36594236 on Build GRCh38
rs144684962 Chr.11: 36594410-36594410 on Build GRCh38
rs751877215 Chr.11: 36594417-36594417 on Build GRCh38
rs537737027 Chr.11: 36594297-36594297 on Build GRCh38
rs143415103 Chr.11: 36594155-36594155 on Build GRCh38
rs535966949 Chr.11: 36594264-36594264 on Build GRCh38
rs997351252 Chr.11: 36594626-36594626 on Build GRCh38
rs954878109 Chr.11: 36594636-36594636 on Build GRCh38
rs904345209 Chr.11: 36594414-36594414 on Build GRCh38
rs969534830 Chr.11: 36594265-36594265 on Build GRCh38
rs564205435 Chr.11: 36594473-36594473 on Build GRCh38
rs866938503 Chr.11: 36594613-36594613 on Build GRCh38
rs893156759 Chr.11: 36594558-36594558 on Build GRCh38
rs1025171350 Chr.11: 36594246-36594246 on Build GRCh38
rs116109232 Chr.11: 36594508-36594508 on Build GRCh38
rs1045843211 Chr.11: 36594520-36594520 on Build GRCh38
rs533857801 Chr.11: 36594268-36594268 on Build GRCh38
rs553405914 Chr.11: 36594332-36594332 on Build GRCh38

View Full Product Details