Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34642485-34642968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHF
Entrez Gene ID
26298
Gene Name
ETS homologous factor
Gene Aliases
ESE3, ESE3B, ESEJ
UniGene
Hs.653859
Gene Chromosome Location
Chr.11: 34621041-34663288 on Build GRCh38
Transcripts
NM_001206615.1 NM_001206616.1 NM_012153.5 XM_005252857.1 XM_005252860.1 XM_005252861.1 XM_005252862.1 XM_011519983.1 XM_011519984.1 XM_011519985.1 XM_017017534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122795 c.3G>A p.M1I Chr.11: 34642633-34642633 on Build GRCh38
5486846 c.34A>C p.N12H Chr.11: 34642664-34642664 on Build GRCh38
4169088 c.2T>C p.M1T Chr.11: 34642632-34642632 on Build GRCh38
428942 c.37C>G p.P13A Chr.11: 34642667-34642667 on Build GRCh38
6068504 c.14G>C p.G5A Chr.11: 34642644-34642644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749238156 Chr.11: 34642775-34642775 on Build GRCh38
rs773194156 Chr.11: 34642774-34642774 on Build GRCh38
rs946292185 Chr.11: 34642678-34642678 on Build GRCh38
rs960443985 Chr.11: 34642834-34642834 on Build GRCh38
rs74370086 Chr.11: 34642874-34642874 on Build GRCh38
rs759554199 Chr.11: 34642808-34642808 on Build GRCh38
rs768559714 Chr.11: 34642670-34642670 on Build GRCh38
rs763642062 Chr.11: 34642880-34642880 on Build GRCh38
rs375755413 Chr.11: 34642932-34642932 on Build GRCh38
rs769491642 Chr.11: 34642773-34642773 on Build GRCh38
rs374539702 Chr.11: 34642621-34642621 on Build GRCh38
rs192520441 Chr.11: 34642929-34642929 on Build GRCh38
rs1057112256 Chr.11: 34642593-34642593 on Build GRCh38
rs972596170 Chr.11: 34642505-34642505 on Build GRCh38
rs902193299 Chr.11: 34642798-34642798 on Build GRCh38
rs759606089 Chr.11: 34642693-34642693 on Build GRCh38
rs746638583 Chr.11: 34642759-34642759 on Build GRCh38
rs866026748 Chr.11: 34642744-34642744 on Build GRCh38
rs952354934 Chr.11: 34642892-34642892 on Build GRCh38
rs371616522 Chr.11: 34642753-34642753 on Build GRCh38
rs764407036 Chr.11: 34642725-34642725 on Build GRCh38
rs370092846 Chr.11: 34642667-34642667 on Build GRCh38
rs960871088 Chr.11: 34642592-34642592 on Build GRCh38
rs751091113 Chr.11: 34642888-34642888 on Build GRCh38
rs757858841 Chr.11: 34642624-34642624 on Build GRCh38
rs940678851 Chr.11: 34642738-34642738 on Build GRCh38
rs748071518 Chr.11: 34642679-34642679 on Build GRCh38
rs151148136 Chr.11: 34642884-34642884 on Build GRCh38
rs200637887 Chr.11: 34642533-34642534 on Build GRCh38
rs142958059 Chr.11: 34642719-34642719 on Build GRCh38
rs377644917 Chr.11: 34642602-34642602 on Build GRCh38
rs999169071 Chr.11: 34642815-34642815 on Build GRCh38
rs1012404849 Chr.11: 34642852-34642852 on Build GRCh38
rs1040873381 Chr.11: 34642745-34642745 on Build GRCh38
rs999155562 Chr.11: 34642863-34642863 on Build GRCh38
rs12721520 Chr.11: 34642771-34642771 on Build GRCh38
rs536235269 Chr.11: 34642512-34642512 on Build GRCh38
rs377054022 Chr.11: 34642622-34642622 on Build GRCh38
rs750897556 Chr.11: 34642747-34642747 on Build GRCh38
rs769818474 Chr.11: 34642687-34642687 on Build GRCh38
rs113397673 Chr.11: 34642590-34642590 on Build GRCh38
rs556415497 Chr.11: 34642518-34642518 on Build GRCh38
rs927331369 Chr.11: 34642575-34642575 on Build GRCh38
rs538728211 Chr.11: 34642576-34642576 on Build GRCh38
rs776617421 Chr.11: 34642671-34642671 on Build GRCh38
rs367875067 Chr.11: 34642740-34642740 on Build GRCh38
rs577868127 Chr.11: 34642669-34642669 on Build GRCh38
rs1008348791 Chr.11: 34642823-34642823 on Build GRCh38
rs371408038 Chr.11: 34642617-34642617 on Build GRCh38
rs879660563 Chr.11: 34642543-34642543 on Build GRCh38
rs779490064 Chr.11: 34642756-34642756 on Build GRCh38
rs572851230 Chr.11: 34642905-34642905 on Build GRCh38
rs754161535 Chr.11: 34642727-34642727 on Build GRCh38
rs1019269484 Chr.11: 34642871-34642871 on Build GRCh38
rs12721522 Chr.11: 34642566-34642566 on Build GRCh38
rs757641129 Chr.11: 34642737-34642737 on Build GRCh38
rs985464840 Chr.11: 34642893-34642893 on Build GRCh38
rs994048431 Chr.11: 34642886-34642886 on Build GRCh38
rs531459124 Chr.11: 34642881-34642881 on Build GRCh38
rs1043419752 Chr.11: 34642788-34642788 on Build GRCh38
rs779692557 Chr.11: 34642639-34642639 on Build GRCh38
rs115122250 Chr.11: 34642816-34642816 on Build GRCh38
rs573721762 Chr.11: 34642787-34642787 on Build GRCh38
rs892080024 Chr.11: 34642637-34642637 on Build GRCh38
rs747907192 Chr.11: 34642772-34642772 on Build GRCh38
rs754718823 Chr.11: 34642766-34642766 on Build GRCh38
rs976070296 Chr.11: 34642940-34642940 on Build GRCh38
rs773048565 Chr.11: 34642692-34642692 on Build GRCh38
rs146140523 Chr.11: 34642720-34642720 on Build GRCh38

View Full Product Details