Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10015480-10016028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC12B
Entrez Gene ID
387837
Gene Name
C-type lectin domain family 12 member B
Gene Aliases
UNQ5782
UniGene
Hs.127937
Gene Chromosome Location
Chr.12: 10006138-10018800 on Build GRCh38
Transcripts
NM_001129998.2 NM_001319241.1 NM_001319242.1 NM_205852.3 NR_135049.1 XM_011520658.2 XM_011520663.2 XM_017019295.1 NR_120484.1

Gene Symbol
LOC102724020
Entrez Gene ID
102724020
Gene Name
uncharacterized LOC102724020
Gene Aliases
-
UniGene
Hs.737731
Gene Chromosome Location
Chr.12: 10015352-10030606 on Build GRCh38
Transcripts
NM_001129998.2 NM_001319241.1 NM_001319242.1 NM_205852.3 NR_135049.1 XM_011520658.2 XM_011520663.2 XM_017019295.1 NR_120484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2063197 c.638G>T p.R213I Chr.12: 10015685-10015685 on Build GRCh38
2063189 c.625G>C p.D209H Chr.12: 10015672-10015672 on Build GRCh38
6070655 c.650G>T p.W217L Chr.12: 10015697-10015697 on Build GRCh38
6070654 c.650G>T p.W217L Chr.12: 10015697-10015697 on Build GRCh38
1492768 c.649T>C p.W217R Chr.12: 10015696-10015696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902835885 Chr.12: 10015548-10015548 on Build GRCh38
rs935600421 Chr.12: 10015564-10015564 on Build GRCh38
rs147703765 Chr.12: 10015731-10015731 on Build GRCh38
rs752841305 Chr.12: 10015576-10015576 on Build GRCh38
rs200102840 Chr.12: 10015626-10015626 on Build GRCh38
rs962025721 Chr.12: 10015650-10015650 on Build GRCh38
rs112220175 Chr.12: 10015563-10015563 on Build GRCh38
rs577474028 Chr.12: 10015579-10015579 on Build GRCh38
rs563573983 Chr.12: 10015788-10015788 on Build GRCh38
rs772987648 Chr.12: 10015637-10015637 on Build GRCh38
rs771673041 Chr.12: 10015714-10015714 on Build GRCh38
rs142428871 Chr.12: 10015736-10015736 on Build GRCh38
rs1045655568 Chr.12: 10015986-10015986 on Build GRCh38
rs968458521 Chr.12: 10015846-10015846 on Build GRCh38
rs117936789 Chr.12: 10015960-10015960 on Build GRCh38
rs753661436 Chr.12: 10015646-10015646 on Build GRCh38
rs1037033610 Chr.12: 10015868-10015868 on Build GRCh38
rs528908804 Chr.12: 10015684-10015684 on Build GRCh38
rs753833584 Chr.12: 10015570-10015570 on Build GRCh38
rs897222176 Chr.12: 10015558-10015558 on Build GRCh38
rs200596115 Chr.12: 10015624-10015624 on Build GRCh38
rs748018777 Chr.12: 10015599-10015599 on Build GRCh38
rs755031891 Chr.12: 10015572-10015572 on Build GRCh38
rs953273731 Chr.12: 10015728-10015728 on Build GRCh38
rs977113921 Chr.12: 10015869-10015869 on Build GRCh38
rs1007361934 Chr.12: 10015958-10015958 on Build GRCh38
rs949695155 Chr.12: 10015857-10015859 on Build GRCh38
rs771479373 Chr.12: 10015585-10015585 on Build GRCh38
rs766214632 Chr.12: 10015644-10015644 on Build GRCh38
rs376861604 Chr.12: 10015692-10015692 on Build GRCh38
rs755844539 Chr.12: 10015602-10015603 on Build GRCh38
rs1020583867 Chr.12: 10015991-10015991 on Build GRCh38
rs778058731 Chr.12: 10015580-10015580 on Build GRCh38
rs772832018 Chr.12: 10015727-10015727 on Build GRCh38
rs896452591 Chr.12: 10015517-10015519 on Build GRCh38
rs370229418 Chr.12: 10015732-10015732 on Build GRCh38
rs979182453 Chr.12: 10015723-10015723 on Build GRCh38
rs751877923 Chr.12: 10015675-10015675 on Build GRCh38
rs764274926 Chr.12: 10015754-10015754 on Build GRCh38
rs370778199 Chr.12: 10015582-10015582 on Build GRCh38
rs531439346 Chr.12: 10015743-10015743 on Build GRCh38
rs1048467394 Chr.12: 10015948-10015948 on Build GRCh38
rs760453499 Chr.12: 10015639-10015639 on Build GRCh38
rs766302345 Chr.12: 10015565-10015565 on Build GRCh38
rs765230820 Chr.12: 10015654-10015654 on Build GRCh38
rs777842910 Chr.12: 10015669-10015669 on Build GRCh38
rs10845019 Chr.12: 10015964-10015964 on Build GRCh38
rs151305465 Chr.12: 10015689-10015689 on Build GRCh38
rs938333507 Chr.12: 10015917-10015917 on Build GRCh38
rs757664427 Chr.12: 10015676-10015676 on Build GRCh38
rs773686863 Chr.12: 10015613-10015613 on Build GRCh38
rs770455435 Chr.12: 10015608-10015608 on Build GRCh38
rs757506410 Chr.12: 10015787-10015787 on Build GRCh38
rs147064442 Chr.12: 10015709-10015709 on Build GRCh38
rs924311814 Chr.12: 10015913-10015913 on Build GRCh38
rs887588910 Chr.12: 10015952-10015953 on Build GRCh38
rs761402142 Chr.12: 10015622-10015622 on Build GRCh38
rs780990478 Chr.12: 10015593-10015594 on Build GRCh38
rs781758990 Chr.12: 10015594-10015594 on Build GRCh38
rs118186444 Chr.12: 10015994-10015994 on Build GRCh38
rs561520318 Chr.12: 10015981-10015981 on Build GRCh38
rs918164327 Chr.12: 10015848-10015848 on Build GRCh38
rs780617394 Chr.12: 10015696-10015696 on Build GRCh38
rs373680539 Chr.12: 10015647-10015647 on Build GRCh38
rs760365286 Chr.12: 10015730-10015730 on Build GRCh38
rs771560100 Chr.12: 10015635-10015635 on Build GRCh38
rs759327362 Chr.12: 10015733-10015733 on Build GRCh38
rs533664439 Chr.12: 10015983-10015983 on Build GRCh38
rs762917656 Chr.12: 10015747-10015747 on Build GRCh38
rs1006338643 Chr.12: 10016002-10016002 on Build GRCh38
rs900144832 Chr.12: 10015910-10015910 on Build GRCh38
rs200349068 Chr.12: 10015575-10015575 on Build GRCh38
rs1017540727 Chr.12: 10015596-10015596 on Build GRCh38
rs751720828 Chr.12: 10015769-10015769 on Build GRCh38
rs746417798 Chr.12: 10015598-10015598 on Build GRCh38
rs752650812 Chr.12: 10015661-10015661 on Build GRCh38
rs767867368 Chr.12: 10015790-10015790 on Build GRCh38
rs897198406 Chr.12: 10015909-10015909 on Build GRCh38
rs140525209 Chr.12: 10015695-10015695 on Build GRCh38
rs1013598707 Chr.12: 10015523-10015523 on Build GRCh38

View Full Product Details