Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110326243-110326563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A2
Entrez Gene ID
488
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Gene Aliases
ATP2B, DAR, DD, SERCA2
UniGene
Hs.506759
Gene Chromosome Location
Chr.12: 110281227-110351093 on Build GRCh38
Transcripts
NM_001681.3 NM_170665.3 XM_005253888.2 XM_011538402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7292360 c.630+4A>G p.? Chr.12: 110326479-110326479 on Build GRCh38
5475605 c.593G>A p.R198Q Chr.12: 110326438-110326438 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751208656 Chr.12: 110326391-110326391 on Build GRCh38
rs923858558 Chr.12: 110326448-110326448 on Build GRCh38
rs140380051 Chr.12: 110326527-110326527 on Build GRCh38
rs767026097 Chr.12: 110326407-110326407 on Build GRCh38
rs768723006 Chr.12: 110326352-110326352 on Build GRCh38
rs1019545890 Chr.12: 110326343-110326343 on Build GRCh38
rs371699814 Chr.12: 110326424-110326424 on Build GRCh38
rs761930681 Chr.12: 110326360-110326360 on Build GRCh38
rs753755593 Chr.12: 110326438-110326438 on Build GRCh38
rs745604029 Chr.12: 110326469-110326469 on Build GRCh38
rs778802900 Chr.12: 110326460-110326460 on Build GRCh38
rs995682220 Chr.12: 110326282-110326282 on Build GRCh38
rs953254791 Chr.12: 110326273-110326273 on Build GRCh38
rs375924802 Chr.12: 110326425-110326425 on Build GRCh38
rs374535528 Chr.12: 110326485-110326485 on Build GRCh38
rs898694020 Chr.12: 110326270-110326270 on Build GRCh38
rs748302089 Chr.12: 110326516-110326516 on Build GRCh38
rs758234347 Chr.12: 110326341-110326365 on Build GRCh38
rs191476843 Chr.12: 110326344-110326344 on Build GRCh38
rs987680100 Chr.12: 110326291-110326291 on Build GRCh38
rs757044025 Chr.12: 110326457-110326457 on Build GRCh38
rs766162395 Chr.12: 110326363-110326364 on Build GRCh38
rs770432663 Chr.12: 110326361-110326361 on Build GRCh38
rs759262783 Chr.12: 110326406-110326406 on Build GRCh38
rs953403434 Chr.12: 110326534-110326534 on Build GRCh38
rs573195762 Chr.12: 110326317-110326317 on Build GRCh38
rs914648136 Chr.12: 110326359-110326359 on Build GRCh38
rs886048952 Chr.12: 110326400-110326400 on Build GRCh38
rs368762109 Chr.12: 110326427-110326427 on Build GRCh38
rs781501530 Chr.12: 110326513-110326513 on Build GRCh38
rs971762641 Chr.12: 110326334-110326334 on Build GRCh38
rs551637459 Chr.12: 110326532-110326532 on Build GRCh38
rs773847727 Chr.12: 110326370-110326370 on Build GRCh38
rs369585470 Chr.12: 110326383-110326383 on Build GRCh38
rs747224258 Chr.12: 110326349-110326349 on Build GRCh38
rs1008651652 Chr.12: 110326332-110326332 on Build GRCh38
rs947687550 Chr.12: 110326537-110326537 on Build GRCh38
rs144121819 Chr.12: 110326531-110326531 on Build GRCh38
rs951620496 Chr.12: 110326329-110326329 on Build GRCh38
rs948275949 Chr.12: 110326385-110326385 on Build GRCh38
rs558110992 Chr.12: 110326313-110326313 on Build GRCh38
rs911673041 Chr.12: 110326300-110326300 on Build GRCh38
rs1052438003 Chr.12: 110326478-110326478 on Build GRCh38
rs1044871403 Chr.12: 110326538-110326538 on Build GRCh38
rs766741286 Chr.12: 110326384-110326384 on Build GRCh38
rs182640040 Chr.12: 110326353-110326353 on Build GRCh38

View Full Product Details