Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112669742-112670297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA1
Entrez Gene ID
829
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Gene Aliases
CAPPA1, CAPZ, CAZ1
UniGene
Hs.744974
Gene Chromosome Location
Chr.1: 112619453-112671619 on Build GRCh38
Transcripts
NM_006135.2 XM_017002424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4945514 c.721-7A>G p.? Chr.1: 112669985-112669985 on Build GRCh38
7116040 c.742C>G p.Q248E Chr.1: 112670013-112670013 on Build GRCh38
4909504 c.728T>G p.I243S Chr.1: 112669999-112669999 on Build GRCh38
6988587 c.775C>T p.R259C Chr.1: 112670046-112670046 on Build GRCh38
3801290 c.852G>T p.Q284H Chr.1: 112670123-112670123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745806008 Chr.1: 112670043-112670043 on Build GRCh38
rs577970194 Chr.1: 112669875-112669875 on Build GRCh38
rs988246019 Chr.1: 112669794-112669794 on Build GRCh38
rs756406634 Chr.1: 112669956-112669956 on Build GRCh38
rs529599201 Chr.1: 112669939-112669939 on Build GRCh38
rs958330224 Chr.1: 112669782-112669782 on Build GRCh38
rs766151613 Chr.1: 112669994-112669994 on Build GRCh38
rs946832682 Chr.1: 112670057-112670057 on Build GRCh38
rs769657219 Chr.1: 112670068-112670068 on Build GRCh38
rs371240455 Chr.1: 112670247-112670247 on Build GRCh38
rs879857260 Chr.1: 112670269-112670269 on Build GRCh38
rs369208203 Chr.1: 112669771-112669771 on Build GRCh38
rs1040595141 Chr.1: 112670174-112670174 on Build GRCh38
rs755175816 Chr.1: 112669970-112669970 on Build GRCh38
rs368445388 Chr.1: 112670157-112670157 on Build GRCh38
rs766960017 Chr.1: 112669958-112669958 on Build GRCh38
rs1060163 Chr.1: 112670051-112670051 on Build GRCh38
rs185470302 Chr.1: 112670272-112670272 on Build GRCh38
rs924563900 Chr.1: 112669822-112669822 on Build GRCh38
rs762414071 Chr.1: 112670072-112670072 on Build GRCh38
rs201104692 Chr.1: 112670067-112670067 on Build GRCh38
rs763850861 Chr.1: 112670076-112670076 on Build GRCh38
rs752864875 Chr.1: 112670154-112670154 on Build GRCh38
rs200171825 Chr.1: 112670180-112670180 on Build GRCh38
rs541169021 Chr.1: 112669865-112669865 on Build GRCh38
rs754385708 Chr.1: 112669959-112669959 on Build GRCh38
rs748189723 Chr.1: 112669992-112669992 on Build GRCh38
rs775007155 Chr.1: 112670070-112670070 on Build GRCh38
rs765798874 Chr.1: 112670138-112670138 on Build GRCh38
rs761161086 Chr.1: 112670113-112670113 on Build GRCh38
rs201851829 Chr.1: 112670027-112670027 on Build GRCh38
rs777824472 Chr.1: 112670007-112670007 on Build GRCh38
rs754189444 Chr.1: 112670120-112670120 on Build GRCh38
rs563996968 Chr.1: 112669935-112669935 on Build GRCh38
rs530446657 Chr.1: 112670249-112670249 on Build GRCh38
rs375232761 Chr.1: 112670156-112670156 on Build GRCh38
rs780975520 Chr.1: 112670179-112670179 on Build GRCh38
rs954745667 Chr.1: 112669866-112669866 on Build GRCh38
rs750833761 Chr.1: 112669952-112669952 on Build GRCh38
rs1049248507 Chr.1: 112670214-112670215 on Build GRCh38
rs758678092 Chr.1: 112670004-112670004 on Build GRCh38
rs570789296 Chr.1: 112669985-112669985 on Build GRCh38
rs760135290 Chr.1: 112670134-112670134 on Build GRCh38
rs773856348 Chr.1: 112670079-112670079 on Build GRCh38
rs772530423 Chr.1: 112669799-112669799 on Build GRCh38
rs199650232 Chr.1: 112670169-112670169 on Build GRCh38
rs763608345 Chr.1: 112669944-112669944 on Build GRCh38
rs111929717 Chr.1: 112669885-112669885 on Build GRCh38
rs746282943 Chr.1: 112669770-112669770 on Build GRCh38
rs956060806 Chr.1: 112669863-112669863 on Build GRCh38
rs558190350 Chr.1: 112669870-112669870 on Build GRCh38
rs971082666 Chr.1: 112669772-112669772 on Build GRCh38
rs551198154 Chr.1: 112670160-112670160 on Build GRCh38
rs559987730 Chr.1: 112670033-112670033 on Build GRCh38

View Full Product Details