Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101025182-101025752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH5
Entrez Gene ID
51611
Gene Name
diphthamide biosynthesis 5
Gene Aliases
AD-018, CGI-30, HSPC143, NPD015
UniGene
Hs.440776
Gene Chromosome Location
Chr.1: 100989623-101025806 on Build GRCh38
Transcripts
NM_001077394.1 NM_001077395.1 NM_015958.2 XM_005270938.1 XM_005270939.4 XM_005270941.3 XM_006710683.3 XM_011541569.1 XM_011541570.2 XM_017001460.1 XM_017001461.1 XM_017001462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1953380 c.122G>A p.G41E Chr.1: 101025322-101025322 on Build GRCh38
1294613 c.12C>T p.L4L Chr.1: 101025432-101025432 on Build GRCh38
5620720 c.112C>A p.L38I Chr.1: 101025332-101025332 on Build GRCh38
3687000 c.16G>A p.G6R Chr.1: 101025428-101025428 on Build GRCh38
4917913 c.100T>A p.Y34N Chr.1: 101025344-101025344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760974191 Chr.1: 101025354-101025354 on Build GRCh38
rs746152114 Chr.1: 101025313-101025314 on Build GRCh38
rs747835119 Chr.1: 101025433-101025433 on Build GRCh38
rs958050528 Chr.1: 101025466-101025466 on Build GRCh38
rs913591291 Chr.1: 101025512-101025512 on Build GRCh38
rs200436425 Chr.1: 101025493-101025493 on Build GRCh38
rs533477724 Chr.1: 101025576-101025576 on Build GRCh38
rs368076446 Chr.1: 101025364-101025364 on Build GRCh38
rs545369662 Chr.1: 101025579-101025579 on Build GRCh38
rs369898111 Chr.1: 101025504-101025504 on Build GRCh38
rs973906162 Chr.1: 101025580-101025580 on Build GRCh38
rs756419656 Chr.1: 101025549-101025549 on Build GRCh38
rs775632795 Chr.1: 101025324-101025324 on Build GRCh38
rs759682647 Chr.1: 101025361-101025361 on Build GRCh38
rs187230132 Chr.1: 101025245-101025245 on Build GRCh38
rs554107170 Chr.1: 101025244-101025244 on Build GRCh38
rs775388935 Chr.1: 101025467-101025467 on Build GRCh38
rs998192985 Chr.1: 101025638-101025638 on Build GRCh38
rs555886953 Chr.1: 101025510-101025510 on Build GRCh38
rs201869101 Chr.1: 101025665-101025665 on Build GRCh38
rs751818278 Chr.1: 101025391-101025391 on Build GRCh38
rs192495268 Chr.1: 101025582-101025582 on Build GRCh38
rs776094357 Chr.1: 101025351-101025352 on Build GRCh38
rs573897914 Chr.1: 101025535-101025535 on Build GRCh38
rs576454336 Chr.1: 101025383-101025383 on Build GRCh38
rs771853622 Chr.1: 101025439-101025439 on Build GRCh38
rs769400196 Chr.1: 101025487-101025487 on Build GRCh38
rs1048517152 Chr.1: 101025233-101025233 on Build GRCh38
rs370546827 Chr.1: 101025411-101025411 on Build GRCh38
rs768430351 Chr.1: 101025426-101025426 on Build GRCh38
rs12090113 Chr.1: 101025658-101025658 on Build GRCh38
rs777976166 Chr.1: 101025304-101025304 on Build GRCh38
rs543567903 Chr.1: 101025414-101025414 on Build GRCh38
rs373682605 Chr.1: 101025463-101025463 on Build GRCh38
rs777655588 Chr.1: 101025488-101025488 on Build GRCh38
rs749280357 Chr.1: 101025423-101025423 on Build GRCh38
rs773978803 Chr.1: 101025347-101025347 on Build GRCh38
rs745316287 Chr.1: 101025321-101025321 on Build GRCh38
rs766516181 Chr.1: 101025264-101025264 on Build GRCh38
rs761442725 Chr.1: 101025460-101025461 on Build GRCh38
rs910040796 Chr.1: 101025562-101025562 on Build GRCh38
rs781443324 Chr.1: 101025375-101025375 on Build GRCh38
rs796666826 Chr.1: 101025380-101025380 on Build GRCh38
rs756946997 Chr.1: 101025394-101025394 on Build GRCh38
rs369798016 Chr.1: 101025456-101025458 on Build GRCh38
rs558728556 Chr.1: 101025403-101025403 on Build GRCh38
rs779327512 Chr.1: 101025285-101025285 on Build GRCh38
rs763535068 Chr.1: 101025479-101025479 on Build GRCh38
rs764546987 Chr.1: 101025388-101025388 on Build GRCh38
rs568196177 Chr.1: 101025682-101025682 on Build GRCh38
rs1037118649 Chr.1: 101025581-101025581 on Build GRCh38
rs1013953000 Chr.1: 101025516-101025516 on Build GRCh38
rs112985750 Chr.1: 101025650-101025650 on Build GRCh38
rs757408859 Chr.1: 101025266-101025266 on Build GRCh38
rs776898562 Chr.1: 101025450-101025450 on Build GRCh38
rs370751772 Chr.1: 101025296-101025296 on Build GRCh38
rs2281828 Chr.1: 101025709-101025709 on Build GRCh38
rs944993107 Chr.1: 101025271-101025272 on Build GRCh38
rs772649250 Chr.1: 101025339-101025341 on Build GRCh38
rs1049344682 Chr.1: 101025205-101025205 on Build GRCh38
rs746714548 Chr.1: 101025307-101025307 on Build GRCh38
rs981709688 Chr.1: 101025532-101025532 on Build GRCh38
rs7521668 Chr.1: 101025552-101025552 on Build GRCh38
rs557895435 Chr.1: 101025363-101025363 on Build GRCh38
rs879002991 Chr.1: 101025607-101025607 on Build GRCh38
rs539828388 Chr.1: 101025316-101025316 on Build GRCh38
rs1007463157 Chr.1: 101025692-101025692 on Build GRCh38
rs760780847 Chr.1: 101025258-101025258 on Build GRCh38
rs373935279 Chr.1: 101025343-101025343 on Build GRCh38
rs759729888 Chr.1: 101025462-101025462 on Build GRCh38
rs766534010 Chr.1: 101025358-101025358 on Build GRCh38
rs780924835 Chr.1: 101025400-101025400 on Build GRCh38
rs769272309 Chr.1: 101025724-101025724 on Build GRCh38
rs1030254094 Chr.1: 101025311-101025311 on Build GRCh38
rs755736052 Chr.1: 101025412-101025412 on Build GRCh38
rs970370354 Chr.1: 101025531-101025531 on Build GRCh38
rs867767657 Chr.1: 101025381-101025381 on Build GRCh38
rs559606945 Chr.1: 101025573-101025573 on Build GRCh38
rs770450078 Chr.1: 101025312-101025312 on Build GRCh38
rs762779877 Chr.1: 101025325-101025325 on Build GRCh38
rs1037471002 Chr.1: 101025255-101025255 on Build GRCh38
rs957888900 Chr.1: 101025719-101025719 on Build GRCh38
rs769855449 Chr.1: 101025322-101025322 on Build GRCh38
rs753199774 Chr.1: 101025289-101025289 on Build GRCh38

View Full Product Details