Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78119241-78119774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC2
Entrez Gene ID
54810
Gene Name
GIPC PDZ domain containing family member 2
Gene Aliases
SEMCAP-2, SEMCAP2
UniGene
Hs.603818
Gene Chromosome Location
Chr.1: 78044962-78138449 on Build GRCh38
Transcripts
NM_001304725.1 NM_017655.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261096 c.646G>T p.E216* Chr.1: 78119431-78119431 on Build GRCh38
6507454 c.703G>A p.V235M Chr.1: 78119488-78119488 on Build GRCh38
5811701 c.679C>T p.L227L Chr.1: 78119464-78119464 on Build GRCh38
5847340 c.667G>A p.A223T Chr.1: 78119452-78119452 on Build GRCh38
4635977 c.623C>G p.S208* Chr.1: 78119408-78119408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143657223 Chr.1: 78119461-78119461 on Build GRCh38
rs767963986 Chr.1: 78119432-78119433 on Build GRCh38
rs747979024 Chr.1: 78119488-78119488 on Build GRCh38
rs921137721 Chr.1: 78119298-78119298 on Build GRCh38
rs932557449 Chr.1: 78119682-78119682 on Build GRCh38
rs912381138 Chr.1: 78119720-78119720 on Build GRCh38
rs771150840 Chr.1: 78119354-78119354 on Build GRCh38
rs1028719051 Chr.1: 78119724-78119724 on Build GRCh38
rs772159949 Chr.1: 78119490-78119490 on Build GRCh38
rs745978605 Chr.1: 78119364-78119364 on Build GRCh38
rs759084482 Chr.1: 78119525-78119525 on Build GRCh38
rs761653875 Chr.1: 78119383-78119383 on Build GRCh38
rs757428096 Chr.1: 78119448-78119448 on Build GRCh38
rs1051389409 Chr.1: 78119707-78119707 on Build GRCh38
rs373924776 Chr.1: 78119395-78119395 on Build GRCh38
rs35488228 Chr.1: 78119375-78119376 on Build GRCh38
rs375698203 Chr.1: 78119676-78119676 on Build GRCh38
rs541952125 Chr.1: 78119470-78119470 on Build GRCh38
rs745938097 Chr.1: 78119467-78119467 on Build GRCh38
rs774201982 Chr.1: 78119366-78119366 on Build GRCh38
rs773449167 Chr.1: 78119505-78119505 on Build GRCh38
rs1008524884 Chr.1: 78119735-78119735 on Build GRCh38
rs777969624 Chr.1: 78119348-78119348 on Build GRCh38
rs908244120 Chr.1: 78119625-78119625 on Build GRCh38
rs765046211 Chr.1: 78119475-78119476 on Build GRCh38
rs763315127 Chr.1: 78119536-78119536 on Build GRCh38
rs765764514 Chr.1: 78119424-78119424 on Build GRCh38
rs758750170 Chr.1: 78119347-78119347 on Build GRCh38
rs371466361 Chr.1: 78119407-78119407 on Build GRCh38
rs752262910 Chr.1: 78119530-78119530 on Build GRCh38
rs540742 Chr.1: 78119402-78119402 on Build GRCh38
rs778051625 Chr.1: 78119441-78119441 on Build GRCh38
rs567944664 Chr.1: 78119440-78119440 on Build GRCh38
rs199817145 Chr.1: 78119496-78119496 on Build GRCh38
rs140087680 Chr.1: 78119431-78119431 on Build GRCh38
rs776489231 Chr.1: 78119359-78119359 on Build GRCh38
rs781196788 Chr.1: 78119357-78119357 on Build GRCh38
rs888887057 Chr.1: 78119725-78119725 on Build GRCh38
rs538925074 Chr.1: 78119443-78119443 on Build GRCh38
rs527650357 Chr.1: 78119375-78119375 on Build GRCh38
rs377606034 Chr.1: 78119547-78119547 on Build GRCh38
rs545756218 Chr.1: 78119575-78119575 on Build GRCh38
rs973673435 Chr.1: 78119643-78119643 on Build GRCh38
rs994577028 Chr.1: 78119706-78119706 on Build GRCh38
rs747164508 Chr.1: 78119353-78119353 on Build GRCh38
rs761778615 Chr.1: 78119472-78119472 on Build GRCh38
rs903172221 Chr.1: 78119455-78119455 on Build GRCh38
rs936754876 Chr.1: 78119444-78119444 on Build GRCh38
rs911790590 Chr.1: 78119271-78119271 on Build GRCh38
rs770688364 Chr.1: 78119505-78119505 on Build GRCh38
rs773102945 Chr.1: 78119491-78119491 on Build GRCh38
rs372526320 Chr.1: 78119361-78119361 on Build GRCh38
rs866467223 Chr.1: 78119504-78119504 on Build GRCh38
rs769821365 Chr.1: 78119469-78119469 on Build GRCh38
rs373994574 Chr.1: 78119384-78119384 on Build GRCh38
rs751889150 Chr.1: 78119549-78119549 on Build GRCh38
rs557751221 Chr.1: 78119624-78119624 on Build GRCh38
rs535572742 Chr.1: 78119320-78119320 on Build GRCh38
rs778904893 Chr.1: 78119483-78119483 on Build GRCh38
rs753214981 Chr.1: 78119429-78119429 on Build GRCh38
rs150748555 Chr.1: 78119270-78119270 on Build GRCh38
rs908193183 Chr.1: 78119273-78119273 on Build GRCh38
rs764613315 Chr.1: 78119546-78119546 on Build GRCh38
rs773156400 Chr.1: 78119386-78119386 on Build GRCh38
rs1038235562 Chr.1: 78119294-78119294 on Build GRCh38
rs764588322 Chr.1: 78119527-78119527 on Build GRCh38
rs1036532287 Chr.1: 78119595-78119595 on Build GRCh38
rs962353342 Chr.1: 78119625-78119625 on Build GRCh38
rs573817246 Chr.1: 78119276-78119276 on Build GRCh38
rs921189859 Chr.1: 78119644-78119644 on Build GRCh38
rs776488195 Chr.1: 78119522-78119522 on Build GRCh38

View Full Product Details