Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153967293-153967847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L4
Entrez Gene ID
148327
Gene Name
cAMP responsive element binding protein 3 like 4
Gene Aliases
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL
UniGene
Hs.372924
Gene Chromosome Location
Chr.1: 153967679-153974364 on Build GRCh38
Transcripts
NM_001255978.1 NM_001255980.1 NM_130898.3 NR_045658.1 XM_017000372.1 NM_001271960.1 NM_014437.4

Gene Symbol
SLC39A1
Entrez Gene ID
27173
Gene Name
solute carrier family 39 member 1
Gene Aliases
ZIP1, ZIRTL
UniGene
Hs.743291
Gene Chromosome Location
Chr.1: 153959099-153968184 on Build GRCh38
Transcripts
NM_001255978.1 NM_001255980.1 NM_130898.3 NR_045658.1 XM_017000372.1 NM_001271960.1 NM_014437.4

SNP Detail

SNP ID Chromosome Location
rs929428106 Chr.1: 153967554-153967554 on Build GRCh38
rs980969169 Chr.1: 153967313-153967313 on Build GRCh38
rs1052964645 Chr.1: 153967778-153967778 on Build GRCh38
rs201712747 Chr.1: 153967669-153967669 on Build GRCh38
rs975419920 Chr.1: 153967416-153967416 on Build GRCh38
rs11288257 Chr.1: 153967669-153967669 on Build GRCh38
rs914144523 Chr.1: 153967486-153967486 on Build GRCh38
rs1051798128 Chr.1: 153967350-153967350 on Build GRCh38
rs397701187 Chr.1: 153967678-153967678 on Build GRCh38
rs1023994849 Chr.1: 153967450-153967450 on Build GRCh38
rs988821045 Chr.1: 153967469-153967469 on Build GRCh38
rs942171575 Chr.1: 153967322-153967322 on Build GRCh38
rs560375831 Chr.1: 153967405-153967405 on Build GRCh38
rs1014403810 Chr.1: 153967448-153967448 on Build GRCh38
rs955399607 Chr.1: 153967435-153967435 on Build GRCh38
rs948813210 Chr.1: 153967620-153967631 on Build GRCh38
rs946887135 Chr.1: 153967503-153967503 on Build GRCh38
rs980998914 Chr.1: 153967495-153967495 on Build GRCh38
rs963593153 Chr.1: 153967513-153967513 on Build GRCh38
rs184126855 Chr.1: 153967814-153967814 on Build GRCh38
rs1000069787 Chr.1: 153967407-153967407 on Build GRCh38
rs963866152 Chr.1: 153967626-153967626 on Build GRCh38
rs549281353 Chr.1: 153967506-153967506 on Build GRCh38
rs968287048 Chr.1: 153967783-153967783 on Build GRCh38
rs1013910231 Chr.1: 153967529-153967529 on Build GRCh38
rs1007493196 Chr.1: 153967372-153967372 on Build GRCh38
rs564878840 Chr.1: 153967549-153967549 on Build GRCh38
rs562543656 Chr.1: 153967780-153967780 on Build GRCh38
rs937055598 Chr.1: 153967508-153967508 on Build GRCh38
rs1038940069 Chr.1: 153967385-153967385 on Build GRCh38
rs1047437258 Chr.1: 153967569-153967569 on Build GRCh38
rs542122764 Chr.1: 153967340-153967340 on Build GRCh38
rs902637958 Chr.1: 153967527-153967527 on Build GRCh38
rs1031502478 Chr.1: 153967421-153967421 on Build GRCh38
rs926807706 Chr.1: 153967321-153967321 on Build GRCh38
rs887009026 Chr.1: 153967582-153967582 on Build GRCh38
rs1029733138 Chr.1: 153967717-153967717 on Build GRCh38
rs1038971338 Chr.1: 153967665-153967665 on Build GRCh38
rs539898851 Chr.1: 153967538-153967538 on Build GRCh38
rs370932487 Chr.1: 153967330-153967330 on Build GRCh38
rs970101812 Chr.1: 153967480-153967480 on Build GRCh38
rs368076810 Chr.1: 153967551-153967551 on Build GRCh38
rs895508997 Chr.1: 153967517-153967517 on Build GRCh38
rs1005803066 Chr.1: 153967593-153967593 on Build GRCh38
rs565987596 Chr.1: 153967712-153967712 on Build GRCh38
rs1005358731 Chr.1: 153967359-153967359 on Build GRCh38
rs922632163 Chr.1: 153967417-153967417 on Build GRCh38
rs911910205 Chr.1: 153967600-153967600 on Build GRCh38
rs1052258387 Chr.1: 153967588-153967588 on Build GRCh38
rs12027029 Chr.1: 153967504-153967504 on Build GRCh38
rs904475660 Chr.1: 153967751-153967751 on Build GRCh38
rs532121100 Chr.1: 153967699-153967699 on Build GRCh38
rs1000101967 Chr.1: 153967761-153967761 on Build GRCh38
rs973575262 Chr.1: 153967514-153967514 on Build GRCh38
rs558480130 Chr.1: 153967424-153967424 on Build GRCh38
rs890432895 Chr.1: 153967358-153967358 on Build GRCh38
rs929371244 Chr.1: 153967343-153967343 on Build GRCh38
rs1017051340 Chr.1: 153967386-153967386 on Build GRCh38
rs904363424 Chr.1: 153967392-153967392 on Build GRCh38
rs867540286 Chr.1: 153967805-153967805 on Build GRCh38
rs955844743 Chr.1: 153967427-153967427 on Build GRCh38
rs998640762 Chr.1: 153967781-153967781 on Build GRCh38
rs540067587 Chr.1: 153967635-153967635 on Build GRCh38
rs1042456919 Chr.1: 153967522-153967522 on Build GRCh38

View Full Product Details