Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37523318-37523917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLCAP
Entrez Gene ID
10904
Gene Name
bladder cancer associated protein
Gene Aliases
BC10
UniGene
Hs.472651
Gene Chromosome Location
Chr.20: 37517417-37527931 on Build GRCh38
Transcripts
NM_001167820.1 NM_001167821.1 NM_001167822.2 NM_001167823.1 NM_006698.3 NM_001322802.1 NM_005386.3 NM_181689.2

Gene Symbol
NNAT
Entrez Gene ID
4826
Gene Name
neuronatin
Gene Aliases
Peg5
UniGene
Hs.504703
Gene Chromosome Location
Chr.20: 37521206-37523690 on Build GRCh38
Transcripts
NM_001167820.1 NM_001167821.1 NM_001167822.2 NM_001167823.1 NM_006698.3 NM_001322802.1 NM_005386.3 NM_181689.2

SNP Detail

SNP ID Chromosome Location
rs1041547451 Chr.20: 37523720-37523720 on Build GRCh38
rs777792534 Chr.20: 37523739-37523739 on Build GRCh38
rs969826623 Chr.20: 37523483-37523483 on Build GRCh38
rs928302641 Chr.20: 37523542-37523542 on Build GRCh38
rs556575719 Chr.20: 37523582-37523582 on Build GRCh38
rs950632740 Chr.20: 37523450-37523450 on Build GRCh38
rs1025656104 Chr.20: 37523774-37523774 on Build GRCh38
rs114560035 Chr.20: 37523752-37523752 on Build GRCh38
rs972435943 Chr.20: 37523725-37523725 on Build GRCh38
rs931261722 Chr.20: 37523389-37523389 on Build GRCh38
rs895612049 Chr.20: 37523851-37523851 on Build GRCh38
rs1016103332 Chr.20: 37523881-37523881 on Build GRCh38
rs985625525 Chr.20: 37523706-37523706 on Build GRCh38
rs865842970 Chr.20: 37523470-37523470 on Build GRCh38
rs1042893771 Chr.20: 37523867-37523867 on Build GRCh38
rs771118625 Chr.20: 37523577-37523577 on Build GRCh38
rs549699444 Chr.20: 37523367-37523367 on Build GRCh38
rs902576926 Chr.20: 37523875-37523875 on Build GRCh38
rs762628 Chr.20: 37523567-37523567 on Build GRCh38
rs147372366 Chr.20: 37523398-37523398 on Build GRCh38
rs1056996997 Chr.20: 37523820-37523820 on Build GRCh38
rs3178311 Chr.20: 37523684-37523684 on Build GRCh38
rs1019704623 Chr.20: 37523709-37523709 on Build GRCh38
rs938376456 Chr.20: 37523572-37523572 on Build GRCh38
rs980299706 Chr.20: 37523529-37523529 on Build GRCh38
rs1013642311 Chr.20: 37523519-37523519 on Build GRCh38
rs112883541 Chr.20: 37523388-37523388 on Build GRCh38
rs1000860076 Chr.20: 37523589-37523589 on Build GRCh38
rs746278720 Chr.20: 37523715-37523715 on Build GRCh38
rs954316407 Chr.20: 37523664-37523664 on Build GRCh38
rs949460613 Chr.20: 37523854-37523854 on Build GRCh38
rs538277386 Chr.20: 37523459-37523459 on Build GRCh38
rs913461662 Chr.20: 37523639-37523640 on Build GRCh38
rs889984733 Chr.20: 37523682-37523682 on Build GRCh38
rs145487894 Chr.20: 37523461-37523461 on Build GRCh38
rs947641335 Chr.20: 37523647-37523648 on Build GRCh38
rs1033030588 Chr.20: 37523645-37523645 on Build GRCh38
rs11469339 Chr.20: 37523386-37523387 on Build GRCh38
rs770265149 Chr.20: 37523742-37523742 on Build GRCh38
rs896607609 Chr.20: 37523502-37523502 on Build GRCh38
rs539585017 Chr.20: 37523405-37523405 on Build GRCh38
rs558126858 Chr.20: 37523806-37523806 on Build GRCh38
rs868544260 Chr.20: 37523424-37523424 on Build GRCh38
rs779556669 Chr.20: 37523780-37523780 on Build GRCh38
rs918273902 Chr.20: 37523726-37523726 on Build GRCh38

View Full Product Details