Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26572998-26573596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
UniGene
Hs.37445
Gene Chromosome Location
Chr.21: 26466209-26573404 on Build GRCh38
Transcripts
NM_001320768.1 NM_052954.3 NR_135472.1 XM_011529450.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211731 c.200G>A p.W67* Chr.21: 26573065-26573065 on Build GRCh38
6639618 c.259C>A p.P87T Chr.21: 26573006-26573006 on Build GRCh38
4629159 c.4A>G p.K2E Chr.21: 26573261-26573261 on Build GRCh38
6858849 c.248C>G p.S83C Chr.21: 26573017-26573017 on Build GRCh38
7259034 c.202C>T p.P68S Chr.21: 26573063-26573063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980556607 Chr.21: 26573515-26573515 on Build GRCh38
rs372128791 Chr.21: 26573088-26573088 on Build GRCh38
rs988980391 Chr.21: 26573563-26573563 on Build GRCh38
rs773852667 Chr.21: 26573052-26573052 on Build GRCh38
rs967266417 Chr.21: 26573181-26573181 on Build GRCh38
rs917336679 Chr.21: 26573221-26573221 on Build GRCh38
rs902617061 Chr.21: 26573378-26573378 on Build GRCh38
rs766477801 Chr.21: 26573019-26573019 on Build GRCh38
rs11909722 Chr.21: 26573434-26573434 on Build GRCh38
rs765500303 Chr.21: 26573021-26573022 on Build GRCh38
rs888235256 Chr.21: 26573401-26573401 on Build GRCh38
rs939771267 Chr.21: 26573535-26573535 on Build GRCh38
rs747913196 Chr.21: 26573028-26573028 on Build GRCh38
rs980224906 Chr.21: 26573251-26573251 on Build GRCh38
rs936222096 Chr.21: 26573074-26573074 on Build GRCh38
rs1046652353 Chr.21: 26573110-26573110 on Build GRCh38
rs926067314 Chr.21: 26573519-26573519 on Build GRCh38
rs977724482 Chr.21: 26573187-26573188 on Build GRCh38
rs920936675 Chr.21: 26573222-26573222 on Build GRCh38
rs996675405 Chr.21: 26573149-26573149 on Build GRCh38
rs113556917 Chr.21: 26573142-26573142 on Build GRCh38
rs11909749 Chr.21: 26573522-26573522 on Build GRCh38
rs892082374 Chr.21: 26573268-26573268 on Build GRCh38
rs912460270 Chr.21: 26573215-26573215 on Build GRCh38
rs1011394797 Chr.21: 26573323-26573323 on Build GRCh38
rs1002413139 Chr.21: 26573285-26573285 on Build GRCh38
rs996970003 Chr.21: 26573359-26573359 on Build GRCh38
rs539035963 Chr.21: 26573365-26573365 on Build GRCh38
rs1015511216 Chr.21: 26573424-26573424 on Build GRCh38
rs767730490 Chr.21: 26573139-26573139 on Build GRCh38
rs1041134389 Chr.21: 26573292-26573292 on Build GRCh38
rs751595053 Chr.21: 26573112-26573112 on Build GRCh38
rs538362698 Chr.21: 26573127-26573127 on Build GRCh38
rs749106796 Chr.21: 26573194-26573194 on Build GRCh38
rs532591284 Chr.21: 26573186-26573186 on Build GRCh38
rs548464418 Chr.21: 26573143-26573143 on Build GRCh38
rs1019317489 Chr.21: 26573114-26573114 on Build GRCh38
rs755995987 Chr.21: 26573033-26573033 on Build GRCh38
rs948839480 Chr.21: 26573228-26573228 on Build GRCh38
rs749208475 Chr.21: 26573045-26573045 on Build GRCh38
rs984210194 Chr.21: 26573498-26573498 on Build GRCh38
rs1053319472 Chr.21: 26573096-26573096 on Build GRCh38
rs751651024 Chr.21: 26573021-26573021 on Build GRCh38
rs80082914 Chr.21: 26573349-26573349 on Build GRCh38
rs752820529 Chr.21: 26573152-26573174 on Build GRCh38
rs376698392 Chr.21: 26573255-26573255 on Build GRCh38
rs1055046183 Chr.21: 26573296-26573296 on Build GRCh38
rs906810433 Chr.21: 26573132-26573132 on Build GRCh38
rs892093088 Chr.21: 26573103-26573103 on Build GRCh38
rs752946614 Chr.21: 26573164-26573164 on Build GRCh38
rs1049004285 Chr.21: 26573100-26573100 on Build GRCh38
rs569358844 Chr.21: 26573025-26573025 on Build GRCh38
rs1024755584 Chr.21: 26573469-26573469 on Build GRCh38
rs1021868952 Chr.21: 26573178-26573178 on Build GRCh38
rs11909726 Chr.21: 26573482-26573482 on Build GRCh38
rs576030731 Chr.21: 26573415-26573415 on Build GRCh38
rs1027985958 Chr.21: 26573475-26573475 on Build GRCh38
rs781279668 Chr.21: 26573027-26573027 on Build GRCh38
rs952586973 Chr.21: 26573171-26573171 on Build GRCh38
rs1028602261 Chr.21: 26573156-26573156 on Build GRCh38
rs931055135 Chr.21: 26573223-26573223 on Build GRCh38
rs756446566 Chr.21: 26573180-26573180 on Build GRCh38
rs771047962 Chr.21: 26573051-26573051 on Build GRCh38
rs982203468 Chr.21: 26573572-26573573 on Build GRCh38
rs755048541 Chr.21: 26573135-26573135 on Build GRCh38
rs369866897 Chr.21: 26573307-26573307 on Build GRCh38
rs11909702 Chr.21: 26573422-26573422 on Build GRCh38
rs554044979 Chr.21: 26573286-26573286 on Build GRCh38
rs926091525 Chr.21: 26573252-26573252 on Build GRCh38
rs775022750 Chr.21: 26573071-26573071 on Build GRCh38
rs1053266657 Chr.21: 26573267-26573267 on Build GRCh38
rs535278986 Chr.21: 26573061-26573061 on Build GRCh38
rs984012632 Chr.21: 26573172-26573172 on Build GRCh38
rs992862046 Chr.21: 26573213-26573213 on Build GRCh38
rs936174917 Chr.21: 26573263-26573263 on Build GRCh38
rs1039406706 Chr.21: 26573044-26573044 on Build GRCh38
rs778747021 Chr.21: 26573250-26573250 on Build GRCh38
rs1049510515 Chr.21: 26573384-26573384 on Build GRCh38
rs900910281 Chr.21: 26573340-26573340 on Build GRCh38
rs766812419 Chr.21: 26573106-26573106 on Build GRCh38
rs147898697 Chr.21: 26573436-26573436 on Build GRCh38
rs748698905 Chr.21: 26573431-26573431 on Build GRCh38
rs760365559 Chr.21: 26573075-26573075 on Build GRCh38
rs574099086 Chr.21: 26573397-26573397 on Build GRCh38
rs763440110 Chr.21: 26573094-26573094 on Build GRCh38
rs893840731 Chr.21: 26573317-26573317 on Build GRCh38
rs773630649 Chr.21: 26573090-26573090 on Build GRCh38
rs900960083 Chr.21: 26573121-26573121 on Build GRCh38
rs777761841 Chr.21: 26573042-26573042 on Build GRCh38
rs568653719 Chr.21: 26573190-26573190 on Build GRCh38
rs534185584 Chr.21: 26573244-26573244 on Build GRCh38
rs745373706 Chr.21: 26573053-26573053 on Build GRCh38
rs931086980 Chr.21: 26573076-26573076 on Build GRCh38

View Full Product Details