Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17796782-17797290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf91
Entrez Gene ID
54149
Gene Name
chromosome 21 open reading frame 91
Gene Aliases
C21orf14, C21orf38, CSSG1, EURL, YG81
UniGene
Hs.293811
Gene Chromosome Location
Chr.21: 17788967-17819386 on Build GRCh38
Transcripts
NM_001100420.1 NM_001100421.1 NM_017447.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612363 c.370A>T p.R124W Chr.21: 17796876-17796876 on Build GRCh38
69935 c.400C>G p.Q134E Chr.21: 17796846-17796846 on Build GRCh38
3549884 c.346C>T p.P116S Chr.21: 17796900-17796900 on Build GRCh38
7097718 c.183C>T p.C61C Chr.21: 17797063-17797063 on Build GRCh38
3719633 c.128-1G>C p.? Chr.21: 17797119-17797119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001726459 Chr.21: 17797215-17797215 on Build GRCh38
rs542207231 Chr.21: 17797178-17797178 on Build GRCh38
rs776870138 Chr.21: 17796883-17796883 on Build GRCh38
rs747048120 Chr.21: 17797004-17797004 on Build GRCh38
rs752652318 Chr.21: 17797068-17797068 on Build GRCh38
rs971282718 Chr.21: 17796873-17796873 on Build GRCh38
rs760182694 Chr.21: 17797125-17797125 on Build GRCh38
rs761751614 Chr.21: 17796997-17796997 on Build GRCh38
rs745977322 Chr.21: 17796837-17796837 on Build GRCh38
rs771590964 Chr.21: 17797149-17797149 on Build GRCh38
rs762588629 Chr.21: 17797022-17797022 on Build GRCh38
rs12626897 Chr.21: 17797236-17797236 on Build GRCh38
rs77760879 Chr.21: 17797031-17797031 on Build GRCh38
rs967665441 Chr.21: 17796922-17796922 on Build GRCh38
rs560422132 Chr.21: 17797146-17797146 on Build GRCh38
rs554277200 Chr.21: 17797206-17797206 on Build GRCh38
rs779318693 Chr.21: 17796967-17796967 on Build GRCh38
rs752456963 Chr.21: 17796958-17796958 on Build GRCh38
rs765829742 Chr.21: 17797124-17797125 on Build GRCh38
rs537781367 Chr.21: 17796988-17796988 on Build GRCh38
rs751781246 Chr.21: 17797166-17797166 on Build GRCh38
rs962158312 Chr.21: 17796812-17796812 on Build GRCh38
rs891717972 Chr.21: 17796833-17796833 on Build GRCh38
rs758702120 Chr.21: 17797094-17797094 on Build GRCh38
rs373695866 Chr.21: 17796909-17796909 on Build GRCh38
rs746069099 Chr.21: 17796982-17796982 on Build GRCh38
rs1047978 Chr.21: 17796838-17796838 on Build GRCh38
rs777151466 Chr.21: 17797008-17797008 on Build GRCh38
rs2824495 Chr.21: 17796901-17796901 on Build GRCh38
rs1047976 Chr.21: 17796896-17796896 on Build GRCh38
rs764899347 Chr.21: 17796902-17796902 on Build GRCh38
rs1002006418 Chr.21: 17796899-17796899 on Build GRCh38
rs201878970 Chr.21: 17797051-17797051 on Build GRCh38
rs1024550360 Chr.21: 17796851-17796851 on Build GRCh38
rs772906179 Chr.21: 17797019-17797019 on Build GRCh38
rs767432023 Chr.21: 17796943-17796943 on Build GRCh38
rs935087463 Chr.21: 17796949-17796949 on Build GRCh38
rs779230611 Chr.21: 17796830-17796830 on Build GRCh38
rs764060216 Chr.21: 17797160-17797160 on Build GRCh38
rs781595131 Chr.21: 17797114-17797114 on Build GRCh38
rs923416599 Chr.21: 17797053-17797053 on Build GRCh38
rs774015396 Chr.21: 17797140-17797140 on Build GRCh38
rs748550451 Chr.21: 17797009-17797009 on Build GRCh38
rs952014448 Chr.21: 17796913-17796913 on Build GRCh38
rs747461880 Chr.21: 17796876-17796876 on Build GRCh38
rs891500621 Chr.21: 17797204-17797204 on Build GRCh38
rs763970150 Chr.21: 17797090-17797090 on Build GRCh38
rs757568024 Chr.21: 17796818-17796818 on Build GRCh38
rs777018700 Chr.21: 17796995-17796997 on Build GRCh38
rs778115058 Chr.21: 17797132-17797132 on Build GRCh38
rs755051627 Chr.21: 17797112-17797112 on Build GRCh38
rs36095555 Chr.21: 17796816-17796816 on Build GRCh38
rs769135295 Chr.21: 17796882-17796882 on Build GRCh38
rs897860505 Chr.21: 17797231-17797231 on Build GRCh38
rs751618119 Chr.21: 17797111-17797111 on Build GRCh38
rs765089007 Chr.21: 17797159-17797159 on Build GRCh38
rs914837838 Chr.21: 17796912-17796912 on Build GRCh38
rs770118588 Chr.21: 17797013-17797013 on Build GRCh38
rs770513389 Chr.21: 17797139-17797139 on Build GRCh38
rs368816028 Chr.21: 17796961-17796961 on Build GRCh38
rs368266262 Chr.21: 17797122-17797122 on Build GRCh38
rs749595779 Chr.21: 17797133-17797133 on Build GRCh38
rs953696365 Chr.21: 17796852-17796852 on Build GRCh38
rs780270000 Chr.21: 17797110-17797110 on Build GRCh38
rs200420591 Chr.21: 17797007-17797007 on Build GRCh38
rs754875633 Chr.21: 17796939-17796939 on Build GRCh38
rs572586117 Chr.21: 17797217-17797217 on Build GRCh38
rs753200483 Chr.21: 17797136-17797139 on Build GRCh38
rs776116159 Chr.21: 17796863-17796863 on Build GRCh38
rs750745092 Chr.21: 17797093-17797093 on Build GRCh38
rs760599397 Chr.21: 17797074-17797074 on Build GRCh38
rs746133121 Chr.21: 17796868-17796868 on Build GRCh38
rs147365430 Chr.21: 17797163-17797163 on Build GRCh38
rs775630389 Chr.21: 17797157-17797157 on Build GRCh38
rs748638721 Chr.21: 17797116-17797116 on Build GRCh38
rs202099162 Chr.21: 17796930-17796930 on Build GRCh38
rs773748068 Chr.21: 17797024-17797024 on Build GRCh38

View Full Product Details