Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29268160-29268625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EWSR1
Entrez Gene ID
2130
Gene Name
EWS RNA binding protein 1
Gene Aliases
EWS, EWS-FLI1, bK984G1.4
UniGene
Hs.374477
Gene Chromosome Location
Chr.22: 29268009-29300525 on Build GRCh38
Transcripts
NM_001163285.1 NM_001163286.1 NM_001163287.1 NM_005243.3 NM_013986.3 XM_005261389.3 XM_005261390.4 XM_011529995.2 XM_011529996.2 XM_011529997.1 XM_011529998.1 XM_011529999.2 XM_011530000.1 XM_011530001.2 XM_011530002.2 XM_017028644.1 XM_017028645.1 XM_017028646.1 XM_017028647.1 XM_017028648.1 XM_017028649.1 XM_017028650.1 XM_017028651.1 XM_017028652.1 XM_017028653.1 XM_017028654.1 XM_017028655.1 XM_017028656.1 XM_017028657.1 XM_017028658.1 XM_017028659.1 XM_017028660.1 XM_017028661.1 XM_017028662.1 XM_017028663.1 XM_017028664.1 XM_017028665.1 XM_017028666.1 XM_011530107.2

Gene Symbol
RHBDD3
Entrez Gene ID
25807
Gene Name
rhomboid domain containing 3
Gene Aliases
C22orf3, HS984G1A, PTAG
UniGene
Hs.106730
Gene Chromosome Location
Chr.22: 29259855-29268196 on Build GRCh38
Transcripts
NM_001163285.1 NM_001163286.1 NM_001163287.1 NM_005243.3 NM_013986.3 XM_005261389.3 XM_005261390.4 XM_011529995.2 XM_011529996.2 XM_011529997.1 XM_011529998.1 XM_011529999.2 XM_011530000.1 XM_011530001.2 XM_011530002.2 XM_017028644.1 XM_017028645.1 XM_017028646.1 XM_017028647.1 XM_017028648.1 XM_017028649.1 XM_017028650.1 XM_017028651.1 XM_017028652.1 XM_017028653.1 XM_017028654.1 XM_017028655.1 XM_017028656.1 XM_017028657.1 XM_017028658.1 XM_017028659.1 XM_017028660.1 XM_017028661.1 XM_017028662.1 XM_017028663.1 XM_017028664.1 XM_017028665.1 XM_017028666.1 XM_011530107.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4939311 c.11C>G p.T4R Chr.22: 29268347-29268347 on Build GRCh38
4939313 c.11C>G p.T4R Chr.22: 29268347-29268347 on Build GRCh38
4939312 c.11C>G p.T4R Chr.22: 29268347-29268347 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772035256 Chr.22: 29268394-29268394 on Build GRCh38
rs904425185 Chr.22: 29268560-29268560 on Build GRCh38
rs752812760 Chr.22: 29268363-29268363 on Build GRCh38
rs371636788 Chr.22: 29268412-29268412 on Build GRCh38
rs909435076 Chr.22: 29268489-29268489 on Build GRCh38
rs367933864 Chr.22: 29268295-29268295 on Build GRCh38
rs774383141 Chr.22: 29268386-29268386 on Build GRCh38
rs918914611 Chr.22: 29268529-29268529 on Build GRCh38
rs761822106 Chr.22: 29268389-29268389 on Build GRCh38
rs770561780 Chr.22: 29268286-29268286 on Build GRCh38
rs900783068 Chr.22: 29268194-29268194 on Build GRCh38
rs754966628 Chr.22: 29268306-29268306 on Build GRCh38
rs796283376 Chr.22: 29268401-29268402 on Build GRCh38
rs773059081 Chr.22: 29268396-29268396 on Build GRCh38
rs147203739 Chr.22: 29268348-29268348 on Build GRCh38
rs372915198 Chr.22: 29268383-29268383 on Build GRCh38
rs998712768 Chr.22: 29268561-29268561 on Build GRCh38
rs755862686 Chr.22: 29268387-29268387 on Build GRCh38
rs570122737 Chr.22: 29268569-29268569 on Build GRCh38
rs757933040 Chr.22: 29268244-29268245 on Build GRCh38
rs111892587 Chr.22: 29268292-29268292 on Build GRCh38
rs774293267 Chr.22: 29268343-29268343 on Build GRCh38
rs758447329 Chr.22: 29268288-29268288 on Build GRCh38
rs377068270 Chr.22: 29268319-29268319 on Build GRCh38
rs201112362 Chr.22: 29268369-29268369 on Build GRCh38
rs923489982 Chr.22: 29268183-29268183 on Build GRCh38
rs551097984 Chr.22: 29268284-29268284 on Build GRCh38
rs953683079 Chr.22: 29268583-29268583 on Build GRCh38
rs754003425 Chr.22: 29268304-29268304 on Build GRCh38
rs368408350 Chr.22: 29268426-29268426 on Build GRCh38
rs968550987 Chr.22: 29268504-29268504 on Build GRCh38
rs779516758 Chr.22: 29268345-29268345 on Build GRCh38
rs981337481 Chr.22: 29268307-29268307 on Build GRCh38
rs894299451 Chr.22: 29268393-29268393 on Build GRCh38
rs762864394 Chr.22: 29268333-29268333 on Build GRCh38
rs199903725 Chr.22: 29268372-29268372 on Build GRCh38
rs147483212 Chr.22: 29268459-29268459 on Build GRCh38
rs1050538999 Chr.22: 29268200-29268200 on Build GRCh38
rs751395202 Chr.22: 29268302-29268302 on Build GRCh38
rs533107413 Chr.22: 29268380-29268380 on Build GRCh38
rs375095904 Chr.22: 29268328-29268328 on Build GRCh38
rs971144500 Chr.22: 29268443-29268443 on Build GRCh38
rs932936416 Chr.22: 29268379-29268379 on Build GRCh38
rs986388259 Chr.22: 29268487-29268487 on Build GRCh38
rs182158627 Chr.22: 29268535-29268535 on Build GRCh38
rs569075074 Chr.22: 29268234-29268234 on Build GRCh38
rs199648590 Chr.22: 29268355-29268355 on Build GRCh38
rs1030380871 Chr.22: 29268582-29268582 on Build GRCh38
rs774952349 Chr.22: 29268604-29268604 on Build GRCh38
rs199589576 Chr.22: 29268312-29268312 on Build GRCh38
rs1030951623 Chr.22: 29268462-29268462 on Build GRCh38
rs1023624461 Chr.22: 29268294-29268294 on Build GRCh38
rs371038010 Chr.22: 29268182-29268182 on Build GRCh38
rs190443563 Chr.22: 29268473-29268473 on Build GRCh38
rs760330065 Chr.22: 29268308-29268308 on Build GRCh38
rs566631691 Chr.22: 29268530-29268530 on Build GRCh38
rs764702775 Chr.22: 29268388-29268388 on Build GRCh38
rs957030826 Chr.22: 29268456-29268456 on Build GRCh38
rs1003536228 Chr.22: 29268592-29268592 on Build GRCh38
rs2301291 Chr.22: 29268434-29268434 on Build GRCh38
rs374736877 Chr.22: 29268400-29268400 on Build GRCh38
rs1000233730 Chr.22: 29268585-29268585 on Build GRCh38
rs878973171 Chr.22: 29268467-29268467 on Build GRCh38
rs758975710 Chr.22: 29268289-29268289 on Build GRCh38
rs972300735 Chr.22: 29268605-29268605 on Build GRCh38
rs1039103872 Chr.22: 29268191-29268191 on Build GRCh38
rs765303745 Chr.22: 29268357-29268357 on Build GRCh38
rs914372843 Chr.22: 29268496-29268496 on Build GRCh38
rs776222972 Chr.22: 29268320-29268320 on Build GRCh38
rs759738889 Chr.22: 29268398-29268398 on Build GRCh38
rs73884709 Chr.22: 29268419-29268419 on Build GRCh38
rs986375036 Chr.22: 29268365-29268365 on Build GRCh38
rs776108501 Chr.22: 29268381-29268382 on Build GRCh38
rs761661070 Chr.22: 29268346-29268346 on Build GRCh38
rs143153949 Chr.22: 29268197-29268197 on Build GRCh38
rs2231392 Chr.22: 29268334-29268334 on Build GRCh38
rs756462778 Chr.22: 29268303-29268303 on Build GRCh38
rs899343528 Chr.22: 29268573-29268573 on Build GRCh38
rs763833724 Chr.22: 29268300-29268300 on Build GRCh38
rs941797679 Chr.22: 29268228-29268228 on Build GRCh38
rs368609428 Chr.22: 29268374-29268374 on Build GRCh38
rs745332947 Chr.22: 29268196-29268196 on Build GRCh38
rs948548306 Chr.22: 29268554-29268554 on Build GRCh38
rs551698500 Chr.22: 29268506-29268506 on Build GRCh38
rs567389929 Chr.22: 29268568-29268568 on Build GRCh38
rs759275041 Chr.22: 29268385-29268385 on Build GRCh38
rs548964089 Chr.22: 29268545-29268545 on Build GRCh38
rs915733462 Chr.22: 29268546-29268546 on Build GRCh38
rs758846327 Chr.22: 29268368-29268368 on Build GRCh38
rs1016396817 Chr.22: 29268249-29268249 on Build GRCh38
rs1003460540 Chr.22: 29268450-29268450 on Build GRCh38
rs770783574 Chr.22: 29268379-29268384 on Build GRCh38
rs1007706725 Chr.22: 29268600-29268600 on Build GRCh38
rs1028491380 Chr.22: 29268361-29268387 on Build GRCh38
rs535706908 Chr.22: 29268602-29268602 on Build GRCh38
rs998049767 Chr.22: 29268581-29268581 on Build GRCh38
rs202208272 Chr.22: 29268378-29268378 on Build GRCh38
rs539670651 Chr.22: 29268280-29268280 on Build GRCh38
rs941695479 Chr.22: 29268508-29268508 on Build GRCh38
rs529315731 Chr.22: 29268433-29268433 on Build GRCh38
rs748647019 Chr.22: 29268316-29268316 on Build GRCh38
rs758943413 Chr.22: 29268317-29268317 on Build GRCh38
rs923446320 Chr.22: 29268352-29268352 on Build GRCh38
rs752261478 Chr.22: 29268401-29268402 on Build GRCh38
rs745403052 Chr.22: 29268322-29268322 on Build GRCh38
rs970730494 Chr.22: 29268290-29268290 on Build GRCh38
rs1036073883 Chr.22: 29268271-29268271 on Build GRCh38
rs757565683 Chr.22: 29268370-29268370 on Build GRCh38
rs201886504 Chr.22: 29268318-29268318 on Build GRCh38
rs201087527 Chr.22: 29268384-29268384 on Build GRCh38
rs971744779 Chr.22: 29268526-29268526 on Build GRCh38
rs552992152 Chr.22: 29268329-29268329 on Build GRCh38
rs779487945 Chr.22: 29268376-29268376 on Build GRCh38
rs938461784 Chr.22: 29268540-29268540 on Build GRCh38
rs375313151 Chr.22: 29268353-29268353 on Build GRCh38
rs878945526 Chr.22: 29268509-29268509 on Build GRCh38
rs1042968658 Chr.22: 29268555-29268555 on Build GRCh38
rs550347784 Chr.22: 29268221-29268221 on Build GRCh38

View Full Product Details