Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19479107-19479655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC45
Entrez Gene ID
8318
Gene Name
cell division cycle 45
Gene Aliases
CDC45L, CDC45L2, PORC-PI-1
UniGene
Hs.474217
Gene Chromosome Location
Chr.22: 19479278-19520612 on Build GRCh38
Transcripts
XM_005261285.2 XM_005261286.3 XM_011530415.1 XM_011530416.1 XM_011530417.2 XM_011530418.2 NM_001035247.2 NM_005659.6

Gene Symbol
UFD1L
Entrez Gene ID
7353
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Gene Aliases
UFD1
UniGene
Hs.474213
Gene Chromosome Location
Chr.22: 19449941-19479215 on Build GRCh38
Transcripts
XM_005261285.2 XM_005261286.3 XM_011530415.1 XM_011530416.1 XM_011530417.2 XM_011530418.2 NM_001035247.2 NM_005659.6

SNP Detail

SNP ID Chromosome Location
rs554052568 Chr.22: 19479466-19479466 on Build GRCh38
rs911828943 Chr.22: 19479425-19479425 on Build GRCh38
rs11392459 Chr.22: 19479234-19479235 on Build GRCh38
rs1036385154 Chr.22: 19479314-19479314 on Build GRCh38
rs941347131 Chr.22: 19479155-19479155 on Build GRCh38
rs1006714617 Chr.22: 19479392-19479392 on Build GRCh38
rs997849047 Chr.22: 19479543-19479543 on Build GRCh38
rs755703333 Chr.22: 19479295-19479295 on Build GRCh38
rs939029721 Chr.22: 19479482-19479482 on Build GRCh38
rs533484130 Chr.22: 19479320-19479320 on Build GRCh38
rs946413813 Chr.22: 19479240-19479240 on Build GRCh38
rs535908897 Chr.22: 19479373-19479373 on Build GRCh38
rs953532103 Chr.22: 19479382-19479382 on Build GRCh38
rs569742524 Chr.22: 19479366-19479366 on Build GRCh38
rs995002873 Chr.22: 19479571-19479571 on Build GRCh38
rs774507480 Chr.22: 19479594-19479594 on Build GRCh38
rs762958311 Chr.22: 19479574-19479574 on Build GRCh38
rs1029058433 Chr.22: 19479368-19479368 on Build GRCh38
rs556169925 Chr.22: 19479592-19479592 on Build GRCh38
rs1017472447 Chr.22: 19479431-19479431 on Build GRCh38
rs545807767 Chr.22: 19479163-19479163 on Build GRCh38
rs963407580 Chr.22: 19479172-19479172 on Build GRCh38
rs1054632881 Chr.22: 19479515-19479515 on Build GRCh38
rs983573290 Chr.22: 19479162-19479162 on Build GRCh38
rs1014976404 Chr.22: 19479167-19479167 on Build GRCh38
rs1015007750 Chr.22: 19479354-19479354 on Build GRCh38
rs1056110476 Chr.22: 19479485-19479485 on Build GRCh38
rs915110436 Chr.22: 19479432-19479432 on Build GRCh38
rs893554456 Chr.22: 19479350-19479350 on Build GRCh38
rs544309728 Chr.22: 19479225-19479225 on Build GRCh38
rs985971548 Chr.22: 19479284-19479284 on Build GRCh38
rs899362171 Chr.22: 19479568-19479568 on Build GRCh38
rs941396349 Chr.22: 19479296-19479296 on Build GRCh38
rs921894368 Chr.22: 19479176-19479176 on Build GRCh38
rs562505955 Chr.22: 19479175-19479175 on Build GRCh38
rs1010569063 Chr.22: 19479214-19479214 on Build GRCh38
rs937545737 Chr.22: 19479497-19479497 on Build GRCh38
rs35642654 Chr.22: 19479620-19479620 on Build GRCh38
rs749347153 Chr.22: 19479348-19479348 on Build GRCh38
rs777109877 Chr.22: 19479133-19479133 on Build GRCh38
rs768979390 Chr.22: 19479506-19479506 on Build GRCh38
rs1003194266 Chr.22: 19479303-19479303 on Build GRCh38
rs760860301 Chr.22: 19479442-19479442 on Build GRCh38
rs1016774366 Chr.22: 19479578-19479578 on Build GRCh38
rs570805453 Chr.22: 19479313-19479313 on Build GRCh38
rs1035944799 Chr.22: 19479157-19479157 on Build GRCh38
rs909892223 Chr.22: 19479141-19479141 on Build GRCh38
rs953464591 Chr.22: 19479596-19479596 on Build GRCh38
rs904897929 Chr.22: 19479300-19479300 on Build GRCh38
rs893560057 Chr.22: 19479232-19479232 on Build GRCh38
rs531417225 Chr.22: 19479178-19479178 on Build GRCh38
rs200993945 Chr.22: 19479240-19479241 on Build GRCh38
rs747384143 Chr.22: 19479463-19479463 on Build GRCh38
rs987817501 Chr.22: 19479206-19479206 on Build GRCh38
rs946464315 Chr.22: 19479520-19479520 on Build GRCh38
rs546149761 Chr.22: 19479582-19479582 on Build GRCh38
rs1036566002 Chr.22: 19479559-19479559 on Build GRCh38
rs904930877 Chr.22: 19479471-19479471 on Build GRCh38
rs771462757 Chr.22: 19479130-19479130 on Build GRCh38
rs906051857 Chr.22: 19479322-19479322 on Build GRCh38
rs889705583 Chr.22: 19479562-19479562 on Build GRCh38
rs777360033 Chr.22: 19479346-19479346 on Build GRCh38
rs963438928 Chr.22: 19479359-19479359 on Build GRCh38
rs367647100 Chr.22: 19479436-19479437 on Build GRCh38
rs935194044 Chr.22: 19479429-19479429 on Build GRCh38
rs994237408 Chr.22: 19479612-19479612 on Build GRCh38
rs1025672423 Chr.22: 19479628-19479628 on Build GRCh38
rs911800374 Chr.22: 19479221-19479221 on Build GRCh38
rs1029561878 Chr.22: 19479371-19479371 on Build GRCh38
rs1033418741 Chr.22: 19479196-19479196 on Build GRCh38
rs960223546 Chr.22: 19479160-19479160 on Build GRCh38
rs868365356 Chr.22: 19479613-19479613 on Build GRCh38
rs746669457 Chr.22: 19479126-19479126 on Build GRCh38
rs188938675 Chr.22: 19479189-19479189 on Build GRCh38
rs139388652 Chr.22: 19479438-19479438 on Build GRCh38
rs770441180 Chr.22: 19479508-19479508 on Build GRCh38
rs5992403 Chr.22: 19479363-19479363 on Build GRCh38
rs761996504 Chr.22: 19479597-19479597 on Build GRCh38
rs918546512 Chr.22: 19479306-19479306 on Build GRCh38
rs770452919 Chr.22: 19479137-19479137 on Build GRCh38
rs950256397 Chr.22: 19479173-19479173 on Build GRCh38
rs1005026365 Chr.22: 19479342-19479342 on Build GRCh38
rs902203795 Chr.22: 19479532-19479532 on Build GRCh38
rs953879484 Chr.22: 19479273-19479273 on Build GRCh38
rs1005058884 Chr.22: 19479553-19479553 on Build GRCh38
rs555588915 Chr.22: 19479387-19479387 on Build GRCh38
rs914711181 Chr.22: 19479519-19479521 on Build GRCh38
rs1037271640 Chr.22: 19479159-19479159 on Build GRCh38
rs111370721 Chr.22: 19479431-19479431 on Build GRCh38
rs759927165 Chr.22: 19479135-19479135 on Build GRCh38
rs181054329 Chr.22: 19479200-19479200 on Build GRCh38
rs1009312818 Chr.22: 19479632-19479632 on Build GRCh38
rs1045960940 Chr.22: 19479318-19479318 on Build GRCh38
rs1029504826 Chr.22: 19479573-19479573 on Build GRCh38
rs13447178 Chr.22: 19479521-19479521 on Build GRCh38
rs548021193 Chr.22: 19479230-19479230 on Build GRCh38
rs982131433 Chr.22: 19479470-19479470 on Build GRCh38
rs772519826 Chr.22: 19479396-19479396 on Build GRCh38
rs530059102 Chr.22: 19479332-19479332 on Build GRCh38

View Full Product Details