Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46360533-46361080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
UniGene
Hs.252387
Gene Chromosome Location
Chr.22: 46360834-46537502 on Build GRCh38
Transcripts
NM_014246.1

SNP Detail

SNP ID Chromosome Location
rs989447762 Chr.22: 46360612-46360612 on Build GRCh38
rs189345987 Chr.22: 46360685-46360685 on Build GRCh38
rs141973202 Chr.22: 46360581-46360581 on Build GRCh38
rs973680113 Chr.22: 46360700-46360700 on Build GRCh38
rs888090753 Chr.22: 46360737-46360737 on Build GRCh38
rs56227926 Chr.22: 46360759-46360760 on Build GRCh38
rs541571372 Chr.22: 46360839-46360839 on Build GRCh38
rs986489199 Chr.22: 46360996-46360996 on Build GRCh38
rs982544627 Chr.22: 46360582-46360582 on Build GRCh38
rs768536167 Chr.22: 46360658-46360658 on Build GRCh38
rs953875668 Chr.22: 46360968-46360968 on Build GRCh38
rs915202253 Chr.22: 46360642-46360642 on Build GRCh38
rs977277482 Chr.22: 46360674-46360674 on Build GRCh38
rs888184130 Chr.22: 46360986-46360986 on Build GRCh38
rs59950551 Chr.22: 46361026-46361026 on Build GRCh38
rs367581785 Chr.22: 46360581-46360582 on Build GRCh38
rs921059810 Chr.22: 46360950-46360950 on Build GRCh38
rs987161604 Chr.22: 46360798-46360798 on Build GRCh38
rs899616136 Chr.22: 46360706-46360706 on Build GRCh38
rs936722716 Chr.22: 46361044-46361044 on Build GRCh38
rs936753574 Chr.22: 46360869-46360869 on Build GRCh38
rs1048094591 Chr.22: 46360733-46360733 on Build GRCh38
rs1055118984 Chr.22: 46360878-46360878 on Build GRCh38
rs76798643 Chr.22: 46361045-46361045 on Build GRCh38
rs966708069 Chr.22: 46360855-46360855 on Build GRCh38
rs185238140 Chr.22: 46361022-46361022 on Build GRCh38
rs578114313 Chr.22: 46360910-46360910 on Build GRCh38
rs977893105 Chr.22: 46361038-46361038 on Build GRCh38
rs982676032 Chr.22: 46360575-46360575 on Build GRCh38
rs895173618 Chr.22: 46360879-46360879 on Build GRCh38
rs932409514 Chr.22: 46360952-46360952 on Build GRCh38
rs537122532 Chr.22: 46360751-46360752 on Build GRCh38
rs943911516 Chr.22: 46360689-46360689 on Build GRCh38
rs775123632 Chr.22: 46360777-46360777 on Build GRCh38
rs1041276051 Chr.22: 46360696-46360696 on Build GRCh38
rs895274055 Chr.22: 46361040-46361040 on Build GRCh38
rs1015264327 Chr.22: 46360664-46360664 on Build GRCh38
rs6007891 Chr.22: 46360936-46360936 on Build GRCh38
rs973816147 Chr.22: 46360923-46360923 on Build GRCh38
rs1004163275 Chr.22: 46361027-46361027 on Build GRCh38
rs978099696 Chr.22: 46361061-46361061 on Build GRCh38
rs762430901 Chr.22: 46360823-46360823 on Build GRCh38
rs564002967 Chr.22: 46360752-46360752 on Build GRCh38
rs184563836 Chr.22: 46360660-46360660 on Build GRCh38
rs1004481303 Chr.22: 46360744-46360744 on Build GRCh38
rs926765973 Chr.22: 46360576-46360576 on Build GRCh38
rs932357382 Chr.22: 46360726-46360726 on Build GRCh38
rs920957585 Chr.22: 46360763-46360763 on Build GRCh38
rs1048145793 Chr.22: 46360952-46360955 on Build GRCh38
rs962423081 Chr.22: 46360922-46360922 on Build GRCh38
rs939691943 Chr.22: 46360997-46360997 on Build GRCh38
rs989498226 Chr.22: 46360825-46360825 on Build GRCh38
rs775438389 Chr.22: 46360951-46360952 on Build GRCh38
rs774410470 Chr.22: 46360859-46360859 on Build GRCh38
rs1013836783 Chr.22: 46360897-46360897 on Build GRCh38
rs571070072 Chr.22: 46361060-46361060 on Build GRCh38
rs947322974 Chr.22: 46360659-46360659 on Build GRCh38
rs1025683118 Chr.22: 46360898-46360898 on Build GRCh38
rs945167909 Chr.22: 46360837-46360837 on Build GRCh38
rs912387744 Chr.22: 46360807-46360807 on Build GRCh38
rs886702443 Chr.22: 46360903-46360903 on Build GRCh38
rs906774900 Chr.22: 46360752-46360752 on Build GRCh38
rs1005423293 Chr.22: 46360915-46360915 on Build GRCh38
rs1003748598 Chr.22: 46360756-46360756 on Build GRCh38
rs558194622 Chr.22: 46360904-46360904 on Build GRCh38
rs773670171 Chr.22: 46361039-46361039 on Build GRCh38
rs150695051 Chr.22: 46360919-46360919 on Build GRCh38
rs180835539 Chr.22: 46361011-46361011 on Build GRCh38
rs1025125116 Chr.22: 46360561-46360561 on Build GRCh38
rs924593477 Chr.22: 46360896-46360896 on Build GRCh38
rs376937278 Chr.22: 46360646-46360646 on Build GRCh38

View Full Product Details