Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24943832-24944342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_017004269.1 XM_017004270.1

SNP Detail

SNP ID Chromosome Location
rs963112244 Chr.2: 24944124-24944124 on Build GRCh38
rs1043174955 Chr.2: 24944279-24944279 on Build GRCh38
rs753650156 Chr.2: 24944028-24944028 on Build GRCh38
rs947972211 Chr.2: 24944164-24944164 on Build GRCh38
rs535943143 Chr.2: 24943988-24943988 on Build GRCh38
rs986113326 Chr.2: 24944223-24944223 on Build GRCh38
rs1014593908 Chr.2: 24944114-24944114 on Build GRCh38
rs764271989 Chr.2: 24943877-24943877 on Build GRCh38
rs184601460 Chr.2: 24944171-24944171 on Build GRCh38
rs1001846142 Chr.2: 24944007-24944007 on Build GRCh38
rs1022571443 Chr.2: 24944263-24944263 on Build GRCh38
rs111759308 Chr.2: 24943906-24943906 on Build GRCh38
rs1053119912 Chr.2: 24944237-24944237 on Build GRCh38
rs905686017 Chr.2: 24944282-24944282 on Build GRCh38
rs906332584 Chr.2: 24944189-24944196 on Build GRCh38
rs1035648076 Chr.2: 24944054-24944054 on Build GRCh38
rs548956689 Chr.2: 24944236-24944236 on Build GRCh38
rs959698638 Chr.2: 24944059-24944059 on Build GRCh38
rs139507282 Chr.2: 24944234-24944234 on Build GRCh38
rs1004510544 Chr.2: 24944092-24944092 on Build GRCh38
rs913294497 Chr.2: 24944266-24944266 on Build GRCh38
rs528969365 Chr.2: 24943905-24943905 on Build GRCh38
rs976429887 Chr.2: 24943966-24943966 on Build GRCh38
rs1010899623 Chr.2: 24944242-24944242 on Build GRCh38
rs189014318 Chr.2: 24944235-24944235 on Build GRCh38
rs147346699 Chr.2: 24943912-24943912 on Build GRCh38
rs954269629 Chr.2: 24944204-24944204 on Build GRCh38
rs533538495 Chr.2: 24944071-24944071 on Build GRCh38
rs540381625 Chr.2: 24944115-24944115 on Build GRCh38
rs909770941 Chr.2: 24944147-24944147 on Build GRCh38
rs895747428 Chr.2: 24944313-24944313 on Build GRCh38
rs766209784 Chr.2: 24943931-24943931 on Build GRCh38
rs747886916 Chr.2: 24944072-24944073 on Build GRCh38
rs947433267 Chr.2: 24944270-24944270 on Build GRCh38
rs575961236 Chr.2: 24944036-24944036 on Build GRCh38
rs754937286 Chr.2: 24944177-24944177 on Build GRCh38
rs1022547261 Chr.2: 24943898-24943898 on Build GRCh38
rs965517130 Chr.2: 24943932-24943932 on Build GRCh38
rs1057400707 Chr.2: 24944303-24944303 on Build GRCh38
rs1045314036 Chr.2: 24944178-24944178 on Build GRCh38
rs984029931 Chr.2: 24944119-24944119 on Build GRCh38
rs139523732 Chr.2: 24943901-24943901 on Build GRCh38
rs1001416261 Chr.2: 24944298-24944298 on Build GRCh38

View Full Product Details