Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52695764-52696362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D1
Entrez Gene ID
55830
Gene Name
glycosyltransferase 8 domain containing 1
Gene Aliases
AD-017, MSTP139
UniGene
Hs.297304
Gene Chromosome Location
Chr.3: 52694484-52706083 on Build GRCh38
Transcripts
NM_001010983.2 NM_001278280.1 NM_001278281.1 NM_018446.3 NM_152932.2 XM_017006857.1 XM_017006858.1 XM_017006859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769966 c.629G>A p.R210H Chr.3: 52695944-52695944 on Build GRCh38
5817147 c.504A>G p.I168M Chr.3: 52696262-52696262 on Build GRCh38
3660656 c.546C>T p.A182A Chr.3: 52696027-52696027 on Build GRCh38
6671822 c.508A>G p.M170V Chr.3: 52696258-52696258 on Build GRCh38
1424561 c.496_498delAAG p.K166delK Chr.3: 52696268-52696270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776174534 Chr.3: 52696047-52696047 on Build GRCh38
rs932695729 Chr.3: 52696151-52696151 on Build GRCh38
rs754701445 Chr.3: 52695948-52695948 on Build GRCh38
rs767101068 Chr.3: 52696054-52696054 on Build GRCh38
rs144225900 Chr.3: 52696031-52696031 on Build GRCh38
rs555358622 Chr.3: 52696210-52696210 on Build GRCh38
rs778832741 Chr.3: 52695951-52695951 on Build GRCh38
rs766023874 Chr.3: 52696057-52696057 on Build GRCh38
rs1049859490 Chr.3: 52696166-52696166 on Build GRCh38
rs149421790 Chr.3: 52695953-52695953 on Build GRCh38
rs995801347 Chr.3: 52696200-52696200 on Build GRCh38
rs749883786 Chr.3: 52696055-52696055 on Build GRCh38
rs986818593 Chr.3: 52696172-52696175 on Build GRCh38
rs753519075 Chr.3: 52696247-52696247 on Build GRCh38
rs759996639 Chr.3: 52696020-52696020 on Build GRCh38
rs760190823 Chr.3: 52695893-52695893 on Build GRCh38
rs753431211 Chr.3: 52696074-52696074 on Build GRCh38
rs750128092 Chr.3: 52695915-52695915 on Build GRCh38
rs539127489 Chr.3: 52695838-52695838 on Build GRCh38
rs765142070 Chr.3: 52696215-52696217 on Build GRCh38
rs972262487 Chr.3: 52696149-52696149 on Build GRCh38
rs889152051 Chr.3: 52695833-52695833 on Build GRCh38
rs933279247 Chr.3: 52695798-52695798 on Build GRCh38
rs979659602 Chr.3: 52696114-52696114 on Build GRCh38
rs777389753 Chr.3: 52695830-52695830 on Build GRCh38
rs369195827 Chr.3: 52695889-52695889 on Build GRCh38
rs780531701 Chr.3: 52696315-52696315 on Build GRCh38
rs903938638 Chr.3: 52695873-52695873 on Build GRCh38
rs745599341 Chr.3: 52696105-52696105 on Build GRCh38
rs1049746033 Chr.3: 52695922-52695922 on Build GRCh38
rs913694469 Chr.3: 52695847-52695847 on Build GRCh38
rs770123883 Chr.3: 52696306-52696306 on Build GRCh38
rs764726100 Chr.3: 52695908-52695909 on Build GRCh38
rs777976926 Chr.3: 52696091-52696091 on Build GRCh38
rs762176092 Chr.3: 52696189-52696189 on Build GRCh38
rs143702907 Chr.3: 52696277-52696277 on Build GRCh38
rs755696406 Chr.3: 52695926-52695926 on Build GRCh38
rs778024760 Chr.3: 52696196-52696196 on Build GRCh38
rs781393266 Chr.3: 52696281-52696281 on Build GRCh38
rs752731050 Chr.3: 52696083-52696083 on Build GRCh38
rs749146959 Chr.3: 52695856-52695856 on Build GRCh38
rs781589483 Chr.3: 52696192-52696192 on Build GRCh38
rs1027132400 Chr.3: 52696046-52696046 on Build GRCh38
rs770318494 Chr.3: 52696194-52696194 on Build GRCh38
rs769006344 Chr.3: 52696335-52696335 on Build GRCh38
rs778557312 Chr.3: 52696052-52696052 on Build GRCh38
rs923475383 Chr.3: 52696153-52696153 on Build GRCh38
rs766818676 Chr.3: 52695882-52695882 on Build GRCh38
rs945208799 Chr.3: 52695854-52695854 on Build GRCh38
rs910536861 Chr.3: 52696184-52696184 on Build GRCh38
rs763569723 Chr.3: 52696204-52696204 on Build GRCh38
rs754787642 Chr.3: 52696075-52696075 on Build GRCh38
rs747314056 Chr.3: 52695987-52695987 on Build GRCh38
rs878906574 Chr.3: 52695996-52695996 on Build GRCh38
rs996076164 Chr.3: 52696042-52696042 on Build GRCh38
rs751467911 Chr.3: 52696276-52696276 on Build GRCh38
rs947464036 Chr.3: 52695862-52695862 on Build GRCh38
rs761053183 Chr.3: 52695886-52695889 on Build GRCh38
rs2276812 Chr.3: 52695944-52695944 on Build GRCh38
rs925137697 Chr.3: 52696118-52696118 on Build GRCh38
rs1037074325 Chr.3: 52696198-52696198 on Build GRCh38
rs778638312 Chr.3: 52696081-52696081 on Build GRCh38
rs1050447572 Chr.3: 52695831-52695831 on Build GRCh38
rs766894865 Chr.3: 52695913-52695913 on Build GRCh38
rs72960225 Chr.3: 52696216-52696216 on Build GRCh38
rs371584519 Chr.3: 52696313-52696313 on Build GRCh38
rs771055063 Chr.3: 52696002-52696002 on Build GRCh38
rs897381004 Chr.3: 52696199-52696199 on Build GRCh38
rs776930236 Chr.3: 52696007-52696007 on Build GRCh38
rs979357568 Chr.3: 52696328-52696328 on Build GRCh38
rs557470468 Chr.3: 52696168-52696168 on Build GRCh38
rs752432253 Chr.3: 52696262-52696262 on Build GRCh38
rs746050763 Chr.3: 52696193-52696193 on Build GRCh38
rs758257520 Chr.3: 52696084-52696084 on Build GRCh38
rs756817900 Chr.3: 52695908-52695908 on Build GRCh38
rs79027420 Chr.3: 52696223-52696223 on Build GRCh38
rs772235053 Chr.3: 52695957-52695957 on Build GRCh38
rs967018639 Chr.3: 52696304-52696304 on Build GRCh38
rs771996544 Chr.3: 52696113-52696113 on Build GRCh38
rs765848089 Chr.3: 52695894-52695894 on Build GRCh38
rs752673790 Chr.3: 52696268-52696270 on Build GRCh38
rs372848323 Chr.3: 52696332-52696332 on Build GRCh38
rs750508333 Chr.3: 52695981-52695981 on Build GRCh38
rs944044791 Chr.3: 52696183-52696183 on Build GRCh38
rs759167757 Chr.3: 52696259-52696259 on Build GRCh38
rs760439503 Chr.3: 52696056-52696056 on Build GRCh38
rs758147539 Chr.3: 52696269-52696269 on Build GRCh38
rs775972057 Chr.3: 52696197-52696197 on Build GRCh38
rs765076471 Chr.3: 52696261-52696261 on Build GRCh38
rs753200246 Chr.3: 52695897-52695897 on Build GRCh38
rs1022615738 Chr.3: 52696087-52696087 on Build GRCh38
rs754137877 Chr.3: 52696077-52696078 on Build GRCh38
rs746139018 Chr.3: 52696291-52696291 on Build GRCh38
rs777589034 Chr.3: 52696270-52696270 on Build GRCh38
rs992053356 Chr.3: 52696100-52696100 on Build GRCh38
rs111818196 Chr.3: 52695976-52695976 on Build GRCh38
rs749077355 Chr.3: 52695945-52695945 on Build GRCh38

View Full Product Details