Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70808466-70809058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4F
Entrez Gene ID
165530
Gene Name
C-type lectin domain family 4 member F
Gene Aliases
CLECSF13, KCLR
UniGene
Hs.681239
Gene Chromosome Location
Chr.2: 70808643-70825235 on Build GRCh38
Transcripts
NM_001258027.1 NM_001321308.1 NM_173535.2 XM_011532635.2 XM_011532637.2 XM_011532638.2 XM_011532639.2 XM_011532640.2 XM_011532642.2 XM_011532643.1 XM_017003519.1

SNP Detail

SNP ID Chromosome Location
rs1035972003 Chr.2: 70809026-70809026 on Build GRCh38
rs782075476 Chr.2: 70808759-70808759 on Build GRCh38
rs187224887 Chr.2: 70808577-70808577 on Build GRCh38
rs1051657809 Chr.2: 70808883-70808883 on Build GRCh38
rs911732878 Chr.2: 70808899-70808899 on Build GRCh38
rs771796035 Chr.2: 70808873-70808873 on Build GRCh38
rs942031575 Chr.2: 70808506-70808506 on Build GRCh38
rs1052034392 Chr.2: 70808571-70808571 on Build GRCh38
rs970719491 Chr.2: 70808751-70808751 on Build GRCh38
rs564787397 Chr.2: 70808615-70808615 on Build GRCh38
rs1037689995 Chr.2: 70808520-70808520 on Build GRCh38
rs553068597 Chr.2: 70808657-70808657 on Build GRCh38
rs549216780 Chr.2: 70808865-70808865 on Build GRCh38
rs1001761611 Chr.2: 70808766-70808766 on Build GRCh38
rs2287100 Chr.2: 70808950-70808950 on Build GRCh38
rs1002214110 Chr.2: 70809017-70809017 on Build GRCh38
rs61739971 Chr.2: 70808941-70808941 on Build GRCh38
rs191252516 Chr.2: 70808644-70808644 on Build GRCh38
rs999243536 Chr.2: 70808652-70808652 on Build GRCh38
rs189634906 Chr.2: 70809006-70809006 on Build GRCh38
rs183779792 Chr.2: 70808796-70808796 on Build GRCh38
rs1010562934 Chr.2: 70808690-70808690 on Build GRCh38
rs931914293 Chr.2: 70808556-70808556 on Build GRCh38
rs775216025 Chr.2: 70808928-70808928 on Build GRCh38
rs150656803 Chr.2: 70808951-70808951 on Build GRCh38
rs536944918 Chr.2: 70808929-70808929 on Build GRCh38
rs571242533 Chr.2: 70808927-70808927 on Build GRCh38
rs782550534 Chr.2: 70808985-70808985 on Build GRCh38
rs907985980 Chr.2: 70808844-70808844 on Build GRCh38
rs782681262 Chr.2: 70808956-70808956 on Build GRCh38
rs113850787 Chr.2: 70809031-70809031 on Build GRCh38
rs528498128 Chr.2: 70808895-70808895 on Build GRCh38
rs374645812 Chr.2: 70808954-70808954 on Build GRCh38
rs147792663 Chr.2: 70808819-70808819 on Build GRCh38
rs983598800 Chr.2: 70808827-70808827 on Build GRCh38
rs529446585 Chr.2: 70808847-70808847 on Build GRCh38
rs377443149 Chr.2: 70808864-70808864 on Build GRCh38
rs782115435 Chr.2: 70808531-70808531 on Build GRCh38
rs544963009 Chr.2: 70808606-70808606 on Build GRCh38
rs141894191 Chr.2: 70808871-70808871 on Build GRCh38
rs1023256188 Chr.2: 70808703-70808703 on Build GRCh38
rs1052109684 Chr.2: 70808653-70808653 on Build GRCh38
rs544060730 Chr.2: 70808778-70808778 on Build GRCh38

View Full Product Details