Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25739962-25740515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL2
Entrez Gene ID
55252
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
ASXH2
UniGene
Hs.119815
Gene Chromosome Location
Chr.2: 25733758-25878485 on Build GRCh38
Transcripts
NM_018263.4 XM_006712039.3 XM_006712040.2 XM_011532950.2 XM_011532951.2 XM_017004429.1 XM_017004430.1

SNP Detail

SNP ID Chromosome Location
rs763267440 Chr.2: 25740346-25740362 on Build GRCh38
rs1023963015 Chr.2: 25740108-25740108 on Build GRCh38
rs10153943 Chr.2: 25740336-25740336 on Build GRCh38
rs893443653 Chr.2: 25740114-25740114 on Build GRCh38
rs556944885 Chr.2: 25740489-25740489 on Build GRCh38
rs550469111 Chr.2: 25740119-25740119 on Build GRCh38
rs567281986 Chr.2: 25740361-25740361 on Build GRCh38
rs780955029 Chr.2: 25740224-25740224 on Build GRCh38
rs964292657 Chr.2: 25740392-25740392 on Build GRCh38
rs185108440 Chr.2: 25740148-25740148 on Build GRCh38
rs1016777078 Chr.2: 25740390-25740390 on Build GRCh38
rs1036297268 Chr.2: 25740329-25740329 on Build GRCh38
rs942680696 Chr.2: 25740159-25740159 on Build GRCh38
rs752818735 Chr.2: 25740127-25740127 on Build GRCh38
rs557995306 Chr.2: 25740107-25740107 on Build GRCh38
rs758684372 Chr.2: 25740206-25740206 on Build GRCh38
rs145599896 Chr.2: 25740166-25740166 on Build GRCh38
rs995053181 Chr.2: 25740083-25740084 on Build GRCh38
rs970899211 Chr.2: 25740124-25740124 on Build GRCh38
rs983966290 Chr.2: 25740147-25740147 on Build GRCh38
rs906890483 Chr.2: 25740436-25740436 on Build GRCh38
rs147872484 Chr.2: 25739995-25739995 on Build GRCh38
rs922655514 Chr.2: 25740218-25740218 on Build GRCh38
rs922590816 Chr.2: 25740435-25740435 on Build GRCh38
rs773457737 Chr.2: 25740280-25740280 on Build GRCh38
rs1033297499 Chr.2: 25740087-25740087 on Build GRCh38
rs1000448618 Chr.2: 25740066-25740066 on Build GRCh38
rs939971963 Chr.2: 25740309-25740309 on Build GRCh38
rs148905796 Chr.2: 25740347-25740347 on Build GRCh38
rs533675397 Chr.2: 25740234-25740234 on Build GRCh38
rs951176523 Chr.2: 25740377-25740377 on Build GRCh38
rs79789849 Chr.2: 25740393-25740393 on Build GRCh38
rs755912422 Chr.2: 25740260-25740260 on Build GRCh38
rs781741036 Chr.2: 25740056-25740056 on Build GRCh38
rs534477861 Chr.2: 25739988-25739988 on Build GRCh38
rs17047134 Chr.2: 25740488-25740488 on Build GRCh38
rs149989272 Chr.2: 25740137-25740137 on Build GRCh38
rs1036650233 Chr.2: 25740003-25740003 on Build GRCh38
rs770248046 Chr.2: 25740225-25740225 on Build GRCh38
rs779914075 Chr.2: 25740349-25740349 on Build GRCh38
rs936162329 Chr.2: 25740235-25740235 on Build GRCh38
rs932758012 Chr.2: 25740167-25740167 on Build GRCh38
rs1054674672 Chr.2: 25740253-25740253 on Build GRCh38
rs1051125957 Chr.2: 25739986-25739986 on Build GRCh38
rs1023710037 Chr.2: 25740331-25740331 on Build GRCh38
rs970995514 Chr.2: 25740116-25740116 on Build GRCh38
rs529106000 Chr.2: 25740111-25740111 on Build GRCh38
rs140460610 Chr.2: 25740123-25740123 on Build GRCh38
rs527669491 Chr.2: 25739987-25739987 on Build GRCh38
rs1011888248 Chr.2: 25740273-25740273 on Build GRCh38
rs1025415402 Chr.2: 25740134-25740134 on Build GRCh38
rs934042316 Chr.2: 25740454-25740454 on Build GRCh38
rs76053652 Chr.2: 25740324-25740324 on Build GRCh38
rs1027426349 Chr.2: 25740093-25740093 on Build GRCh38
rs981914961 Chr.2: 25740129-25740129 on Build GRCh38
rs374645958 Chr.2: 25740095-25740098 on Build GRCh38
rs771167152 Chr.2: 25740482-25740482 on Build GRCh38
rs1008858213 Chr.2: 25739997-25739997 on Build GRCh38
rs930560539 Chr.2: 25740338-25740338 on Build GRCh38
rs983935186 Chr.2: 25740384-25740384 on Build GRCh38
rs1025353818 Chr.2: 25740356-25740356 on Build GRCh38
rs912569928 Chr.2: 25740262-25740263 on Build GRCh38
rs998902169 Chr.2: 25740471-25740471 on Build GRCh38
rs981403384 Chr.2: 25740217-25740217 on Build GRCh38

View Full Product Details