Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97657848-97658384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR1B
Entrez Gene ID
10120
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
Gene Aliases
ARP1B, CTRN2, PC3
UniGene
Hs.98791
Gene Chromosome Location
Chr.2: 97655939-97664098 on Build GRCh38
Transcripts
NM_005735.3 XM_005263854.4 XM_017003116.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238367 c.840C>T p.A280A Chr.2: 97658028-97658028 on Build GRCh38
384378 c.813G>C p.E271D Chr.2: 97658055-97658055 on Build GRCh38
5863043 c.724A>G p.T242A Chr.2: 97658250-97658250 on Build GRCh38
1283244 c.726G>T p.T242T Chr.2: 97658248-97658248 on Build GRCh38
5863042 c.898C>T p.L300F Chr.2: 97657970-97657970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377460048 Chr.2: 97658147-97658147 on Build GRCh38
rs368032697 Chr.2: 97657974-97657974 on Build GRCh38
rs754426761 Chr.2: 97657916-97657916 on Build GRCh38
rs769565201 Chr.2: 97658034-97658034 on Build GRCh38
rs765048686 Chr.2: 97658316-97658316 on Build GRCh38
rs200870080 Chr.2: 97658215-97658215 on Build GRCh38
rs551047888 Chr.2: 97658170-97658170 on Build GRCh38
rs750675813 Chr.2: 97658195-97658195 on Build GRCh38
rs751531607 Chr.2: 97658098-97658098 on Build GRCh38
rs751159878 Chr.2: 97657899-97657899 on Build GRCh38
rs779911304 Chr.2: 97658205-97658205 on Build GRCh38
rs1016512288 Chr.2: 97658031-97658031 on Build GRCh38
rs757328064 Chr.2: 97658105-97658105 on Build GRCh38
rs771691922 Chr.2: 97658163-97658163 on Build GRCh38
rs757416072 Chr.2: 97658016-97658016 on Build GRCh38
rs146624887 Chr.2: 97658239-97658239 on Build GRCh38
rs766871606 Chr.2: 97657989-97657989 on Build GRCh38
rs140435232 Chr.2: 97658066-97658066 on Build GRCh38
rs769045495 Chr.2: 97658249-97658249 on Build GRCh38
rs200969439 Chr.2: 97657910-97657910 on Build GRCh38
rs535608880 Chr.2: 97658238-97658238 on Build GRCh38
rs773219286 Chr.2: 97658266-97658266 on Build GRCh38
rs780973427 Chr.2: 97658021-97658021 on Build GRCh38
rs773384868 Chr.2: 97658100-97658102 on Build GRCh38
rs775404286 Chr.2: 97657973-97657973 on Build GRCh38
rs929905927 Chr.2: 97658020-97658020 on Build GRCh38
rs763875630 Chr.2: 97657895-97657895 on Build GRCh38
rs760033478 Chr.2: 97658067-97658067 on Build GRCh38
rs765672091 Chr.2: 97658073-97658073 on Build GRCh38
rs756781263 Chr.2: 97658343-97658343 on Build GRCh38
rs565812380 Chr.2: 97658267-97658267 on Build GRCh38
rs747173514 Chr.2: 97657954-97657954 on Build GRCh38
rs151155439 Chr.2: 97658226-97658226 on Build GRCh38
rs574561031 Chr.2: 97657881-97657881 on Build GRCh38
rs767606853 Chr.2: 97658259-97658259 on Build GRCh38
rs765908883 Chr.2: 97657996-97657996 on Build GRCh38
rs763325139 Chr.2: 97658090-97658090 on Build GRCh38
rs756280709 Chr.2: 97658196-97658196 on Build GRCh38
rs754677033 Chr.2: 97658077-97658077 on Build GRCh38
rs1002580055 Chr.2: 97658167-97658167 on Build GRCh38
rs753509567 Chr.2: 97658213-97658213 on Build GRCh38
rs770749832 Chr.2: 97657956-97657956 on Build GRCh38
rs200672180 Chr.2: 97657953-97657953 on Build GRCh38
rs745732438 Chr.2: 97658024-97658024 on Build GRCh38
rs764578250 Chr.2: 97658179-97658179 on Build GRCh38
rs1042705 Chr.2: 97658064-97658064 on Build GRCh38
rs1054596091 Chr.2: 97658121-97658121 on Build GRCh38
rs551464888 Chr.2: 97658160-97658160 on Build GRCh38
rs775662923 Chr.2: 97658177-97658177 on Build GRCh38
rs781453544 Chr.2: 97658108-97658108 on Build GRCh38
rs924698234 Chr.2: 97658085-97658085 on Build GRCh38
rs959282737 Chr.2: 97658045-97658045 on Build GRCh38
rs893350580 Chr.2: 97657903-97657903 on Build GRCh38
rs745504508 Chr.2: 97658353-97658353 on Build GRCh38
rs748190438 Chr.2: 97657937-97657937 on Build GRCh38
rs543225703 Chr.2: 97658099-97658099 on Build GRCh38
rs763287500 Chr.2: 97658248-97658248 on Build GRCh38
rs749270339 Chr.2: 97658139-97658139 on Build GRCh38
rs766577056 Chr.2: 97658269-97658269 on Build GRCh38
rs772468833 Chr.2: 97657983-97657983 on Build GRCh38
rs1024871070 Chr.2: 97658302-97658302 on Build GRCh38
rs767396304 Chr.2: 97658134-97658135 on Build GRCh38
rs775547948 Chr.2: 97658049-97658049 on Build GRCh38
rs752709465 Chr.2: 97658319-97658319 on Build GRCh38
rs138142670 Chr.2: 97657982-97657982 on Build GRCh38
rs753127424 Chr.2: 97657997-97657997 on Build GRCh38
rs760368632 Chr.2: 97658169-97658169 on Build GRCh38
rs760396356 Chr.2: 97658223-97658223 on Build GRCh38
rs777679337 Chr.2: 97658009-97658009 on Build GRCh38
rs752051108 Chr.2: 97658185-97658185 on Build GRCh38
rs1026006041 Chr.2: 97658006-97658006 on Build GRCh38
rs191369953 Chr.2: 97657994-97657994 on Build GRCh38
rs775439012 Chr.2: 97658221-97658222 on Build GRCh38
rs754195764 Chr.2: 97657990-97657990 on Build GRCh38
rs577318131 Chr.2: 97658005-97658005 on Build GRCh38
rs766532607 Chr.2: 97658153-97658153 on Build GRCh38
rs7608385 Chr.2: 97658320-97658320 on Build GRCh38
rs770221814 Chr.2: 97658173-97658173 on Build GRCh38
rs762269186 Chr.2: 97658255-97658255 on Build GRCh38
rs756551537 Chr.2: 97657909-97657909 on Build GRCh38
rs754795306 Chr.2: 97658214-97658214 on Build GRCh38
rs1046962974 Chr.2: 97658025-97658025 on Build GRCh38
rs189465867 Chr.2: 97658321-97658321 on Build GRCh38
rs893108204 Chr.2: 97658334-97658334 on Build GRCh38
rs779674723 Chr.2: 97658133-97658133 on Build GRCh38
rs367614205 Chr.2: 97658231-97658231 on Build GRCh38
rs769762491 Chr.2: 97658018-97658020 on Build GRCh38
rs1020422462 Chr.2: 97658150-97658150 on Build GRCh38
rs780760031 Chr.2: 97658345-97658345 on Build GRCh38
rs773684866 Chr.2: 97658061-97658061 on Build GRCh38
rs572322477 Chr.2: 97657971-97657971 on Build GRCh38
rs759449958 Chr.2: 97658133-97658134 on Build GRCh38
rs761931268 Chr.2: 97658189-97658189 on Build GRCh38
rs1040444916 Chr.2: 97658065-97658065 on Build GRCh38
rs757632610 Chr.2: 97658311-97658311 on Build GRCh38
rs376524220 Chr.2: 97658294-97658294 on Build GRCh38
rs755492665 Chr.2: 97657933-97657933 on Build GRCh38
rs368596295 Chr.2: 97658362-97658362 on Build GRCh38
rs755544675 Chr.2: 97657993-97657993 on Build GRCh38
rs201642763 Chr.2: 97657919-97657919 on Build GRCh38
rs762732953 Chr.2: 97657980-97657980 on Build GRCh38
rs759824041 Chr.2: 97658241-97658241 on Build GRCh38
rs951391155 Chr.2: 97658209-97658209 on Build GRCh38
rs144577602 Chr.2: 97658058-97658058 on Build GRCh38
rs373737375 Chr.2: 97658230-97658230 on Build GRCh38
rs184588007 Chr.2: 97658161-97658161 on Build GRCh38
rs758401310 Chr.2: 97657945-97657945 on Build GRCh38
rs747281452 Chr.2: 97658010-97658010 on Build GRCh38
rs370608191 Chr.2: 97658190-97658190 on Build GRCh38
rs761570777 Chr.2: 97658063-97658063 on Build GRCh38
rs748073377 Chr.2: 97658229-97658229 on Build GRCh38
rs750623469 Chr.2: 97658128-97658128 on Build GRCh38
rs559759747 Chr.2: 97658131-97658131 on Build GRCh38
rs530938033 Chr.2: 97658168-97658168 on Build GRCh38
rs997109120 Chr.2: 97658223-97658223 on Build GRCh38
rs149355636 Chr.2: 97657934-97657934 on Build GRCh38
rs753284851 Chr.2: 97658089-97658089 on Build GRCh38
rs768244227 Chr.2: 97658060-97658060 on Build GRCh38
rs1020391201 Chr.2: 97657928-97657928 on Build GRCh38
rs780795599 Chr.2: 97658091-97658091 on Build GRCh38

View Full Product Details