Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 121362886-121363379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
UniGene
Hs.469840
Gene Chromosome Location
Chr.2: 121337776-121649476 on Build GRCh38
Transcripts
NM_001142273.1 NM_001142274.1 NM_001207051.1 NM_015282.2 XM_006712377.1 XM_006712381.1 XM_006712382.1 XM_006712383.1 XM_006712385.1 XM_011510848.1 XM_011510849.1 XM_017003657.1 XM_017003658.1 XM_017003659.1 XM_017003660.1 XM_017003661.1 XM_017003662.1 XM_017003663.1 XM_017003664.1 XM_017003665.1 XM_017003666.1 XM_017003667.1 XM_017003668.1 XM_017003669.1 XM_017003670.1 XM_017003671.1 XM_017003672.1 XM_017003673.1 XM_017003674.1 XM_017003675.1 XM_017003676.1 XM_017003677.1 XM_017003678.1 XM_017003679.1 XM_017003680.1 XM_017003681.1 XM_017003682.1 XM_017003683.1 XM_017003684.1 XM_017003685.1 XM_017003686.1 XM_017003687.1 XM_017003688.1 XM_017003689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4861468 c.3973G>A p.A1325T Chr.2: 121363225-121363225 on Build GRCh38
6153551 c.3247G>A p.A1083T Chr.2: 121363282-121363282 on Build GRCh38
3749593 c.3238C>T p.R1080* Chr.2: 121363291-121363291 on Build GRCh38
4874145 c.3243A>G p.A1081A Chr.2: 121363286-121363286 on Build GRCh38
260120 c.4087C>T p.R1363* Chr.2: 121363291-121363291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750162677 Chr.2: 121363152-121363152 on Build GRCh38
rs982373402 Chr.2: 121363201-121363201 on Build GRCh38
rs900543985 Chr.2: 121363051-121363051 on Build GRCh38
rs1011524504 Chr.2: 121363017-121363017 on Build GRCh38
rs749035943 Chr.2: 121363173-121363173 on Build GRCh38
rs1015465277 Chr.2: 121363079-121363079 on Build GRCh38
rs935152739 Chr.2: 121363241-121363241 on Build GRCh38
rs548342902 Chr.2: 121363280-121363280 on Build GRCh38
rs951051665 Chr.2: 121363104-121363104 on Build GRCh38
rs556977582 Chr.2: 121363160-121363160 on Build GRCh38
rs755594869 Chr.2: 121363281-121363281 on Build GRCh38
rs1034026698 Chr.2: 121363088-121363088 on Build GRCh38
rs759165442 Chr.2: 121363254-121363254 on Build GRCh38
rs1039000170 Chr.2: 121363285-121363285 on Build GRCh38
rs775611138 Chr.2: 121363213-121363213 on Build GRCh38
rs773534307 Chr.2: 121363036-121363036 on Build GRCh38
rs780740078 Chr.2: 121363153-121363153 on Build GRCh38
rs777133463 Chr.2: 121363223-121363223 on Build GRCh38
rs778833854 Chr.2: 121363342-121363342 on Build GRCh38
rs758118874 Chr.2: 121363309-121363310 on Build GRCh38
rs567506326 Chr.2: 121362982-121362982 on Build GRCh38
rs1017682501 Chr.2: 121363026-121363026 on Build GRCh38
rs750843630 Chr.2: 121363244-121363244 on Build GRCh38
rs370486612 Chr.2: 121363136-121363136 on Build GRCh38
rs145127425 Chr.2: 121363291-121363291 on Build GRCh38
rs763356841 Chr.2: 121363077-121363077 on Build GRCh38
rs900056383 Chr.2: 121362999-121362999 on Build GRCh38
rs1003353624 Chr.2: 121363083-121363083 on Build GRCh38
rs767005164 Chr.2: 121363266-121363266 on Build GRCh38
rs765577807 Chr.2: 121363243-121363243 on Build GRCh38
rs374987543 Chr.2: 121363156-121363156 on Build GRCh38
rs1041715903 Chr.2: 121362995-121362995 on Build GRCh38
rs548485147 Chr.2: 121362962-121362962 on Build GRCh38
rs757326320 Chr.2: 121363310-121363310 on Build GRCh38
rs768711928 Chr.2: 121362928-121362928 on Build GRCh38
rs746077657 Chr.2: 121363184-121363184 on Build GRCh38
rs534901294 Chr.2: 121363013-121363013 on Build GRCh38
rs777595356 Chr.2: 121363164-121363164 on Build GRCh38
rs373992151 Chr.2: 121363182-121363182 on Build GRCh38
rs769153880 Chr.2: 121363215-121363215 on Build GRCh38
rs779811270 Chr.2: 121363319-121363319 on Build GRCh38
rs760703210 Chr.2: 121363214-121363214 on Build GRCh38
rs770853916 Chr.2: 121363179-121363179 on Build GRCh38
rs754691427 Chr.2: 121363138-121363138 on Build GRCh38
rs35300231 Chr.2: 121363103-121363103 on Build GRCh38
rs949246440 Chr.2: 121362955-121362955 on Build GRCh38
rs753822900 Chr.2: 121363305-121363305 on Build GRCh38
rs772236411 Chr.2: 121363190-121363190 on Build GRCh38
rs1054918485 Chr.2: 121362931-121362931 on Build GRCh38
rs182286427 Chr.2: 121363135-121363135 on Build GRCh38
rs893720863 Chr.2: 121363270-121363270 on Build GRCh38
rs770314496 Chr.2: 121363016-121363016 on Build GRCh38
rs752180868 Chr.2: 121363344-121363344 on Build GRCh38
rs141331716 Chr.2: 121363226-121363226 on Build GRCh38
rs1004292927 Chr.2: 121363034-121363034 on Build GRCh38
rs530685813 Chr.2: 121362957-121362957 on Build GRCh38
rs906346762 Chr.2: 121363081-121363081 on Build GRCh38
rs916333304 Chr.2: 121362932-121362932 on Build GRCh38

View Full Product Details