Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201182454-201183042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
UniGene
Hs.5353
Gene Chromosome Location
Chr.2: 201182881-201229406 on Build GRCh38
Transcripts
NM_001206524.1 NM_001206542.1 NM_001230.4 NM_001306083.1 NM_032974.4 NM_032976.3 NM_032977.3 XM_006712796.3

SNP Detail

SNP ID Chromosome Location
rs923875521 Chr.2: 201182656-201182656 on Build GRCh38
rs1036555972 Chr.2: 201182731-201182731 on Build GRCh38
rs535021474 Chr.2: 201182537-201182537 on Build GRCh38
rs934468717 Chr.2: 201182786-201182786 on Build GRCh38
rs373304324 Chr.2: 201182496-201182496 on Build GRCh38
rs998883600 Chr.2: 201182805-201182805 on Build GRCh38
rs958852777 Chr.2: 201182595-201182595 on Build GRCh38
rs41429849 Chr.2: 201182956-201182956 on Build GRCh38
rs899826702 Chr.2: 201182918-201182918 on Build GRCh38
rs377330874 Chr.2: 201182642-201182642 on Build GRCh38
rs1036461443 Chr.2: 201182584-201182584 on Build GRCh38
rs867056381 Chr.2: 201182733-201182733 on Build GRCh38
rs930805181 Chr.2: 201182765-201182765 on Build GRCh38
rs971021903 Chr.2: 201182652-201182652 on Build GRCh38
rs989275741 Chr.2: 201182965-201182965 on Build GRCh38
rs998437972 Chr.2: 201182636-201182636 on Build GRCh38
rs41440544 Chr.2: 201182798-201182798 on Build GRCh38
rs1049259215 Chr.2: 201182775-201182775 on Build GRCh38
rs1009161729 Chr.2: 201182581-201182581 on Build GRCh38
rs776724879 Chr.2: 201182658-201182658 on Build GRCh38
rs557540629 Chr.2: 201182628-201182628 on Build GRCh38
rs1039706066 Chr.2: 201182605-201182605 on Build GRCh38
rs536704741 Chr.2: 201182667-201182667 on Build GRCh38
rs898111867 Chr.2: 201182756-201182756 on Build GRCh38
rs887330075 Chr.2: 201183019-201183019 on Build GRCh38
rs765256703 Chr.2: 201182688-201182688 on Build GRCh38
rs935042882 Chr.2: 201182511-201182511 on Build GRCh38
rs539027305 Chr.2: 201182624-201182624 on Build GRCh38
rs769070649 Chr.2: 201182653-201182653 on Build GRCh38
rs914970535 Chr.2: 201182687-201182687 on Build GRCh38
rs1040202935 Chr.2: 201182817-201182817 on Build GRCh38
rs559580881 Chr.2: 201182936-201182936 on Build GRCh38
rs41525953 Chr.2: 201182638-201182638 on Build GRCh38
rs147431455 Chr.2: 201182726-201182726 on Build GRCh38
rs991201317 Chr.2: 201182712-201182712 on Build GRCh38
rs1052502522 Chr.2: 201182569-201182569 on Build GRCh38
rs956511900 Chr.2: 201182664-201182664 on Build GRCh38
rs763491325 Chr.2: 201182807-201182807 on Build GRCh38
rs1030364350 Chr.2: 201182644-201182644 on Build GRCh38
rs867548103 Chr.2: 201182693-201182693 on Build GRCh38
rs562148669 Chr.2: 201182674-201182674 on Build GRCh38
rs62191478 Chr.2: 201182560-201182560 on Build GRCh38
rs887263639 Chr.2: 201182646-201182646 on Build GRCh38
rs994991872 Chr.2: 201182590-201182590 on Build GRCh38
rs1053512995 Chr.2: 201182518-201182518 on Build GRCh38
rs577258169 Chr.2: 201182952-201182952 on Build GRCh38
rs547241685 Chr.2: 201182592-201182592 on Build GRCh38
rs894517223 Chr.2: 201182777-201182777 on Build GRCh38
rs371809125 Chr.2: 201182505-201182505 on Build GRCh38
rs41422644 Chr.2: 201182527-201182527 on Build GRCh38
rs748888342 Chr.2: 201182689-201182689 on Build GRCh38
rs934006847 Chr.2: 201182953-201182953 on Build GRCh38
rs573753670 Chr.2: 201182787-201182787 on Build GRCh38
rs946797605 Chr.2: 201182602-201182602 on Build GRCh38
rs1022139540 Chr.2: 201183007-201183007 on Build GRCh38
rs898016962 Chr.2: 201182586-201182586 on Build GRCh38
rs1013015787 Chr.2: 201182785-201182785 on Build GRCh38
rs759395240 Chr.2: 201182559-201182559 on Build GRCh38
rs571587910 Chr.2: 201182593-201182593 on Build GRCh38
rs71424285 Chr.2: 201182745-201182745 on Build GRCh38
rs924548149 Chr.2: 201182755-201182755 on Build GRCh38
rs1022595997 Chr.2: 201182719-201182719 on Build GRCh38
rs890665877 Chr.2: 201182577-201182577 on Build GRCh38
rs899836174 Chr.2: 201182626-201182626 on Build GRCh38
rs371263182 Chr.2: 201182736-201182736 on Build GRCh38

View Full Product Details