Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51998279-51998705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf166
Entrez Gene ID
51637
Gene Name
chromosome 14 open reading frame 166
Gene Aliases
CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1
UniGene
Hs.534457
Gene Chromosome Location
Chr.14: 51989510-52004702 on Build GRCh38
Transcripts
NM_016039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246481 c.301G>C p.D101H Chr.14: 51998508-51998508 on Build GRCh38
333949 c.307G>T p.V103L Chr.14: 51998514-51998514 on Build GRCh38
4430942 c.314A>G p.D105G Chr.14: 51998521-51998521 on Build GRCh38
1629336 c.340A>G p.T114A Chr.14: 51998547-51998547 on Build GRCh38
6404398 c.326C>A p.T109N Chr.14: 51998533-51998533 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969152772 Chr.14: 51998557-51998557 on Build GRCh38
rs763112081 Chr.14: 51998564-51998564 on Build GRCh38
rs369423657 Chr.14: 51998611-51998611 on Build GRCh38
rs762979293 Chr.14: 51998598-51998598 on Build GRCh38
rs555138778 Chr.14: 51998317-51998317 on Build GRCh38
rs753508006 Chr.14: 51998621-51998621 on Build GRCh38
rs543865285 Chr.14: 51998409-51998409 on Build GRCh38
rs1012806206 Chr.14: 51998633-51998633 on Build GRCh38
rs765023268 Chr.14: 51998372-51998372 on Build GRCh38
rs192321991 Chr.14: 51998546-51998546 on Build GRCh38
rs762155548 Chr.14: 51998489-51998489 on Build GRCh38
rs114700982 Chr.14: 51998368-51998368 on Build GRCh38
rs35006177 Chr.14: 51998537-51998537 on Build GRCh38
rs982969622 Chr.14: 51998650-51998650 on Build GRCh38
rs373333013 Chr.14: 51998610-51998610 on Build GRCh38
rs780290809 Chr.14: 51998404-51998404 on Build GRCh38
rs748865536 Chr.14: 51998548-51998548 on Build GRCh38
rs376608717 Chr.14: 51998456-51998456 on Build GRCh38
rs765487249 Chr.14: 51998501-51998501 on Build GRCh38
rs745690303 Chr.14: 51998461-51998461 on Build GRCh38
rs1037876936 Chr.14: 51998402-51998402 on Build GRCh38
rs763428379 Chr.14: 51998602-51998602 on Build GRCh38
rs34968560 Chr.14: 51998360-51998360 on Build GRCh38
rs773436638 Chr.14: 51998511-51998511 on Build GRCh38
rs375943562 Chr.14: 51998588-51998588 on Build GRCh38
rs149288575 Chr.14: 51998518-51998518 on Build GRCh38
rs370052868 Chr.14: 51998608-51998608 on Build GRCh38
rs777272455 Chr.14: 51998450-51998450 on Build GRCh38
rs750904253 Chr.14: 51998629-51998631 on Build GRCh38
rs763776841 Chr.14: 51998615-51998615 on Build GRCh38
rs1018815314 Chr.14: 51998577-51998577 on Build GRCh38
rs777392887 Chr.14: 51998547-51998547 on Build GRCh38
rs778765729 Chr.14: 51998570-51998570 on Build GRCh38
rs766858321 Chr.14: 51998519-51998519 on Build GRCh38
rs760096002 Chr.14: 51998609-51998609 on Build GRCh38
rs771864500 Chr.14: 51998572-51998572 on Build GRCh38
rs745839804 Chr.14: 51998571-51998571 on Build GRCh38
rs1020309824 Chr.14: 51998549-51998549 on Build GRCh38
rs34455622 Chr.14: 51998379-51998380 on Build GRCh38
rs971261480 Chr.14: 51998336-51998336 on Build GRCh38
rs775428230 Chr.14: 51998578-51998578 on Build GRCh38
rs373814435 Chr.14: 51998613-51998613 on Build GRCh38
rs143561618 Chr.14: 51998520-51998520 on Build GRCh38
rs112285188 Chr.14: 51998484-51998484 on Build GRCh38
rs771877315 Chr.14: 51998466-51998466 on Build GRCh38
rs930195081 Chr.14: 51998337-51998337 on Build GRCh38
rs143904107 Chr.14: 51998581-51998581 on Build GRCh38
rs756809153 Chr.14: 51998631-51998631 on Build GRCh38
rs1693189 Chr.14: 51998531-51998531 on Build GRCh38
rs981925071 Chr.14: 51998338-51998338 on Build GRCh38
rs763561229 Chr.14: 51998633-51998633 on Build GRCh38
rs771068057 Chr.14: 51998601-51998601 on Build GRCh38
rs770834114 Chr.14: 51998459-51998459 on Build GRCh38
rs561626901 Chr.14: 51998542-51998542 on Build GRCh38
rs371686137 Chr.14: 51998460-51998460 on Build GRCh38

View Full Product Details