Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51697985-51698568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
UniGene
Hs.434914
Gene Chromosome Location
Chr.14: 51489100-51730727 on Build GRCh38
Transcripts
NM_001042481.2 NM_001267046.1 NM_152330.3 XM_006720029.1 XM_006720030.1 XM_011536423.1 XM_011536424.1 XM_017020967.1 XM_017020968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7242785 c.147C>G p.L49L Chr.14: 51698189-51698189 on Build GRCh38
6294515 c.150G>T p.R50S Chr.14: 51698192-51698192 on Build GRCh38
1252792 c.178A>G p.S60G Chr.14: 51698220-51698220 on Build GRCh38
7242784 c.147C>G p.L49L Chr.14: 51698189-51698189 on Build GRCh38
6258507 c.144G>T p.L48L Chr.14: 51698186-51698186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193210556 Chr.14: 51698170-51698170 on Build GRCh38
rs928913213 Chr.14: 51698549-51698549 on Build GRCh38
rs527606277 Chr.14: 51698394-51698394 on Build GRCh38
rs934242642 Chr.14: 51698015-51698015 on Build GRCh38
rs765692816 Chr.14: 51698094-51698094 on Build GRCh38
rs985693234 Chr.14: 51698042-51698042 on Build GRCh38
rs569321477 Chr.14: 51698088-51698088 on Build GRCh38
rs756018793 Chr.14: 51698246-51698246 on Build GRCh38
rs548099819 Chr.14: 51698548-51698548 on Build GRCh38
rs886882592 Chr.14: 51698384-51698384 on Build GRCh38
rs765958718 Chr.14: 51698192-51698192 on Build GRCh38
rs561497530 Chr.14: 51698059-51698059 on Build GRCh38
rs752277577 Chr.14: 51698253-51698255 on Build GRCh38
rs1029957332 Chr.14: 51698413-51698413 on Build GRCh38
rs763360286 Chr.14: 51698191-51698191 on Build GRCh38
rs780992849 Chr.14: 51698121-51698121 on Build GRCh38
rs911456834 Chr.14: 51698479-51698479 on Build GRCh38
rs776988920 Chr.14: 51698499-51698499 on Build GRCh38
rs777072709 Chr.14: 51698175-51698175 on Build GRCh38
rs750148111 Chr.14: 51698101-51698101 on Build GRCh38
rs533160244 Chr.14: 51698424-51698424 on Build GRCh38
rs114289355 Chr.14: 51698269-51698269 on Build GRCh38
rs561695942 Chr.14: 51698388-51698388 on Build GRCh38
rs375152381 Chr.14: 51698204-51698204 on Build GRCh38
rs986853380 Chr.14: 51698250-51698250 on Build GRCh38
rs377502742 Chr.14: 51698283-51698283 on Build GRCh38
rs546740554 Chr.14: 51698524-51698524 on Build GRCh38
rs115992811 Chr.14: 51698461-51698461 on Build GRCh38
rs111333361 Chr.14: 51698378-51698378 on Build GRCh38
rs139287449 Chr.14: 51698352-51698355 on Build GRCh38
rs1045144260 Chr.14: 51698026-51698026 on Build GRCh38
rs148502201 Chr.14: 51698421-51698421 on Build GRCh38
rs911389628 Chr.14: 51698052-51698052 on Build GRCh38
rs955333351 Chr.14: 51698239-51698239 on Build GRCh38
rs142526761 Chr.14: 51698169-51698169 on Build GRCh38
rs375692856 Chr.14: 51698123-51698123 on Build GRCh38
rs767037450 Chr.14: 51698223-51698223 on Build GRCh38
rs541860482 Chr.14: 51698055-51698055 on Build GRCh38
rs926285534 Chr.14: 51698369-51698369 on Build GRCh38
rs533393308 Chr.14: 51698447-51698447 on Build GRCh38
rs896447283 Chr.14: 51698247-51698247 on Build GRCh38
rs74713569 Chr.14: 51698060-51698060 on Build GRCh38
rs202093591 Chr.14: 51698120-51698120 on Build GRCh38
rs534480876 Chr.14: 51698184-51698184 on Build GRCh38
rs530504352 Chr.14: 51698067-51698067 on Build GRCh38
rs766087637 Chr.14: 51698110-51698110 on Build GRCh38
rs752628637 Chr.14: 51698227-51698227 on Build GRCh38
rs777525624 Chr.14: 51698128-51698128 on Build GRCh38
rs1028161584 Chr.14: 51698420-51698420 on Build GRCh38
rs188166227 Chr.14: 51698280-51698280 on Build GRCh38
rs183842555 Chr.14: 51698274-51698274 on Build GRCh38
rs6572784 Chr.14: 51698365-51698365 on Build GRCh38
rs751051671 Chr.14: 51698118-51698118 on Build GRCh38
rs746132408 Chr.14: 51698138-51698138 on Build GRCh38
rs980671346 Chr.14: 51698544-51698544 on Build GRCh38
rs142888574 Chr.14: 51698495-51698495 on Build GRCh38
rs1008221348 Chr.14: 51698450-51698450 on Build GRCh38
rs527680165 Chr.14: 51698022-51698023 on Build GRCh38
rs1031614482 Chr.14: 51698452-51698452 on Build GRCh38
rs758093335 Chr.14: 51698103-51698103 on Build GRCh38
rs6572785 Chr.14: 51698408-51698408 on Build GRCh38
rs939770967 Chr.14: 51698389-51698390 on Build GRCh38
rs918712984 Chr.14: 51698535-51698535 on Build GRCh38
rs748108678 Chr.14: 51698122-51698122 on Build GRCh38
rs562232035 Chr.14: 51698299-51698299 on Build GRCh38
rs773741489 Chr.14: 51698190-51698190 on Build GRCh38
rs760678358 Chr.14: 51698376-51698376 on Build GRCh38
rs370580833 Chr.14: 51698165-51698165 on Build GRCh38
rs956449459 Chr.14: 51698396-51698396 on Build GRCh38
rs1056194755 Chr.14: 51698077-51698077 on Build GRCh38
rs150568838 Chr.14: 51698198-51698198 on Build GRCh38
rs180768518 Chr.14: 51698372-51698372 on Build GRCh38
rs914718564 Chr.14: 51698314-51698314 on Build GRCh38

View Full Product Details